Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 133274710-133274965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYA4
Entrez Gene ID
2070
Gene Name
EYA transcriptional coactivator and phosphatase 4
Gene Aliases
CMD1J, DFNA10
UniGene
Hs.596680
Gene Chromosome Location
Chr.6: 133240340-133532120 on Build GRCh38
Transcripts
NM_001301012.1 NM_001301013.1 NM_004100.4 NM_172103.3 NM_172105.3 XM_005266851.4 XM_005266853.4 XM_017010368.1 XM_017010369.1 XM_017010370.1 XM_017010371.1 XM_017010372.1 XM_017010373.1 XM_017010374.1 XM_017010375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
228090 c.16G>A p.D6N Chr.6: 133274796-133274796 on Build GRCh38
3174039 c.4G>A p.E2K Chr.6: 133274784-133274784 on Build GRCh38
6293548 c.14A>T p.Q5L Chr.6: 133274794-133274794 on Build GRCh38
4530454 c.16G>A p.D6N Chr.6: 133274796-133274796 on Build GRCh38
3174040 c.15G>A p.Q5Q Chr.6: 133274795-133274795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759852514 Chr.6: 133274757-133274757 on Build GRCh38
rs374322196 Chr.6: 133274789-133274789 on Build GRCh38
rs776128099 Chr.6: 133274838-133274838 on Build GRCh38
rs753899876 Chr.6: 133274844-133274844 on Build GRCh38
rs76139839 Chr.6: 133274840-133274840 on Build GRCh38
rs765607552 Chr.6: 133274849-133274849 on Build GRCh38
rs761938713 Chr.6: 133274847-133274847 on Build GRCh38
rs911536575 Chr.6: 133274886-133274886 on Build GRCh38
rs753119255 Chr.6: 133274766-133274766 on Build GRCh38
rs760845191 Chr.6: 133274767-133274767 on Build GRCh38
rs926133339 Chr.6: 133274922-133274922 on Build GRCh38
rs751952960 Chr.6: 133274837-133274860 on Build GRCh38
rs965754514 Chr.6: 133274888-133274888 on Build GRCh38
rs913488990 Chr.6: 133274896-133274896 on Build GRCh38
rs953313876 Chr.6: 133274783-133274783 on Build GRCh38
rs375532786 Chr.6: 133274873-133274873 on Build GRCh38
rs183259468 Chr.6: 133274874-133274874 on Build GRCh38
rs746916198 Chr.6: 133274786-133274786 on Build GRCh38
rs772801278 Chr.6: 133274823-133274823 on Build GRCh38
rs986089427 Chr.6: 133274788-133274788 on Build GRCh38
rs753901240 Chr.6: 133274769-133274769 on Build GRCh38
rs200947384 Chr.6: 133274764-133274764 on Build GRCh38
rs977385813 Chr.6: 133274890-133274890 on Build GRCh38
rs3734278 Chr.6: 133274925-133274925 on Build GRCh38
rs772392856 Chr.6: 133274832-133274832 on Build GRCh38
rs774700638 Chr.6: 133274754-133274754 on Build GRCh38
rs566465018 Chr.6: 133274913-133274913 on Build GRCh38
rs191747564 Chr.6: 133274850-133274850 on Build GRCh38
rs747628106 Chr.6: 133274798-133274798 on Build GRCh38
rs371119361 Chr.6: 133274742-133274742 on Build GRCh38
rs780580161 Chr.6: 133274794-133274794 on Build GRCh38
rs750268684 Chr.6: 133274779-133274779 on Build GRCh38
rs779238999 Chr.6: 133274777-133274777 on Build GRCh38
rs759940308 Chr.6: 133274825-133274825 on Build GRCh38
rs780081930 Chr.6: 133274785-133274785 on Build GRCh38
rs200593099 Chr.6: 133274782-133274782 on Build GRCh38
rs531917929 Chr.6: 133274768-133274768 on Build GRCh38
rs764598296 Chr.6: 133274843-133274843 on Build GRCh38
rs370459996 Chr.6: 133274857-133274857 on Build GRCh38
rs946163861 Chr.6: 133274908-133274908 on Build GRCh38
rs143613363 Chr.6: 133274813-133274813 on Build GRCh38
rs751310321 Chr.6: 133274851-133274851 on Build GRCh38
rs761069787 Chr.6: 133274841-133274841 on Build GRCh38

View Full Product Details