Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154899777-154900050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F8
Entrez Gene ID
2157
Gene Name
coagulation factor VIII
Gene Aliases
AHF, DXS1253E, F8B, F8C, FVIII, HEMA
UniGene
Hs.632836
Gene Chromosome Location
Chr.X: 154835788-155022723 on Build GRCh38
Transcripts
NM_000132.3

SNP Detail

SNP ID Chromosome Location
rs137852455 Chr.X: 154899946-154899946 on Build GRCh38
rs781988865 Chr.X: 154899977-154899977 on Build GRCh38
rs782201175 Chr.X: 154899922-154899922 on Build GRCh38
rs371317608 Chr.X: 154899919-154899919 on Build GRCh38
rs782656088 Chr.X: 154899841-154899841 on Build GRCh38
rs782353868 Chr.X: 154899926-154899926 on Build GRCh38
rs781917172 Chr.X: 154899960-154899960 on Build GRCh38
rs369060351 Chr.X: 154899819-154899819 on Build GRCh38
rs781794885 Chr.X: 154899817-154899817 on Build GRCh38
rs782454730 Chr.X: 154899838-154899838 on Build GRCh38
rs137852456 Chr.X: 154899876-154899876 on Build GRCh38
rs376228821 Chr.X: 154899963-154899963 on Build GRCh38
rs782559263 Chr.X: 154899914-154899914 on Build GRCh38
rs782286433 Chr.X: 154899913-154899913 on Build GRCh38

View Full Product Details