Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5774013-5774272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMP1
Entrez Gene ID
79956
Gene Name
endoplasmic reticulum metallopeptidase 1
Gene Aliases
FXNA, KIAA1815, bA207C16.3
UniGene
Hs.591078
Gene Chromosome Location
Chr.9: 5764061-5867091 on Build GRCh38
Transcripts
NM_001206557.1 NM_020829.3 XM_005251523.3 XM_011517967.2 XM_011517968.2 XM_017014934.1 XM_017014935.1 XM_017014936.1

Gene Symbol
RIC1
Entrez Gene ID
57589
Gene Name
RIC1 homolog, RAB6A GEF complex partner 1
Gene Aliases
CIP150, KIAA1432, bA207C16.1
UniGene
Hs.211520
Gene Chromosome Location
Chr.9: 5629030-5778633 on Build GRCh38
Transcripts
NM_001206557.1 NM_020829.3 XM_005251523.3 XM_011517967.2 XM_011517968.2 XM_017014934.1 XM_017014935.1 XM_017014936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4927013 c.3811G>A p.E1271K Chr.9: 5774022-5774022 on Build GRCh38
1239237 c.4012A>G p.T1338A Chr.9: 5774223-5774223 on Build GRCh38
6694067 c.3876C>T p.S1292S Chr.9: 5774087-5774087 on Build GRCh38
462392 c.3814G>C p.E1272Q Chr.9: 5774025-5774025 on Build GRCh38
6293732 c.3951G>T p.M1317I Chr.9: 5774162-5774162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775573960 Chr.9: 5774229-5774229 on Build GRCh38
rs778873124 Chr.9: 5774158-5774158 on Build GRCh38
rs878936538 Chr.9: 5774111-5774111 on Build GRCh38
rs372491933 Chr.9: 5774186-5774186 on Build GRCh38
rs1056048451 Chr.9: 5774062-5774062 on Build GRCh38
rs757865916 Chr.9: 5774165-5774165 on Build GRCh38
rs3739647 Chr.9: 5774159-5774159 on Build GRCh38
rs757410696 Chr.9: 5774218-5774218 on Build GRCh38
rs751215810 Chr.9: 5774195-5774203 on Build GRCh38
rs552633335 Chr.9: 5774052-5774052 on Build GRCh38
rs145856254 Chr.9: 5774146-5774146 on Build GRCh38
rs141047465 Chr.9: 5774075-5774075 on Build GRCh38
rs776553031 Chr.9: 5774161-5774161 on Build GRCh38
rs770868925 Chr.9: 5774092-5774092 on Build GRCh38
rs1015903622 Chr.9: 5774087-5774087 on Build GRCh38
rs748026732 Chr.9: 5774209-5774209 on Build GRCh38
rs745627904 Chr.9: 5774228-5774228 on Build GRCh38
rs764450150 Chr.9: 5774134-5774134 on Build GRCh38
rs759931673 Chr.9: 5774109-5774109 on Build GRCh38
rs780772814 Chr.9: 5774055-5774055 on Build GRCh38
rs774562859 Chr.9: 5774247-5774247 on Build GRCh38
rs571170082 Chr.9: 5774061-5774061 on Build GRCh38
rs746594252 Chr.9: 5774170-5774170 on Build GRCh38
rs763586601 Chr.9: 5774175-5774177 on Build GRCh38
rs780205253 Chr.9: 5774150-5774150 on Build GRCh38
rs769197957 Chr.9: 5774245-5774245 on Build GRCh38
rs777464077 Chr.9: 5774036-5774036 on Build GRCh38
rs760965880 Chr.9: 5774091-5774091 on Build GRCh38
rs913175539 Chr.9: 5774096-5774096 on Build GRCh38
rs762303239 Chr.9: 5774253-5774253 on Build GRCh38
rs554530071 Chr.9: 5774204-5774204 on Build GRCh38
rs375146459 Chr.9: 5774073-5774073 on Build GRCh38
rs751159023 Chr.9: 5774140-5774140 on Build GRCh38
rs768512952 Chr.9: 5774156-5774156 on Build GRCh38
rs974353752 Chr.9: 5774044-5774044 on Build GRCh38
rs751958383 Chr.9: 5774177-5774177 on Build GRCh38
rs763379165 Chr.9: 5774132-5774132 on Build GRCh38
rs749895946 Chr.9: 5774180-5774180 on Build GRCh38
rs949448093 Chr.9: 5774192-5774192 on Build GRCh38
rs763468870 Chr.9: 5774175-5774175 on Build GRCh38
rs753779392 Chr.9: 5774189-5774189 on Build GRCh38
rs369951068 Chr.9: 5774154-5774154 on Build GRCh38
rs776805324 Chr.9: 5774102-5774102 on Build GRCh38
rs376097440 Chr.9: 5774208-5774208 on Build GRCh38
rs767256306 Chr.9: 5774178-5774178 on Build GRCh38
rs756659856 Chr.9: 5774050-5774050 on Build GRCh38
rs1018206273 Chr.9: 5774163-5774163 on Build GRCh38
rs775836744 Chr.9: 5774087-5774087 on Build GRCh38
rs779971821 Chr.9: 5774074-5774074 on Build GRCh38
rs376378176 Chr.9: 5774169-5774169 on Build GRCh38
rs772418488 Chr.9: 5774078-5774078 on Build GRCh38
rs754997905 Chr.9: 5774201-5774201 on Build GRCh38
rs897528541 Chr.9: 5774088-5774088 on Build GRCh38
rs1002740101 Chr.9: 5774060-5774060 on Build GRCh38
rs149844862 Chr.9: 5774080-5774080 on Build GRCh38
rs527983176 Chr.9: 5774045-5774045 on Build GRCh38
rs753050335 Chr.9: 5774122-5774122 on Build GRCh38
rs762578948 Chr.9: 5774110-5774111 on Build GRCh38
rs113236827 Chr.9: 5774089-5774089 on Build GRCh38
rs3739648 Chr.9: 5774225-5774225 on Build GRCh38
rs765824248 Chr.9: 5774121-5774121 on Build GRCh38
rs756961310 Chr.9: 5774144-5774144 on Build GRCh38
rs771225515 Chr.9: 5774160-5774160 on Build GRCh38
rs369200435 Chr.9: 5774181-5774181 on Build GRCh38
rs550417804 Chr.9: 5774149-5774149 on Build GRCh38
rs534087193 Chr.9: 5774240-5774240 on Build GRCh38
rs913904754 Chr.9: 5774190-5774190 on Build GRCh38
rs778493235 Chr.9: 5774043-5774043 on Build GRCh38
rs896482257 Chr.9: 5774191-5774191 on Build GRCh38
rs770980698 Chr.9: 5774196-5774196 on Build GRCh38

View Full Product Details