Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 55267658-55267929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLF
Entrez Gene ID
3131
Gene Name
HLF, PAR bZIP transcription factor
Gene Aliases
-
UniGene
Hs.196952
Gene Chromosome Location
Chr.17: 55264960-55325065 on Build GRCh38
Transcripts
NM_002126.4 XM_005257269.2 XM_011524705.1 XM_017024556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3194091 c.161G>C p.S54T Chr.17: 55267796-55267796 on Build GRCh38
281721 c.231C>T p.D77D Chr.17: 55267866-55267866 on Build GRCh38
6439273 c.157G>A p.E53K Chr.17: 55267792-55267792 on Build GRCh38
5853023 c.197G>A p.G66E Chr.17: 55267832-55267832 on Build GRCh38
3717482 c.150G>A p.L50L Chr.17: 55267785-55267785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761427708 Chr.17: 55267808-55267808 on Build GRCh38
rs762403571 Chr.17: 55267715-55267716 on Build GRCh38
rs201209755 Chr.17: 55267739-55267739 on Build GRCh38
rs778071353 Chr.17: 55267872-55267872 on Build GRCh38
rs183911087 Chr.17: 55267855-55267855 on Build GRCh38
rs945568063 Chr.17: 55267768-55267768 on Build GRCh38
rs777681481 Chr.17: 55267711-55267711 on Build GRCh38
rs755022700 Chr.17: 55267854-55267854 on Build GRCh38
rs924364210 Chr.17: 55267694-55267694 on Build GRCh38
rs765631500 Chr.17: 55267716-55267720 on Build GRCh38
rs200291249 Chr.17: 55267893-55267893 on Build GRCh38
rs553666718 Chr.17: 55267730-55267730 on Build GRCh38
rs776962498 Chr.17: 55267716-55267716 on Build GRCh38
rs563496609 Chr.17: 55267745-55267745 on Build GRCh38
rs760106013 Chr.17: 55267797-55267797 on Build GRCh38
rs749260477 Chr.17: 55267878-55267878 on Build GRCh38
rs760757135 Chr.17: 55267833-55267833 on Build GRCh38
rs916151819 Chr.17: 55267798-55267798 on Build GRCh38
rs143697299 Chr.17: 55267809-55267809 on Build GRCh38
rs751538271 Chr.17: 55267852-55267852 on Build GRCh38
rs769173900 Chr.17: 55267702-55267706 on Build GRCh38
rs181297521 Chr.17: 55267823-55267823 on Build GRCh38
rs368847041 Chr.17: 55267779-55267779 on Build GRCh38
rs749851297 Chr.17: 55267821-55267821 on Build GRCh38
rs935656428 Chr.17: 55267683-55267683 on Build GRCh38
rs2302739 Chr.17: 55267690-55267690 on Build GRCh38
rs767704148 Chr.17: 55267766-55267766 on Build GRCh38
rs775884594 Chr.17: 55267805-55267805 on Build GRCh38
rs766159669 Chr.17: 55267842-55267842 on Build GRCh38
rs906658784 Chr.17: 55267887-55267887 on Build GRCh38
rs772417385 Chr.17: 55267804-55267804 on Build GRCh38
rs752578837 Chr.17: 55267859-55267859 on Build GRCh38
rs771286163 Chr.17: 55267752-55267752 on Build GRCh38
rs577401506 Chr.17: 55267688-55267688 on Build GRCh38
rs202162317 Chr.17: 55267866-55267866 on Build GRCh38

View Full Product Details