Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44784806-44785075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST4
Entrez Gene ID
266675
Gene Name
bestrophin 4
Gene Aliases
VMD2L2
UniGene
Hs.302513
Gene Chromosome Location
Chr.1: 44781836-44792828 on Build GRCh38
Transcripts
NM_153274.2 XM_017001020.1 XM_017001021.1 XM_017001022.1 XM_017001023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4187044 c.970C>T p.Q324* Chr.1: 44784928-44784928 on Build GRCh38
3369782 c.992A>G p.Q331R Chr.1: 44784906-44784906 on Build GRCh38
2171830 c.946G>T p.D316Y Chr.1: 44784952-44784952 on Build GRCh38
4423667 c.954C>T p.D318D Chr.1: 44784944-44784944 on Build GRCh38
1667522 c.983G>A p.R328H Chr.1: 44784915-44784915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762198297 Chr.1: 44784883-44784883 on Build GRCh38
rs1036635036 Chr.1: 44785025-44785025 on Build GRCh38
rs778549996 Chr.1: 44784916-44784916 on Build GRCh38
rs372498770 Chr.1: 44784912-44784912 on Build GRCh38
rs777525386 Chr.1: 44784989-44784989 on Build GRCh38
rs369629759 Chr.1: 44785037-44785037 on Build GRCh38
rs773705288 Chr.1: 44784858-44784858 on Build GRCh38
rs150797829 Chr.1: 44784923-44784923 on Build GRCh38
rs772916692 Chr.1: 44784831-44784831 on Build GRCh38
rs769208659 Chr.1: 44785030-44785030 on Build GRCh38
rs894204398 Chr.1: 44784846-44784846 on Build GRCh38
rs748074273 Chr.1: 44784931-44784931 on Build GRCh38
rs762584742 Chr.1: 44784832-44784832 on Build GRCh38
rs773936755 Chr.1: 44785010-44785010 on Build GRCh38
rs758671494 Chr.1: 44784889-44784889 on Build GRCh38
rs780927449 Chr.1: 44784916-44784916 on Build GRCh38
rs750730789 Chr.1: 44784887-44784887 on Build GRCh38
rs769636382 Chr.1: 44784948-44784948 on Build GRCh38
rs780220883 Chr.1: 44784896-44784896 on Build GRCh38
rs1012656124 Chr.1: 44784877-44784877 on Build GRCh38
rs369252209 Chr.1: 44784886-44784886 on Build GRCh38
rs201461479 Chr.1: 44784862-44784862 on Build GRCh38
rs16832241 Chr.1: 44784907-44784907 on Build GRCh38
rs937440816 Chr.1: 44784904-44784904 on Build GRCh38
rs201994250 Chr.1: 44784906-44784906 on Build GRCh38
rs377207881 Chr.1: 44784921-44784921 on Build GRCh38
rs373156176 Chr.1: 44785002-44785002 on Build GRCh38
rs765967549 Chr.1: 44784835-44784835 on Build GRCh38
rs758908781 Chr.1: 44785015-44785015 on Build GRCh38
rs375787125 Chr.1: 44784865-44784865 on Build GRCh38
rs369961513 Chr.1: 44785045-44785045 on Build GRCh38
rs375771977 Chr.1: 44785046-44785046 on Build GRCh38
rs369088764 Chr.1: 44784944-44784944 on Build GRCh38
rs745436002 Chr.1: 44785031-44785031 on Build GRCh38
rs754241463 Chr.1: 44784881-44784881 on Build GRCh38

View Full Product Details