Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43774599-43774824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTB
Entrez Gene ID
1476
Gene Name
cystatin B
Gene Aliases
CPI-B, CST6, EPM1, EPM1A, PME, STFB, ULD
UniGene
Hs.695
Gene Chromosome Location
Chr.21: 43773665-43776375 on Build GRCh38
Transcripts
NM_000100.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4469948 c.162C>T p.F54F Chr.21: 43774664-43774664 on Build GRCh38
5898625 c.67-4C>T p.? Chr.21: 43774763-43774763 on Build GRCh38
7363110 c.121G>C p.V41L Chr.21: 43774705-43774705 on Build GRCh38
4675225 c.119C>G p.A40G Chr.21: 43774707-43774707 on Build GRCh38
7264217 c.72G>A p.R24R Chr.21: 43774754-43774754 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs878971833 Chr.21: 43774654-43774654 on Build GRCh38
rs764244824 Chr.21: 43774618-43774618 on Build GRCh38
rs140799752 Chr.21: 43774687-43774687 on Build GRCh38
rs745589113 Chr.21: 43774639-43774639 on Build GRCh38
rs312262707 Chr.21: 43774640-43774657 on Build GRCh38
rs879246734 Chr.21: 43774652-43774652 on Build GRCh38
rs760098381 Chr.21: 43774773-43774773 on Build GRCh38
rs370404096 Chr.21: 43774626-43774626 on Build GRCh38
rs201046688 Chr.21: 43774629-43774629 on Build GRCh38
rs772315538 Chr.21: 43774753-43774753 on Build GRCh38
rs377595258 Chr.21: 43774794-43774794 on Build GRCh38
rs375008755 Chr.21: 43774691-43774691 on Build GRCh38
rs754421704 Chr.21: 43774680-43774680 on Build GRCh38
rs750620672 Chr.21: 43774728-43774730 on Build GRCh38
rs374092254 Chr.21: 43774785-43774785 on Build GRCh38
rs369957957 Chr.21: 43774623-43774623 on Build GRCh38
rs745678958 Chr.21: 43774689-43774689 on Build GRCh38
rs312262708 Chr.21: 43774677-43774677 on Build GRCh38
rs767709881 Chr.21: 43774636-43774653 on Build GRCh38
rs765593211 Chr.21: 43774780-43774780 on Build GRCh38
rs748162136 Chr.21: 43774720-43774720 on Build GRCh38
rs199873087 Chr.21: 43774769-43774769 on Build GRCh38
rs776841907 Chr.21: 43774771-43774771 on Build GRCh38
rs751856832 Chr.21: 43774621-43774621 on Build GRCh38
rs202139907 Chr.21: 43774627-43774627 on Build GRCh38
rs770690422 Chr.21: 43774634-43774634 on Build GRCh38
rs559906825 Chr.21: 43774681-43774681 on Build GRCh38
rs372787200 Chr.21: 43774676-43774676 on Build GRCh38
rs760102906 Chr.21: 43774708-43774708 on Build GRCh38
rs143153487 Chr.21: 43774705-43774705 on Build GRCh38
rs766654703 Chr.21: 43774679-43774679 on Build GRCh38
rs766829227 Chr.21: 43774765-43774765 on Build GRCh38
rs6382 Chr.21: 43774767-43774767 on Build GRCh38
rs746739128 Chr.21: 43774631-43774631 on Build GRCh38
rs878898846 Chr.21: 43774655-43774655 on Build GRCh38
rs545986367 Chr.21: 43774690-43774690 on Build GRCh38
rs781145714 Chr.21: 43774638-43774638 on Build GRCh38
rs777111664 Chr.21: 43774630-43774630 on Build GRCh38
rs202096395 Chr.21: 43774682-43774682 on Build GRCh38
rs1057518464 Chr.21: 43774659-43774659 on Build GRCh38
rs386833439 Chr.21: 43774701-43774701 on Build GRCh38
rs386833440 Chr.21: 43774658-43774658 on Build GRCh38
rs368198839 Chr.21: 43774706-43774706 on Build GRCh38
rs1045956757 Chr.21: 43774719-43774719 on Build GRCh38
rs200236513 Chr.21: 43774669-43774669 on Build GRCh38
rs774504790 Chr.21: 43774715-43774715 on Build GRCh38
rs768820634 Chr.21: 43774657-43774657 on Build GRCh38
rs556221258 Chr.21: 43774650-43774650 on Build GRCh38
rs774291632 Chr.21: 43774666-43774666 on Build GRCh38
rs541671661 Chr.21: 43774671-43774671 on Build GRCh38
rs775447833 Chr.21: 43774653-43774653 on Build GRCh38
rs762082236 Chr.21: 43774668-43774668 on Build GRCh38
rs761992444 Chr.21: 43774622-43774622 on Build GRCh38
rs762632973 Chr.21: 43774736-43774737 on Build GRCh38
rs864309482 Chr.21: 43774637-43774656 on Build GRCh38
rs749157471 Chr.21: 43774656-43774656 on Build GRCh38
rs6383 Chr.21: 43774762-43774762 on Build GRCh38
rs910966552 Chr.21: 43774645-43774645 on Build GRCh38
rs935774172 Chr.21: 43774659-43774659 on Build GRCh38
rs147484110 Chr.21: 43774760-43774760 on Build GRCh38
rs201959779 Chr.21: 43774782-43774782 on Build GRCh38

View Full Product Details