Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157430378-157430647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTIP
Entrez Gene ID
9595
Gene Name
cytohesin 1 interacting protein
Gene Aliases
B3-1, CASP, CYBR, CYTHIP, HE, PSCDBP
UniGene
Hs.270
Gene Chromosome Location
Chr.2: 157412781-157444092 on Build GRCh38
Transcripts
NM_004288.4 XM_017005386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1691177 c.458C>T p.S153L Chr.2: 157430577-157430577 on Build GRCh38
4085810 c.440T>G p.V147G Chr.2: 157430595-157430595 on Build GRCh38
3299803 c.463G>A p.G155R Chr.2: 157430572-157430572 on Build GRCh38
255451 c.462C>G p.S154S Chr.2: 157430573-157430573 on Build GRCh38
3961331 c.476+1G>T p.? Chr.2: 157430558-157430558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026458051 Chr.2: 157430592-157430592 on Build GRCh38
rs569718078 Chr.2: 157430500-157430500 on Build GRCh38
rs537094302 Chr.2: 157430506-157430506 on Build GRCh38
rs138461242 Chr.2: 157430593-157430593 on Build GRCh38
rs112966992 Chr.2: 157430537-157430537 on Build GRCh38
rs540605984 Chr.2: 157430606-157430606 on Build GRCh38
rs890660596 Chr.2: 157430518-157430519 on Build GRCh38
rs376116326 Chr.2: 157430533-157430533 on Build GRCh38
rs921142058 Chr.2: 157430443-157430443 on Build GRCh38
rs534886532 Chr.2: 157430559-157430559 on Build GRCh38
rs762663690 Chr.2: 157430577-157430577 on Build GRCh38
rs376309452 Chr.2: 157430566-157430566 on Build GRCh38
rs773163736 Chr.2: 157430576-157430576 on Build GRCh38
rs369807419 Chr.2: 157430519-157430519 on Build GRCh38
rs113434180 Chr.2: 157430543-157430543 on Build GRCh38
rs962662460 Chr.2: 157430411-157430411 on Build GRCh38
rs145695913 Chr.2: 157430425-157430425 on Build GRCh38
rs148213749 Chr.2: 157430522-157430522 on Build GRCh38
rs552825547 Chr.2: 157430460-157430460 on Build GRCh38
rs144643138 Chr.2: 157430450-157430450 on Build GRCh38
rs767200421 Chr.2: 157430510-157430510 on Build GRCh38
rs779016880 Chr.2: 157430551-157430551 on Build GRCh38
rs548142718 Chr.2: 157430499-157430499 on Build GRCh38
rs370533583 Chr.2: 157430538-157430538 on Build GRCh38
rs185432543 Chr.2: 157430552-157430552 on Build GRCh38
rs769643425 Chr.2: 157430573-157430573 on Build GRCh38
rs747072083 Chr.2: 157430572-157430572 on Build GRCh38
rs767253791 Chr.2: 157430611-157430611 on Build GRCh38
rs777007288 Chr.2: 157430587-157430587 on Build GRCh38
rs754659685 Chr.2: 157430508-157430508 on Build GRCh38
rs775588319 Chr.2: 157430568-157430568 on Build GRCh38
rs140813062 Chr.2: 157430442-157430442 on Build GRCh38
rs933817317 Chr.2: 157430444-157430444 on Build GRCh38
rs763976944 Chr.2: 157430583-157430585 on Build GRCh38
rs372835396 Chr.2: 157430544-157430544 on Build GRCh38
rs148810128 Chr.2: 157430601-157430601 on Build GRCh38

View Full Product Details