Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7042875-7043122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC96
Entrez Gene ID
257236
Gene Name
coiled-coil domain containing 96
Gene Aliases
-
UniGene
Hs.656757
Gene Chromosome Location
Chr.4: 7040849-7043001 on Build GRCh38
Transcripts
NM_153376.2 NR_033828.1

Gene Symbol
LOC100129931
Entrez Gene ID
100129931
Gene Name
uncharacterized LOC100129931
Gene Aliases
-
UniGene
Hs.707753
Gene Chromosome Location
Chr.4: 7030554-7046231 on Build GRCh38
Transcripts
NM_153376.2 NR_033828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622713 c.58A>C p.S20R Chr.4: 7042881-7042881 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768424517 Chr.4: 7042976-7042976 on Build GRCh38
rs893642487 Chr.4: 7043045-7043045 on Build GRCh38
rs750735210 Chr.4: 7042902-7042902 on Build GRCh38
rs935937358 Chr.4: 7042995-7042995 on Build GRCh38
rs535061373 Chr.4: 7042903-7042903 on Build GRCh38
rs912904774 Chr.4: 7043006-7043006 on Build GRCh38
rs751076362 Chr.4: 7042967-7042967 on Build GRCh38
rs554177563 Chr.4: 7043042-7043042 on Build GRCh38
rs762041425 Chr.4: 7042951-7042951 on Build GRCh38
rs549950164 Chr.4: 7042959-7042959 on Build GRCh38
rs1026284998 Chr.4: 7043066-7043066 on Build GRCh38
rs775553910 Chr.4: 7042944-7042944 on Build GRCh38
rs966634646 Chr.4: 7043067-7043067 on Build GRCh38
rs754289282 Chr.4: 7042923-7042923 on Build GRCh38
rs746673348 Chr.4: 7042933-7042933 on Build GRCh38
rs905943175 Chr.4: 7043015-7043015 on Build GRCh38
rs775068998 Chr.4: 7042970-7042971 on Build GRCh38
rs374155375 Chr.4: 7042947-7042947 on Build GRCh38
rs566428698 Chr.4: 7043049-7043049 on Build GRCh38
rs1010797758 Chr.4: 7043048-7043048 on Build GRCh38
rs749178458 Chr.4: 7042946-7042946 on Build GRCh38
rs950742188 Chr.4: 7042915-7042915 on Build GRCh38
rs906674908 Chr.4: 7043033-7043033 on Build GRCh38
rs530149078 Chr.4: 7042973-7042973 on Build GRCh38
rs773389273 Chr.4: 7042956-7042956 on Build GRCh38
rs1019187065 Chr.4: 7042955-7042955 on Build GRCh38
rs756642380 Chr.4: 7042912-7042912 on Build GRCh38
rs748264473 Chr.4: 7042989-7042989 on Build GRCh38
rs533573428 Chr.4: 7043100-7043100 on Build GRCh38
rs745699246 Chr.4: 7042941-7042941 on Build GRCh38
rs754378410 Chr.4: 7042963-7042963 on Build GRCh38
rs760279490 Chr.4: 7042985-7042985 on Build GRCh38
rs192060165 Chr.4: 7043003-7043003 on Build GRCh38
rs997805530 Chr.4: 7043096-7043096 on Build GRCh38
rs985576469 Chr.4: 7042935-7042935 on Build GRCh38
rs781039462 Chr.4: 7042970-7042970 on Build GRCh38
rs774443476 Chr.4: 7042977-7042977 on Build GRCh38
rs755383641 Chr.4: 7042964-7042964 on Build GRCh38
rs1055716298 Chr.4: 7043038-7043038 on Build GRCh38
rs756003221 Chr.4: 7042972-7042972 on Build GRCh38
rs938223224 Chr.4: 7043036-7043036 on Build GRCh38
rs949794164 Chr.4: 7043010-7043010 on Build GRCh38
rs1001641969 Chr.4: 7043021-7043021 on Build GRCh38
rs1029310018 Chr.4: 7043099-7043099 on Build GRCh38
rs745500794 Chr.4: 7042971-7042971 on Build GRCh38
rs567402403 Chr.4: 7042931-7042931 on Build GRCh38
rs757996689 Chr.4: 7042929-7042929 on Build GRCh38
rs539270710 Chr.4: 7043039-7043039 on Build GRCh38
rs780662498 Chr.4: 7042919-7042919 on Build GRCh38
rs932443709 Chr.4: 7042906-7042906 on Build GRCh38
rs756798163 Chr.4: 7042968-7042968 on Build GRCh38
rs370057062 Chr.4: 7042937-7042937 on Build GRCh38
rs770659366 Chr.4: 7042936-7042936 on Build GRCh38

View Full Product Details