Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37833094-37833321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD54
Entrez Gene ID
129138
Gene Name
ankyrin repeat domain 54
Gene Aliases
LIAR
UniGene
Hs.135259
Gene Chromosome Location
Chr.22: 37830854-37848994 on Build GRCh38
Transcripts
NM_138797.2 NR_036556.1 XM_006724136.1 XM_006724137.1 XM_011529877.2 XM_011529878.2 XM_011529879.2 XM_011529880.2 XM_011529882.2 XM_011529883.2 XM_011529884.2 XM_017028590.1 XM_017028591.1 XM_017028593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182950 c.589C>T p.R197* Chr.22: 37833165-37833165 on Build GRCh38
4445355 c.329G>A p.Q110Q Chr.22: 37833096-37833096 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs13056924 Chr.22: 37833290-37833290 on Build GRCh38
rs756051275 Chr.22: 37833285-37833286 on Build GRCh38
rs779228558 Chr.22: 37833216-37833218 on Build GRCh38
rs780130290 Chr.22: 37833121-37833121 on Build GRCh38
rs755300603 Chr.22: 37833136-37833137 on Build GRCh38
rs559520754 Chr.22: 37833279-37833279 on Build GRCh38
rs138041109 Chr.22: 37833299-37833299 on Build GRCh38
rs199708999 Chr.22: 37833131-37833131 on Build GRCh38
rs970124648 Chr.22: 37833261-37833261 on Build GRCh38
rs748117077 Chr.22: 37833248-37833248 on Build GRCh38
rs941550472 Chr.22: 37833292-37833292 on Build GRCh38
rs955919221 Chr.22: 37833155-37833155 on Build GRCh38
rs545207906 Chr.22: 37833298-37833298 on Build GRCh38
rs754947629 Chr.22: 37833129-37833129 on Build GRCh38
rs577772176 Chr.22: 37833294-37833294 on Build GRCh38
rs986094057 Chr.22: 37833285-37833285 on Build GRCh38
rs1014919873 Chr.22: 37833163-37833163 on Build GRCh38
rs966108500 Chr.22: 37833286-37833286 on Build GRCh38
rs772812047 Chr.22: 37833154-37833154 on Build GRCh38
rs767955748 Chr.22: 37833205-37833205 on Build GRCh38
rs1034438423 Chr.22: 37833124-37833124 on Build GRCh38
rs753983003 Chr.22: 37833223-37833223 on Build GRCh38
rs371302913 Chr.22: 37833136-37833136 on Build GRCh38
rs201355073 Chr.22: 37833184-37833184 on Build GRCh38
rs746685471 Chr.22: 37833162-37833162 on Build GRCh38
rs574259793 Chr.22: 37833189-37833189 on Build GRCh38
rs755035634 Chr.22: 37833245-37833245 on Build GRCh38
rs771910028 Chr.22: 37833146-37833146 on Build GRCh38
rs541401539 Chr.22: 37833220-37833220 on Build GRCh38
rs750703715 Chr.22: 37833206-37833206 on Build GRCh38
rs543772480 Chr.22: 37833123-37833123 on Build GRCh38
rs756288645 Chr.22: 37833120-37833120 on Build GRCh38
rs143148264 Chr.22: 37833169-37833169 on Build GRCh38
rs748264001 Chr.22: 37833228-37833229 on Build GRCh38
rs979876731 Chr.22: 37833262-37833262 on Build GRCh38
rs759114130 Chr.22: 37833183-37833183 on Build GRCh38
rs772821802 Chr.22: 37833172-37833172 on Build GRCh38
rs778796407 Chr.22: 37833246-37833246 on Build GRCh38
rs770688448 Chr.22: 37833164-37833164 on Build GRCh38
rs766556154 Chr.22: 37833221-37833221 on Build GRCh38

View Full Product Details