Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70049669-70049898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AWAT2
Entrez Gene ID
158835
Gene Name
acyl-CoA wax alcohol acyltransferase 2
Gene Aliases
ARAT, DC4, DGAT2L4, MFAT, WS
UniGene
Hs.661507
Gene Chromosome Location
Chr.X: 70040542-70049970 on Build GRCh38
Transcripts
NM_001002254.1 XM_011530876.2

SNP Detail

SNP ID Chromosome Location
rs745880837 Chr.X: 70049849-70049849 on Build GRCh38
rs757612942 Chr.X: 70049816-70049816 on Build GRCh38
rs62604276 Chr.X: 70049717-70049717 on Build GRCh38
rs756539713 Chr.X: 70049836-70049836 on Build GRCh38
rs887239377 Chr.X: 70049856-70049856 on Build GRCh38
rs1047077526 Chr.X: 70049829-70049829 on Build GRCh38
rs747590269 Chr.X: 70049858-70049858 on Build GRCh38
rs770439246 Chr.X: 70049754-70049754 on Build GRCh38
rs1006041919 Chr.X: 70049868-70049868 on Build GRCh38
rs147621322 Chr.X: 70049744-70049744 on Build GRCh38
rs780662978 Chr.X: 70049840-70049840 on Build GRCh38
rs750811516 Chr.X: 70049820-70049820 on Build GRCh38
rs775088835 Chr.X: 70049695-70049695 on Build GRCh38
rs191921246 Chr.X: 70049807-70049807 on Build GRCh38
rs768318748 Chr.X: 70049730-70049730 on Build GRCh38
rs761670321 Chr.X: 70049803-70049803 on Build GRCh38
rs895837911 Chr.X: 70049752-70049752 on Build GRCh38

View Full Product Details