Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6385447-6385716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT7
Entrez Gene ID
11332
Gene Name
acyl-CoA thioesterase 7
Gene Aliases
ACH1, ACT, BACH, CTE-II, LACH, LACH1, hBACH
UniGene
Hs.126137
Gene Chromosome Location
Chr.1: 6264272-6393766 on Build GRCh38
Transcripts
NM_007274.3 NM_181864.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5952587 c.97C>T p.R33* Chr.1: 6385570-6385570 on Build GRCh38
3790203 c.149C>G p.P50R Chr.1: 6385518-6385518 on Build GRCh38
2238831 c.109G>A p.A37T Chr.1: 6385558-6385558 on Build GRCh38
5053635 c.74G>A p.G25D Chr.1: 6385593-6385593 on Build GRCh38
6274589 c.142C>A p.L48I Chr.1: 6385525-6385525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780569034 Chr.1: 6385497-6385497 on Build GRCh38
rs922288826 Chr.1: 6385690-6385690 on Build GRCh38
rs746634199 Chr.1: 6385620-6385620 on Build GRCh38
rs781670466 Chr.1: 6385485-6385485 on Build GRCh38
rs373815001 Chr.1: 6385558-6385558 on Build GRCh38
rs371732190 Chr.1: 6385572-6385572 on Build GRCh38
rs776749790 Chr.1: 6385553-6385553 on Build GRCh38
rs767747865 Chr.1: 6385569-6385569 on Build GRCh38
rs149893425 Chr.1: 6385573-6385573 on Build GRCh38
rs978568399 Chr.1: 6385615-6385615 on Build GRCh38
rs773050219 Chr.1: 6385538-6385541 on Build GRCh38
rs775447654 Chr.1: 6385633-6385633 on Build GRCh38
rs559053195 Chr.1: 6385559-6385559 on Build GRCh38
rs911213277 Chr.1: 6385486-6385486 on Build GRCh38
rs558396989 Chr.1: 6385546-6385546 on Build GRCh38
rs746242908 Chr.1: 6385491-6385491 on Build GRCh38
rs369930242 Chr.1: 6385536-6385536 on Build GRCh38
rs747897842 Chr.1: 6385510-6385510 on Build GRCh38
rs757302904 Chr.1: 6385568-6385568 on Build GRCh38
rs537543895 Chr.1: 6385641-6385641 on Build GRCh38
rs199893009 Chr.1: 6385627-6385627 on Build GRCh38
rs757408867 Chr.1: 6385477-6385477 on Build GRCh38
rs747627222 Chr.1: 6385586-6385586 on Build GRCh38
rs777221235 Chr.1: 6385557-6385557 on Build GRCh38
rs749763389 Chr.1: 6385498-6385498 on Build GRCh38
rs577253801 Chr.1: 6385570-6385570 on Build GRCh38
rs779407085 Chr.1: 6385583-6385583 on Build GRCh38
rs529640665 Chr.1: 6385608-6385608 on Build GRCh38
rs768258189 Chr.1: 6385501-6385501 on Build GRCh38
rs751589406 Chr.1: 6385560-6385560 on Build GRCh38
rs373937124 Chr.1: 6385642-6385642 on Build GRCh38
rs915864790 Chr.1: 6385598-6385598 on Build GRCh38
rs773033812 Chr.1: 6385618-6385618 on Build GRCh38
rs376250754 Chr.1: 6385623-6385623 on Build GRCh38
rs776555854 Chr.1: 6385625-6385625 on Build GRCh38
rs993269089 Chr.1: 6385564-6385564 on Build GRCh38
rs960337803 Chr.1: 6385575-6385575 on Build GRCh38
rs770093912 Chr.1: 6385493-6385493 on Build GRCh38
rs750692435 Chr.1: 6385664-6385664 on Build GRCh38
rs542134906 Chr.1: 6385660-6385660 on Build GRCh38
rs771625387 Chr.1: 6385517-6385517 on Build GRCh38
rs115915823 Chr.1: 6385502-6385502 on Build GRCh38
rs576668181 Chr.1: 6385552-6385552 on Build GRCh38
rs746632593 Chr.1: 6385642-6385643 on Build GRCh38
rs754154470 Chr.1: 6385672-6385672 on Build GRCh38
rs767285472 Chr.1: 6385488-6385488 on Build GRCh38
rs377549870 Chr.1: 6385671-6385671 on Build GRCh38
rs1032732342 Chr.1: 6385647-6385647 on Build GRCh38
rs759587518 Chr.1: 6385626-6385626 on Build GRCh38
rs755432796 Chr.1: 6385580-6385580 on Build GRCh38
rs968624994 Chr.1: 6385601-6385601 on Build GRCh38
rs549941128 Chr.1: 6385694-6385694 on Build GRCh38
rs749848196 Chr.1: 6385579-6385579 on Build GRCh38
rs369668675 Chr.1: 6385592-6385592 on Build GRCh38

View Full Product Details