Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142873323-142873570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP26
Entrez Gene ID
23092
Gene Name
Rho GTPase activating protein 26
Gene Aliases
GRAF, GRAF1, OPHN1L, OPHN1L1
UniGene
Hs.654668
Gene Chromosome Location
Chr.5: 142770370-143229007 on Build GRCh38
Transcripts
NM_001135608.1 NM_015071.4 XM_005268398.4 XM_005268399.4 XM_005268400.4 XM_005268402.4 XM_006714774.3 XM_011537610.2 XM_011537611.2 XM_011537612.2 XM_017009247.1 XM_017009248.1 XM_017009249.1 XM_017009250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4531324 c.177G>A p.K59K Chr.5: 142873422-142873422 on Build GRCh38
6603328 c.196G>A p.E66K Chr.5: 142873441-142873441 on Build GRCh38
1245342 c.208C>G p.Q70E Chr.5: 142873453-142873453 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377359847 Chr.5: 142873507-142873507 on Build GRCh38
rs777084250 Chr.5: 142873392-142873392 on Build GRCh38
rs556866282 Chr.5: 142873358-142873358 on Build GRCh38
rs745414431 Chr.5: 142873357-142873357 on Build GRCh38
rs376354527 Chr.5: 142873544-142873544 on Build GRCh38
rs933750942 Chr.5: 142873501-142873501 on Build GRCh38
rs779633329 Chr.5: 142873364-142873364 on Build GRCh38
rs376170795 Chr.5: 142873383-142873383 on Build GRCh38
rs766608479 Chr.5: 142873413-142873413 on Build GRCh38
rs779692526 Chr.5: 142873530-142873530 on Build GRCh38
rs748677239 Chr.5: 142873396-142873396 on Build GRCh38
rs761816054 Chr.5: 142873470-142873470 on Build GRCh38
rs764549933 Chr.5: 142873498-142873498 on Build GRCh38
rs769014324 Chr.5: 142873387-142873387 on Build GRCh38
rs773309825 Chr.5: 142873403-142873403 on Build GRCh38
rs765158622 Chr.5: 142873478-142873478 on Build GRCh38
rs200276995 Chr.5: 142873457-142873457 on Build GRCh38
rs770251969 Chr.5: 142873401-142873401 on Build GRCh38
rs1038258366 Chr.5: 142873549-142873549 on Build GRCh38
rs138620429 Chr.5: 142873412-142873412 on Build GRCh38
rs982598003 Chr.5: 142873517-142873517 on Build GRCh38
rs576416706 Chr.5: 142873372-142873372 on Build GRCh38
rs751244793 Chr.5: 142873531-142873531 on Build GRCh38
rs989730276 Chr.5: 142873381-142873381 on Build GRCh38
rs764008846 Chr.5: 142873461-142873461 on Build GRCh38
rs755287291 Chr.5: 142873534-142873534 on Build GRCh38
rs922352544 Chr.5: 142873486-142873486 on Build GRCh38
rs975495910 Chr.5: 142873418-142873418 on Build GRCh38
rs753775256 Chr.5: 142873465-142873465 on Build GRCh38
rs757862982 Chr.5: 142873518-142873518 on Build GRCh38
rs763483140 Chr.5: 142873533-142873533 on Build GRCh38
rs774726416 Chr.5: 142873417-142873417 on Build GRCh38

View Full Product Details