Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132679271-132679514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C4
Entrez Gene ID
9329
Gene Name
general transcription factor IIIC subunit 4
Gene Aliases
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA
UniGene
Hs.22302
Gene Chromosome Location
Chr.9: 132670035-132694955 on Build GRCh38
Transcripts
NM_012204.3 NR_133925.1 XM_017015317.1 XM_017015318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6676836 c.1685A>G p.E562G Chr.9: 132679304-132679304 on Build GRCh38
210975 c.1841_1842insG p.N616fs*3 Chr.9: 132679460-132679461 on Build GRCh38
3847945 c.1784C>T p.A595V Chr.9: 132679403-132679403 on Build GRCh38
1209177 c.1832C>T p.S611L Chr.9: 132679451-132679451 on Build GRCh38
6182675 c.1870G>C p.E624Q Chr.9: 132679489-132679489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758442546 Chr.9: 132679390-132679390 on Build GRCh38
rs145388767 Chr.9: 132679353-132679353 on Build GRCh38
rs567122373 Chr.9: 132679320-132679320 on Build GRCh38
rs759943928 Chr.9: 132679381-132679381 on Build GRCh38
rs1050022135 Chr.9: 132679451-132679451 on Build GRCh38
rs370378462 Chr.9: 132679430-132679430 on Build GRCh38
rs199574706 Chr.9: 132679437-132679437 on Build GRCh38
rs753145398 Chr.9: 132679389-132679389 on Build GRCh38
rs751790621 Chr.9: 132679410-132679410 on Build GRCh38
rs765836358 Chr.9: 132679383-132679383 on Build GRCh38
rs534512256 Chr.9: 132679426-132679426 on Build GRCh38
rs757590948 Chr.9: 132679423-132679423 on Build GRCh38
rs766693428 Chr.9: 132679361-132679361 on Build GRCh38
rs1031666448 Chr.9: 132679452-132679452 on Build GRCh38
rs199853514 Chr.9: 132679473-132679473 on Build GRCh38
rs149046524 Chr.9: 132679419-132679419 on Build GRCh38
rs754243448 Chr.9: 132679371-132679371 on Build GRCh38
rs772794978 Chr.9: 132679341-132679341 on Build GRCh38
rs778020564 Chr.9: 132679404-132679404 on Build GRCh38
rs771488209 Chr.9: 132679323-132679323 on Build GRCh38
rs778198525 Chr.9: 132679377-132679377 on Build GRCh38
rs145764040 Chr.9: 132679314-132679314 on Build GRCh38
rs577534312 Chr.9: 132679444-132679444 on Build GRCh38

View Full Product Details