Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138556499-138556744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA9
Entrez Gene ID
3313
Gene Name
heat shock protein family A (Hsp70) member 9
Gene Aliases
CRP40, CSA, EVPLS, GRP-75, GRP75, HEL-S-124m, HSPA9B, MOT, MOT2, MTHSP75, PBP74, SAAN, SIDBA4
UniGene
Hs.184233
Gene Chromosome Location
Chr.5: 138554882-138575629 on Build GRCh38
Transcripts
NM_004134.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7140852 c.1833G>C p.L611L Chr.5: 138556581-138556581 on Build GRCh38
6682669 c.1830G>T p.K610N Chr.5: 138556584-138556584 on Build GRCh38
7102083 c.1863C>G p.L621L Chr.5: 138556551-138556551 on Build GRCh38
281787 c.1822-8T>G p.? Chr.5: 138556600-138556600 on Build GRCh38
6682665 c.1881C>T p.S627S Chr.5: 138556533-138556533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978678662 Chr.5: 138556662-138556662 on Build GRCh38
rs760535812 Chr.5: 138556592-138556592 on Build GRCh38
rs770647682 Chr.5: 138556596-138556596 on Build GRCh38
rs368659266 Chr.5: 138556604-138556604 on Build GRCh38
rs1042686 Chr.5: 138556533-138556533 on Build GRCh38
rs773524708 Chr.5: 138556596-138556596 on Build GRCh38
rs772946816 Chr.5: 138556643-138556643 on Build GRCh38
rs995805734 Chr.5: 138556642-138556642 on Build GRCh38
rs748079161 Chr.5: 138556570-138556570 on Build GRCh38
rs780426518 Chr.5: 138556542-138556542 on Build GRCh38
rs748824826 Chr.5: 138556716-138556716 on Build GRCh38
rs968003320 Chr.5: 138556647-138556647 on Build GRCh38
rs528118611 Chr.5: 138556672-138556672 on Build GRCh38
rs377697898 Chr.5: 138556614-138556614 on Build GRCh38
rs779025678 Chr.5: 138556569-138556569 on Build GRCh38
rs566717208 Chr.5: 138556532-138556532 on Build GRCh38
rs759141353 Chr.5: 138556603-138556603 on Build GRCh38
rs1009572159 Chr.5: 138556620-138556620 on Build GRCh38
rs776296627 Chr.5: 138556599-138556599 on Build GRCh38
rs772942312 Chr.5: 138556585-138556585 on Build GRCh38
rs772044929 Chr.5: 138556573-138556573 on Build GRCh38
rs373475418 Chr.5: 138556540-138556540 on Build GRCh38
rs768876845 Chr.5: 138556559-138556559 on Build GRCh38
rs749478409 Chr.5: 138556549-138556549 on Build GRCh38
rs756575313 Chr.5: 138556637-138556637 on Build GRCh38
rs768082547 Chr.5: 138556631-138556631 on Build GRCh38
rs372765581 Chr.5: 138556636-138556636 on Build GRCh38
rs760971853 Chr.5: 138556596-138556599 on Build GRCh38
rs762283564 Chr.5: 138556629-138556629 on Build GRCh38
rs778321149 Chr.5: 138556599-138556603 on Build GRCh38

View Full Product Details