Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22472229-22472458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD5
Entrez Gene ID
9847
Gene Name
C2 calcium dependent domain containing 5
Gene Aliases
CDP138, KIAA0528
UniGene
Hs.271014
Gene Chromosome Location
Chr.12: 22448546-22544546 on Build GRCh38
Transcripts
NM_001286173.1 NM_001286174.1 NM_001286175.1 NM_001286176.1 NM_001286177.1 NM_014802.2 XM_005253538.2 XM_005253539.3 XM_006719170.2 XM_017020264.1 XM_017020265.1 XM_017020266.1 XM_017020267.1 XM_017020268.1 XM_017020269.1 XM_017020270.1 XM_017020271.1 XM_017020272.1 XM_017020273.1 XM_017020274.1 XM_017020275.1 XM_017020276.1 XM_017020277.1 XM_017020278.1 XM_017020279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4477479 c.1602C>T p.T534T Chr.12: 22472298-22472298 on Build GRCh38
5170012 c.1553-7T>G p.? Chr.12: 22472354-22472354 on Build GRCh38
4477478 c.2157C>T p.T719T Chr.12: 22472298-22472298 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1047864100 Chr.12: 22472321-22472321 on Build GRCh38
rs765762043 Chr.12: 22472317-22472317 on Build GRCh38
rs202165258 Chr.12: 22472341-22472341 on Build GRCh38
rs189750753 Chr.12: 22472406-22472406 on Build GRCh38
rs954501292 Chr.12: 22472402-22472402 on Build GRCh38
rs749283585 Chr.12: 22472388-22472388 on Build GRCh38
rs780727140 Chr.12: 22472363-22472363 on Build GRCh38
rs760064122 Chr.12: 22472305-22472305 on Build GRCh38
rs766527258 Chr.12: 22472302-22472302 on Build GRCh38
rs756981827 Chr.12: 22472350-22472350 on Build GRCh38
rs760692248 Chr.12: 22472273-22472273 on Build GRCh38
rs750048468 Chr.12: 22472371-22472371 on Build GRCh38
rs548237076 Chr.12: 22472271-22472271 on Build GRCh38
rs1014830341 Chr.12: 22472419-22472419 on Build GRCh38
rs755622091 Chr.12: 22472375-22472375 on Build GRCh38
rs529036598 Chr.12: 22472420-22472420 on Build GRCh38
rs752971808 Chr.12: 22472346-22472346 on Build GRCh38
rs16924983 Chr.12: 22472304-22472304 on Build GRCh38
rs906496896 Chr.12: 22472330-22472330 on Build GRCh38
rs768579011 Chr.12: 22472398-22472398 on Build GRCh38
rs899914013 Chr.12: 22472323-22472323 on Build GRCh38
rs375777831 Chr.12: 22472380-22472380 on Build GRCh38

View Full Product Details