Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176645781-176646050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E1B
Entrez Gene ID
253314
Gene Name
eukaryotic translation initiation factor 4E family member 1B
Gene Aliases
-
UniGene
Hs.448362
Gene Chromosome Location
Chr.5: 176630613-176646644 on Build GRCh38
Transcripts
NM_001099408.1 XM_011534499.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6513665 c.631C>A p.R211S Chr.5: 176645882-176645882 on Build GRCh38
4745779 c.625A>T p.K209* Chr.5: 176645876-176645876 on Build GRCh38
160681 c.642C>T p.L214L Chr.5: 176645893-176645893 on Build GRCh38
5908481 c.644C>T p.S215F Chr.5: 176645895-176645895 on Build GRCh38
4920721 c.626A>G p.K209R Chr.5: 176645877-176645877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779041907 Chr.5: 176646023-176646023 on Build GRCh38
rs540602818 Chr.5: 176646019-176646019 on Build GRCh38
rs555063008 Chr.5: 176645843-176645843 on Build GRCh38
rs779221982 Chr.5: 176645904-176645904 on Build GRCh38
rs757870734 Chr.5: 176645889-176645889 on Build GRCh38
rs746706986 Chr.5: 176645928-176645928 on Build GRCh38
rs766151683 Chr.5: 176645995-176645995 on Build GRCh38
rs372249540 Chr.5: 176645856-176645856 on Build GRCh38
rs761785957 Chr.5: 176645862-176645862 on Build GRCh38
rs530950799 Chr.5: 176645923-176645923 on Build GRCh38
rs758911475 Chr.5: 176646029-176646029 on Build GRCh38
rs760499683 Chr.5: 176645824-176645826 on Build GRCh38
rs753850774 Chr.5: 176645931-176645932 on Build GRCh38
rs182336246 Chr.5: 176645867-176645867 on Build GRCh38
rs761002677 Chr.5: 176646008-176646008 on Build GRCh38
rs369228216 Chr.5: 176645848-176645848 on Build GRCh38
rs756399527 Chr.5: 176645883-176645883 on Build GRCh38
rs917717354 Chr.5: 176645820-176645820 on Build GRCh38
rs749853369 Chr.5: 176645839-176645839 on Build GRCh38
rs13183830 Chr.5: 176645976-176645976 on Build GRCh38
rs766548962 Chr.5: 176645843-176645844 on Build GRCh38
rs756203622 Chr.5: 176645822-176645822 on Build GRCh38
rs779126575 Chr.5: 176645865-176645865 on Build GRCh38
rs762933520 Chr.5: 176645994-176645994 on Build GRCh38
rs992930595 Chr.5: 176645814-176645814 on Build GRCh38
rs774951072 Chr.5: 176645851-176645851 on Build GRCh38
rs760310232 Chr.5: 176645855-176645855 on Build GRCh38
rs561324254 Chr.5: 176645989-176645989 on Build GRCh38
rs1047630727 Chr.5: 176645823-176645823 on Build GRCh38
rs771544393 Chr.5: 176645841-176645841 on Build GRCh38
rs761268817 Chr.5: 176645963-176645963 on Build GRCh38
rs780464331 Chr.5: 176645922-176645922 on Build GRCh38
rs13163938 Chr.5: 176645930-176645930 on Build GRCh38
rs573376055 Chr.5: 176646018-176646018 on Build GRCh38
rs753922679 Chr.5: 176645888-176645888 on Build GRCh38
rs778049289 Chr.5: 176645887-176645887 on Build GRCh38
rs746178693 Chr.5: 176645847-176645847 on Build GRCh38
rs543131273 Chr.5: 176645945-176645945 on Build GRCh38
rs746260963 Chr.5: 176645910-176645910 on Build GRCh38
rs777984762 Chr.5: 176645835-176645835 on Build GRCh38
rs772381239 Chr.5: 176645912-176645912 on Build GRCh38
rs776084974 Chr.5: 176645857-176645857 on Build GRCh38
rs772964599 Chr.5: 176645991-176645991 on Build GRCh38
rs969892994 Chr.5: 176645825-176645825 on Build GRCh38
rs369631741 Chr.5: 176645868-176645868 on Build GRCh38
rs751379082 Chr.5: 176645996-176645996 on Build GRCh38
rs752860930 Chr.5: 176645882-176645882 on Build GRCh38
rs755209254 Chr.5: 176645980-176645981 on Build GRCh38
rs900197990 Chr.5: 176645890-176645890 on Build GRCh38
rs764372571 Chr.5: 176646010-176646010 on Build GRCh38
rs1040146907 Chr.5: 176646003-176646003 on Build GRCh38
rs995845205 Chr.5: 176645917-176645917 on Build GRCh38
rs750961557 Chr.5: 176646028-176646028 on Build GRCh38
rs980769156 Chr.5: 176645828-176645828 on Build GRCh38

View Full Product Details