Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63820780-63821019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTSJ3
Entrez Gene ID
117246
Gene Name
FtsJ homolog 3
Gene Aliases
EPCS3, SPB1
UniGene
Hs.463785
Gene Chromosome Location
Chr.17: 63819433-63827671 on Build GRCh38
Transcripts
NM_017647.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
437122 c.2069A>G p.N690S Chr.17: 63820842-63820842 on Build GRCh38
6876971 c.1989G>A p.L663L Chr.17: 63820922-63820922 on Build GRCh38
5016586 c.2040delC p.K681fs*5 Chr.17: 63820871-63820871 on Build GRCh38
6876970 c.1990G>A p.D664N Chr.17: 63820921-63820921 on Build GRCh38
3773006 c.2022T>A p.I674I Chr.17: 63820889-63820889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749932773 Chr.17: 63820937-63820937 on Build GRCh38
rs763053621 Chr.17: 63820912-63820912 on Build GRCh38
rs372742570 Chr.17: 63820825-63820825 on Build GRCh38
rs781520729 Chr.17: 63820947-63820947 on Build GRCh38
rs752574892 Chr.17: 63820829-63820829 on Build GRCh38
rs779693529 Chr.17: 63820938-63820938 on Build GRCh38
rs974075865 Chr.17: 63820814-63820814 on Build GRCh38
rs753764737 Chr.17: 63820849-63820849 on Build GRCh38
rs192706486 Chr.17: 63820970-63820970 on Build GRCh38
rs374821474 Chr.17: 63820984-63820984 on Build GRCh38
rs545130754 Chr.17: 63820926-63820926 on Build GRCh38
rs943858693 Chr.17: 63820900-63820900 on Build GRCh38
rs140503316 Chr.17: 63820870-63820870 on Build GRCh38
rs375155785 Chr.17: 63820978-63820978 on Build GRCh38
rs76727299 Chr.17: 63820993-63820993 on Build GRCh38
rs758495269 Chr.17: 63820807-63820807 on Build GRCh38
rs377649318 Chr.17: 63820980-63820980 on Build GRCh38
rs769972059 Chr.17: 63820899-63820899 on Build GRCh38
rs767371045 Chr.17: 63820812-63820812 on Build GRCh38
rs1027154395 Chr.17: 63820862-63820862 on Build GRCh38
rs1023383396 Chr.17: 63820942-63820942 on Build GRCh38
rs751776325 Chr.17: 63820916-63820916 on Build GRCh38
rs753656347 Chr.17: 63820989-63820989 on Build GRCh38
rs771488349 Chr.17: 63820852-63820852 on Build GRCh38
rs114729867 Chr.17: 63820890-63820890 on Build GRCh38
rs760935264 Chr.17: 63820829-63820830 on Build GRCh38
rs768950577 Chr.17: 63820875-63820875 on Build GRCh38
rs750666120 Chr.17: 63820803-63820803 on Build GRCh38
rs199519029 Chr.17: 63820840-63820840 on Build GRCh38
rs934846941 Chr.17: 63820959-63820959 on Build GRCh38
rs746415246 Chr.17: 63820812-63820813 on Build GRCh38
rs778274921 Chr.17: 63820861-63820861 on Build GRCh38
rs772447463 Chr.17: 63820816-63820818 on Build GRCh38
rs779716903 Chr.17: 63820868-63820868 on Build GRCh38
rs565074305 Chr.17: 63820941-63820941 on Build GRCh38
rs200211760 Chr.17: 63820958-63820958 on Build GRCh38
rs200615117 Chr.17: 63820915-63820915 on Build GRCh38
rs762301427 Chr.17: 63820892-63820892 on Build GRCh38
rs199801932 Chr.17: 63820981-63820981 on Build GRCh38
rs764163051 Chr.17: 63820989-63820989 on Build GRCh38
rs775781377 Chr.17: 63820819-63820820 on Build GRCh38
rs188934049 Chr.17: 63820968-63820968 on Build GRCh38
rs773724045 Chr.17: 63820963-63820963 on Build GRCh38
rs770165747 Chr.17: 63820952-63820952 on Build GRCh38
rs771592744 Chr.17: 63820864-63820864 on Build GRCh38
rs761629210 Chr.17: 63820990-63820990 on Build GRCh38
rs200808492 Chr.17: 63820932-63820932 on Build GRCh38
rs763978409 Chr.17: 63820929-63820929 on Build GRCh38
rs757121437 Chr.17: 63820936-63820936 on Build GRCh38
rs138285537 Chr.17: 63820909-63820909 on Build GRCh38
rs756860868 Chr.17: 63820857-63820857 on Build GRCh38

View Full Product Details