Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 125039682-125039955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0196
Entrez Gene ID
9897
Gene Name
KIAA0196
Gene Aliases
RTSC, RTSC1, SPG8
UniGene
Hs.270043
Gene Chromosome Location
Chr.8: 125024260-125091819 on Build GRCh38
Transcripts
NM_014846.3 XM_005251120.3 XM_011517409.1 XM_017014113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014818 c.2948T>C p.L983P Chr.8: 125039801-125039801 on Build GRCh38
1095956 c.2933C>T p.A978V Chr.8: 125039816-125039816 on Build GRCh38
6365857 c.2942A>G p.E981G Chr.8: 125039807-125039807 on Build GRCh38
4934759 c.2882T>C p.I961T Chr.8: 125039867-125039867 on Build GRCh38
3834030 c.2862G>A p.M954I Chr.8: 125039887-125039887 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745549627 Chr.8: 125039744-125039744 on Build GRCh38
rs768316958 Chr.8: 125039848-125039848 on Build GRCh38
rs774438456 Chr.8: 125039766-125039766 on Build GRCh38
rs748167994 Chr.8: 125039861-125039861 on Build GRCh38
rs759625490 Chr.8: 125039798-125039798 on Build GRCh38
rs769176746 Chr.8: 125039752-125039752 on Build GRCh38
rs765321662 Chr.8: 125039922-125039922 on Build GRCh38
rs927990562 Chr.8: 125039785-125039785 on Build GRCh38
rs760999456 Chr.8: 125039791-125039791 on Build GRCh38
rs745705899 Chr.8: 125039720-125039720 on Build GRCh38
rs979606809 Chr.8: 125039719-125039719 on Build GRCh38
rs778801994 Chr.8: 125039717-125039717 on Build GRCh38
rs548400961 Chr.8: 125039915-125039915 on Build GRCh38
rs759570613 Chr.8: 125039920-125039920 on Build GRCh38
rs111281783 Chr.8: 125039739-125039739 on Build GRCh38
rs777800320 Chr.8: 125039849-125039853 on Build GRCh38
rs773286131 Chr.8: 125039779-125039779 on Build GRCh38
rs565875405 Chr.8: 125039806-125039806 on Build GRCh38
rs139669979 Chr.8: 125039845-125039845 on Build GRCh38
rs768660279 Chr.8: 125039765-125039765 on Build GRCh38
rs900850009 Chr.8: 125039745-125039745 on Build GRCh38
rs563786689 Chr.8: 125039916-125039916 on Build GRCh38
rs368458656 Chr.8: 125039814-125039814 on Build GRCh38
rs749228892 Chr.8: 125039756-125039756 on Build GRCh38
rs766366067 Chr.8: 125039792-125039792 on Build GRCh38
rs771083273 Chr.8: 125039918-125039918 on Build GRCh38
rs753232489 Chr.8: 125039810-125039810 on Build GRCh38
rs771802610 Chr.8: 125039821-125039821 on Build GRCh38
rs1040993525 Chr.8: 125039740-125039740 on Build GRCh38
rs886062651 Chr.8: 125039782-125039782 on Build GRCh38
rs867751848 Chr.8: 125039917-125039917 on Build GRCh38
rs771735577 Chr.8: 125039862-125039862 on Build GRCh38
rs775211008 Chr.8: 125039755-125039755 on Build GRCh38
rs552149149 Chr.8: 125039770-125039770 on Build GRCh38
rs776843021 Chr.8: 125039919-125039919 on Build GRCh38
rs773126796 Chr.8: 125039882-125039882 on Build GRCh38
rs755858522 Chr.8: 125039840-125039840 on Build GRCh38
rs753976948 Chr.8: 125039796-125039796 on Build GRCh38
rs778948449 Chr.8: 125039850-125039850 on Build GRCh38
rs750010364 Chr.8: 125039824-125039824 on Build GRCh38
rs779693936 Chr.8: 125039841-125039841 on Build GRCh38

View Full Product Details