Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97859299-97859564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAC1
Entrez Gene ID
55118
Gene Name
cartilage acidic protein 1
Gene Aliases
ASPIC, ASPIC1, CEP-68
UniGene
Hs.500736
Gene Chromosome Location
Chr.10: 97854711-98030828 on Build GRCh38
Transcripts
XM_011539917.1 NM_001010917.2

Gene Symbol
GOLGA7B
Entrez Gene ID
401647
Gene Name
golgin A7 family member B
Gene Aliases
C10orf132, C10orf133, bA451M19.3, bA459F3.4
UniGene
Hs.567524
Gene Chromosome Location
Chr.10: 97850238-97871578 on Build GRCh38
Transcripts
XM_011539917.1 NM_001010917.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1963563 c.101A>G p.Q34R Chr.10: 97859546-97859546 on Build GRCh38
922074 c.27G>T p.Q9H Chr.10: 97859472-97859472 on Build GRCh38
3442446 c.76G>A p.D26N Chr.10: 97859521-97859521 on Build GRCh38
6569022 c.33C>G p.L11L Chr.10: 97859478-97859478 on Build GRCh38
4017109 c.85G>A p.D29N Chr.10: 97859530-97859530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143455176 Chr.10: 97859492-97859492 on Build GRCh38
rs567531286 Chr.10: 97859494-97859494 on Build GRCh38
rs765913755 Chr.10: 97859498-97859498 on Build GRCh38
rs531579996 Chr.10: 97859473-97859473 on Build GRCh38
rs200347657 Chr.10: 97859532-97859532 on Build GRCh38
rs747038840 Chr.10: 97859481-97859481 on Build GRCh38
rs951908987 Chr.10: 97859344-97859344 on Build GRCh38
rs985883780 Chr.10: 97859354-97859354 on Build GRCh38
rs377015068 Chr.10: 97859410-97859410 on Build GRCh38
rs758951956 Chr.10: 97859522-97859522 on Build GRCh38
rs139595752 Chr.10: 97859429-97859429 on Build GRCh38
rs201805184 Chr.10: 97859445-97859445 on Build GRCh38
rs778400554 Chr.10: 97859448-97859448 on Build GRCh38
rs556487533 Chr.10: 97859345-97859345 on Build GRCh38
rs754308200 Chr.10: 97859452-97859452 on Build GRCh38
rs760701630 Chr.10: 97859428-97859428 on Build GRCh38
rs149717226 Chr.10: 97859444-97859444 on Build GRCh38
rs370419153 Chr.10: 97859479-97859479 on Build GRCh38
rs776461048 Chr.10: 97859432-97859432 on Build GRCh38
rs200719626 Chr.10: 97859530-97859530 on Build GRCh38
rs904511795 Chr.10: 97859437-97859437 on Build GRCh38
rs772148450 Chr.10: 97859480-97859480 on Build GRCh38
rs574731720 Chr.10: 97859359-97859359 on Build GRCh38
rs761671560 Chr.10: 97859483-97859483 on Build GRCh38
rs757688740 Chr.10: 97859463-97859463 on Build GRCh38
rs772210529 Chr.10: 97859414-97859414 on Build GRCh38
rs148717823 Chr.10: 97859384-97859385 on Build GRCh38
rs1040054896 Chr.10: 97859538-97859538 on Build GRCh38
rs756682474 Chr.10: 97859447-97859447 on Build GRCh38
rs746126130 Chr.10: 97859475-97859475 on Build GRCh38
rs764936617 Chr.10: 97859439-97859439 on Build GRCh38
rs750080047 Chr.10: 97859440-97859440 on Build GRCh38
rs757930495 Chr.10: 97859443-97859443 on Build GRCh38
rs576743164 Chr.10: 97859339-97859339 on Build GRCh38
rs1027376844 Chr.10: 97859334-97859334 on Build GRCh38
rs768638204 Chr.10: 97859482-97859482 on Build GRCh38
rs761583597 Chr.10: 97859434-97859434 on Build GRCh38
rs913793165 Chr.10: 97859419-97859419 on Build GRCh38
rs775385022 Chr.10: 97859424-97859424 on Build GRCh38
rs910418236 Chr.10: 97859364-97859364 on Build GRCh38
rs769589632 Chr.10: 97859488-97859488 on Build GRCh38
rs746014428 Chr.10: 97859411-97859411 on Build GRCh38
rs146085693 Chr.10: 97859341-97859341 on Build GRCh38
rs944519166 Chr.10: 97859390-97859390 on Build GRCh38
rs199703901 Chr.10: 97859521-97859521 on Build GRCh38
rs199523057 Chr.10: 97859413-97859413 on Build GRCh38
rs374417789 Chr.10: 97859489-97859489 on Build GRCh38
rs751126796 Chr.10: 97859506-97859506 on Build GRCh38
rs764761052 Chr.10: 97859529-97859529 on Build GRCh38

View Full Product Details