Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44772687-44772926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
UniGene
Hs.6088
Gene Chromosome Location
Chr.17: 44759037-44781846 on Build GRCh38
Transcripts
NM_001318933.1 NM_002390.5 XM_005257373.3 XM_017024647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5587121 c.745C>T p.H249Y Chr.17: 44772923-44772923 on Build GRCh38
249652 c.734T>C p.V245A Chr.17: 44772912-44772912 on Build GRCh38
187787 c.683G>A p.R228H Chr.17: 44772861-44772861 on Build GRCh38
7216543 c.741C>T p.N247N Chr.17: 44772919-44772919 on Build GRCh38
1302898 c.738C>T p.I246I Chr.17: 44772916-44772916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376634027 Chr.17: 44772844-44772844 on Build GRCh38
rs752220717 Chr.17: 44772873-44772873 on Build GRCh38
rs773292017 Chr.17: 44772849-44772849 on Build GRCh38
rs760584511 Chr.17: 44772853-44772853 on Build GRCh38
rs980348960 Chr.17: 44772845-44772845 on Build GRCh38
rs1038804311 Chr.17: 44772768-44772768 on Build GRCh38
rs900593575 Chr.17: 44772729-44772729 on Build GRCh38
rs749792475 Chr.17: 44772821-44772821 on Build GRCh38
rs569481370 Chr.17: 44772802-44772802 on Build GRCh38
rs754927296 Chr.17: 44772864-44772864 on Build GRCh38
rs994840377 Chr.17: 44772772-44772772 on Build GRCh38
rs769282688 Chr.17: 44772820-44772824 on Build GRCh38
rs1027652439 Chr.17: 44772780-44772780 on Build GRCh38
rs772613698 Chr.17: 44772839-44772840 on Build GRCh38
rs979571590 Chr.17: 44772752-44772752 on Build GRCh38
rs191624738 Chr.17: 44772843-44772843 on Build GRCh38
rs751143606 Chr.17: 44772881-44772881 on Build GRCh38
rs1034093827 Chr.17: 44772847-44772847 on Build GRCh38
rs551518918 Chr.17: 44772835-44772835 on Build GRCh38
rs187592549 Chr.17: 44772804-44772804 on Build GRCh38
rs753819242 Chr.17: 44772863-44772863 on Build GRCh38
rs765115407 Chr.17: 44772867-44772867 on Build GRCh38
rs943189917 Chr.17: 44772765-44772765 on Build GRCh38
rs200895725 Chr.17: 44772861-44772861 on Build GRCh38
rs777253895 Chr.17: 44772895-44772895 on Build GRCh38
rs549588089 Chr.17: 44772801-44772801 on Build GRCh38
rs1004785592 Chr.17: 44772839-44772839 on Build GRCh38
rs200695565 Chr.17: 44772875-44772875 on Build GRCh38
rs555358248 Chr.17: 44772904-44772904 on Build GRCh38
rs1006337507 Chr.17: 44772882-44772882 on Build GRCh38
rs769236614 Chr.17: 44772823-44772823 on Build GRCh38
rs147644154 Chr.17: 44772877-44772877 on Build GRCh38
rs200118240 Chr.17: 44772820-44772820 on Build GRCh38
rs925169314 Chr.17: 44772764-44772765 on Build GRCh38
rs531467805 Chr.17: 44772781-44772781 on Build GRCh38
rs886339219 Chr.17: 44772813-44772813 on Build GRCh38
rs945162707 Chr.17: 44772737-44772737 on Build GRCh38

View Full Product Details