Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56412338-56412611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASS4
Entrez Gene ID
57091
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
UniGene
Hs.473144
Gene Chromosome Location
Chr.20: 56411550-56460240 on Build GRCh38
Transcripts
NM_001164114.1 NM_001164115.1 NM_001164116.1 NM_020356.3 XM_006723831.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3701643 c.36G>T p.K12N Chr.20: 56412494-56412494 on Build GRCh38
6244078 c.13G>T p.G5C Chr.20: 56412471-56412471 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755597485 Chr.20: 56412529-56412529 on Build GRCh38
rs752416290 Chr.20: 56412523-56412523 on Build GRCh38
rs568097945 Chr.20: 56412455-56412455 on Build GRCh38
rs955506351 Chr.20: 56412395-56412395 on Build GRCh38
rs766264727 Chr.20: 56412488-56412488 on Build GRCh38
rs890799897 Chr.20: 56412469-56412469 on Build GRCh38
rs138191448 Chr.20: 56412474-56412474 on Build GRCh38
rs142858434 Chr.20: 56412530-56412530 on Build GRCh38
rs571011329 Chr.20: 56412584-56412584 on Build GRCh38
rs755654980 Chr.20: 56412438-56412438 on Build GRCh38
rs200155192 Chr.20: 56412456-56412456 on Build GRCh38
rs745826641 Chr.20: 56412515-56412518 on Build GRCh38
rs773960659 Chr.20: 56412491-56412491 on Build GRCh38
rs145646754 Chr.20: 56412423-56412423 on Build GRCh38
rs190767176 Chr.20: 56412429-56412429 on Build GRCh38
rs1054502523 Chr.20: 56412555-56412555 on Build GRCh38
rs775780911 Chr.20: 56412466-56412466 on Build GRCh38
rs904084702 Chr.20: 56412434-56412434 on Build GRCh38
rs757957308 Chr.20: 56412383-56412383 on Build GRCh38
rs183155890 Chr.20: 56412450-56412450 on Build GRCh38
rs1002017792 Chr.20: 56412552-56412552 on Build GRCh38
rs202007264 Chr.20: 56412432-56412432 on Build GRCh38
rs1046975336 Chr.20: 56412494-56412494 on Build GRCh38
rs989021212 Chr.20: 56412369-56412369 on Build GRCh38
rs750395450 Chr.20: 56412415-56412415 on Build GRCh38
rs778196586 Chr.20: 56412448-56412448 on Build GRCh38
rs773256357 Chr.20: 56412477-56412477 on Build GRCh38
rs752109065 Chr.20: 56412436-56412436 on Build GRCh38
rs754634575 Chr.20: 56412431-56412431 on Build GRCh38
rs759381098 Chr.20: 56412509-56412509 on Build GRCh38
rs374168804 Chr.20: 56412487-56412487 on Build GRCh38
rs767037963 Chr.20: 56412514-56412514 on Build GRCh38
rs369235380 Chr.20: 56412422-56412422 on Build GRCh38
rs1020414005 Chr.20: 56412378-56412378 on Build GRCh38
rs747015969 Chr.20: 56412473-56412473 on Build GRCh38
rs746086661 Chr.20: 56412459-56412459 on Build GRCh38
rs916718022 Chr.20: 56412401-56412401 on Build GRCh38
rs772124079 Chr.20: 56412464-56412464 on Build GRCh38

View Full Product Details