Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230843608-230843879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf198
Entrez Gene ID
84886
Gene Name
chromosome 1 open reading frame 198
Gene Aliases
-
UniGene
Hs.520494
Gene Chromosome Location
Chr.1: 230837119-230869589 on Build GRCh38
Transcripts
NM_001136494.1 NM_001136495.1 NM_032800.2 XM_017002599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4029399 c.538G>T p.G180C Chr.1: 230843743-230843743 on Build GRCh38
1238358 c.590G>A p.R197Q Chr.1: 230843691-230843691 on Build GRCh38
4029398 c.564G>A p.A188A Chr.1: 230843717-230843717 on Build GRCh38
5779631 c.478C>T p.Q160* Chr.1: 230843803-230843803 on Build GRCh38
1501816 c.409G>A p.A137T Chr.1: 230843872-230843872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777302924 Chr.1: 230843835-230843835 on Build GRCh38
rs757027201 Chr.1: 230843756-230843756 on Build GRCh38
rs746984139 Chr.1: 230843852-230843852 on Build GRCh38
rs554195668 Chr.1: 230843853-230843853 on Build GRCh38
rs116129449 Chr.1: 230843690-230843690 on Build GRCh38
rs564936718 Chr.1: 230843686-230843686 on Build GRCh38
rs190484507 Chr.1: 230843718-230843718 on Build GRCh38
rs759429065 Chr.1: 230843692-230843692 on Build GRCh38
rs369794287 Chr.1: 230843841-230843841 on Build GRCh38
rs758911473 Chr.1: 230843666-230843666 on Build GRCh38
rs200717843 Chr.1: 230843765-230843765 on Build GRCh38
rs182151613 Chr.1: 230843766-230843766 on Build GRCh38
rs115333854 Chr.1: 230843786-230843786 on Build GRCh38
rs752204291 Chr.1: 230843760-230843760 on Build GRCh38
rs758890119 Chr.1: 230843755-230843755 on Build GRCh38
rs541415344 Chr.1: 230843654-230843654 on Build GRCh38
rs752009848 Chr.1: 230843830-230843830 on Build GRCh38
rs571278510 Chr.1: 230843845-230843845 on Build GRCh38
rs138642377 Chr.1: 230843783-230843783 on Build GRCh38
rs766899398 Chr.1: 230843825-230843825 on Build GRCh38
rs759681546 Chr.1: 230843628-230843628 on Build GRCh38
rs370293584 Chr.1: 230843750-230843750 on Build GRCh38
rs115242372 Chr.1: 230843846-230843846 on Build GRCh38
rs772780914 Chr.1: 230843677-230843677 on Build GRCh38
rs779974069 Chr.1: 230843850-230843850 on Build GRCh38
rs1022843803 Chr.1: 230843800-230843800 on Build GRCh38
rs764442305 Chr.1: 230843747-230843747 on Build GRCh38
rs549059928 Chr.1: 230843778-230843778 on Build GRCh38
rs150666760 Chr.1: 230843814-230843814 on Build GRCh38
rs776743827 Chr.1: 230843806-230843806 on Build GRCh38
rs61734648 Chr.1: 230843748-230843748 on Build GRCh38
rs753381372 Chr.1: 230843836-230843836 on Build GRCh38
rs151055746 Chr.1: 230843655-230843655 on Build GRCh38
rs185973061 Chr.1: 230843831-230843831 on Build GRCh38
rs142860003 Chr.1: 230843633-230843633 on Build GRCh38
rs779498094 Chr.1: 230843659-230843659 on Build GRCh38
rs375002821 Chr.1: 230843763-230843763 on Build GRCh38
rs767556367 Chr.1: 230843710-230843710 on Build GRCh38
rs116084579 Chr.1: 230843676-230843676 on Build GRCh38
rs750799062 Chr.1: 230843753-230843753 on Build GRCh38
rs775769596 Chr.1: 230843717-230843717 on Build GRCh38
rs780164639 Chr.1: 230843840-230843840 on Build GRCh38
rs762010027 Chr.1: 230843811-230843811 on Build GRCh38
rs768907418 Chr.1: 230843804-230843804 on Build GRCh38
rs150339183 Chr.1: 230843669-230843669 on Build GRCh38
rs922152010 Chr.1: 230843742-230843742 on Build GRCh38
rs114652630 Chr.1: 230843818-230843818 on Build GRCh38
rs746413033 Chr.1: 230843665-230843665 on Build GRCh38
rs778638875 Chr.1: 230843770-230843770 on Build GRCh38
rs375540477 Chr.1: 230843749-230843749 on Build GRCh38
rs767541103 Chr.1: 230843629-230843629 on Build GRCh38
rs760861127 Chr.1: 230843803-230843803 on Build GRCh38
rs749135657 Chr.1: 230843685-230843685 on Build GRCh38
rs143886432 Chr.1: 230843817-230843817 on Build GRCh38
rs977960521 Chr.1: 230843828-230843828 on Build GRCh38
rs756917091 Chr.1: 230843839-230843839 on Build GRCh38

View Full Product Details