Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96197007-96197280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLNK
Entrez Gene ID
29760
Gene Name
B-cell linker
Gene Aliases
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca
UniGene
Hs.665244
Gene Chromosome Location
Chr.10: 96189171-96271576 on Build GRCh38
Transcripts
NM_001114094.1 NM_001258440.1 NM_001258441.1 NM_001258442.1 NM_013314.3 NR_047680.1 NR_047681.1 NR_047682.1 NR_047683.1 XM_011539728.2 XM_017016159.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4016966 c.1115G>A p.R372Q Chr.10: 96197044-96197044 on Build GRCh38
5946426 c.1096-3C>G p.? Chr.10: 96197066-96197066 on Build GRCh38
6413338 c.1109T>C p.L370P Chr.10: 96197050-96197050 on Build GRCh38
4902404 c.1122C>T p.S374S Chr.10: 96197037-96197037 on Build GRCh38
4901884 c.1114C>T p.R372W Chr.10: 96197045-96197045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587742115 Chr.10: 96197212-96197212 on Build GRCh38
rs782452195 Chr.10: 96197151-96197151 on Build GRCh38
rs782349549 Chr.10: 96197069-96197069 on Build GRCh38
rs782566012 Chr.10: 96197045-96197045 on Build GRCh38
rs782160429 Chr.10: 96197101-96197101 on Build GRCh38
rs142226223 Chr.10: 96197206-96197206 on Build GRCh38
rs74151283 Chr.10: 96197074-96197074 on Build GRCh38
rs80026049 Chr.10: 96197079-96197079 on Build GRCh38
rs10491068 Chr.10: 96197146-96197146 on Build GRCh38
rs782018939 Chr.10: 96197093-96197093 on Build GRCh38
rs782570122 Chr.10: 96197044-96197044 on Build GRCh38
rs199983704 Chr.10: 96197080-96197080 on Build GRCh38
rs1019427725 Chr.10: 96197167-96197167 on Build GRCh38
rs781837707 Chr.10: 96197210-96197210 on Build GRCh38
rs782205113 Chr.10: 96197061-96197061 on Build GRCh38
rs199715227 Chr.10: 96197112-96197112 on Build GRCh38
rs956922601 Chr.10: 96197191-96197191 on Build GRCh38
rs782197882 Chr.10: 96197097-96197097 on Build GRCh38
rs895691306 Chr.10: 96197053-96197053 on Build GRCh38
rs782635269 Chr.10: 96197037-96197037 on Build GRCh38
rs781823661 Chr.10: 96197071-96197072 on Build GRCh38
rs782480599 Chr.10: 96197072-96197072 on Build GRCh38
rs782248760 Chr.10: 96197038-96197038 on Build GRCh38
rs113051625 Chr.10: 96197155-96197155 on Build GRCh38
rs201454460 Chr.10: 96197080-96197080 on Build GRCh38

View Full Product Details