Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83970548-83970803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPK
Entrez Gene ID
3190
Gene Name
heterogeneous nuclear ribonucleoprotein K
Gene Aliases
AUKS, CSBP, HNRPK, TUNP
UniGene
Hs.522257
Gene Chromosome Location
Chr.9: 83968083-83980782 on Build GRCh38
Transcripts
NM_001318186.1 NM_001318187.1 NM_001318188.1 NM_002140.4 NM_031262.3 NM_031263.3 XM_005251960.2 XM_005251963.3 XM_005251965.2 XM_011518616.1 XM_017014668.1 XM_017014669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3703582 c.1163T>C p.I388T Chr.9: 83970765-83970765 on Build GRCh38
5816466 c.1154G>T p.G385V Chr.9: 83970774-83970774 on Build GRCh38
6268154 c.1129C>A p.R377S Chr.9: 83970799-83970799 on Build GRCh38
6268155 c.1129C>A p.R377S Chr.9: 83970799-83970799 on Build GRCh38
5816465 c.1154G>T p.G385V Chr.9: 83970774-83970774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980772629 Chr.9: 83970680-83970680 on Build GRCh38
rs1025342664 Chr.9: 83970600-83970600 on Build GRCh38
rs184361719 Chr.9: 83970621-83970621 on Build GRCh38
rs535426539 Chr.9: 83970673-83970673 on Build GRCh38
rs781177925 Chr.9: 83970714-83970714 on Build GRCh38
rs755597367 Chr.9: 83970716-83970716 on Build GRCh38
rs754814694 Chr.9: 83970710-83970710 on Build GRCh38
rs942829364 Chr.9: 83970781-83970781 on Build GRCh38
rs1034339468 Chr.9: 83970681-83970681 on Build GRCh38
rs995579822 Chr.9: 83970585-83970585 on Build GRCh38
rs774517631 Chr.9: 83970768-83970768 on Build GRCh38
rs1049381888 Chr.9: 83970583-83970583 on Build GRCh38
rs777424249 Chr.9: 83970717-83970717 on Build GRCh38
rs397687683 Chr.9: 83970646-83970647 on Build GRCh38
rs751523268 Chr.9: 83970709-83970709 on Build GRCh38
rs752617416 Chr.9: 83970582-83970583 on Build GRCh38
rs1010396279 Chr.9: 83970638-83970638 on Build GRCh38
rs762893180 Chr.9: 83970718-83970723 on Build GRCh38
rs751422995 Chr.9: 83970719-83970719 on Build GRCh38
rs779676856 Chr.9: 83970703-83970703 on Build GRCh38
rs889464020 Chr.9: 83970597-83970597 on Build GRCh38
rs747681079 Chr.9: 83970715-83970715 on Build GRCh38
rs758293796 Chr.9: 83970702-83970702 on Build GRCh38
rs371386785 Chr.9: 83970728-83970728 on Build GRCh38
rs140741394 Chr.9: 83970668-83970668 on Build GRCh38
rs971594875 Chr.9: 83970674-83970674 on Build GRCh38
rs771289643 Chr.9: 83970745-83970745 on Build GRCh38
rs537862835 Chr.9: 83970617-83970617 on Build GRCh38
rs748898288 Chr.9: 83970720-83970720 on Build GRCh38
rs760830318 Chr.9: 83970686-83970686 on Build GRCh38
rs3214660 Chr.9: 83970643-83970644 on Build GRCh38
rs188765983 Chr.9: 83970697-83970697 on Build GRCh38
rs767805806 Chr.9: 83970723-83970724 on Build GRCh38
rs750677424 Chr.9: 83970724-83970725 on Build GRCh38
rs981343438 Chr.9: 83970610-83970610 on Build GRCh38
rs78280596 Chr.9: 83970607-83970607 on Build GRCh38
rs552255559 Chr.9: 83970589-83970589 on Build GRCh38
rs952164009 Chr.9: 83970682-83970682 on Build GRCh38
rs201503271 Chr.9: 83970726-83970726 on Build GRCh38
rs775395655 Chr.9: 83970696-83970697 on Build GRCh38
rs759644134 Chr.9: 83970774-83970776 on Build GRCh38
rs763018954 Chr.9: 83970634-83970634 on Build GRCh38

View Full Product Details