Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168901203-168901434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC2
Entrez Gene ID
57818
Gene Name
glucose-6-phosphatase catalytic subunit 2
Gene Aliases
IGRP
UniGene
Hs.283963
Gene Chromosome Location
Chr.2: 168901223-168910000 on Build GRCh38
Transcripts
NM_001081686.1 NM_021176.2 XM_011511564.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
441434 c.84T>A p.F28L Chr.2: 168901415-168901415 on Build GRCh38
4532621 c.18G>A p.R6R Chr.2: 168901349-168901349 on Build GRCh38
4935043 c.33A>C p.I11I Chr.2: 168901364-168901364 on Build GRCh38
1691347 c.17G>A p.R6K Chr.2: 168901348-168901348 on Build GRCh38
1009099 c.81T>G p.N27K Chr.2: 168901412-168901412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030771046 Chr.2: 168901274-168901274 on Build GRCh38
rs368619514 Chr.2: 168901283-168901283 on Build GRCh38
rs775748050 Chr.2: 168901307-168901307 on Build GRCh38
rs999742726 Chr.2: 168901241-168901241 on Build GRCh38
rs747911600 Chr.2: 168901374-168901374 on Build GRCh38
rs754146984 Chr.2: 168901312-168901312 on Build GRCh38
rs965878348 Chr.2: 168901275-168901275 on Build GRCh38
rs764510066 Chr.2: 168901311-168901311 on Build GRCh38
rs746671578 Chr.2: 168901371-168901371 on Build GRCh38
rs761008499 Chr.2: 168901308-168901308 on Build GRCh38
rs1006880618 Chr.2: 168901287-168901287 on Build GRCh38
rs143670077 Chr.2: 168901390-168901390 on Build GRCh38
rs779614984 Chr.2: 168901361-168901361 on Build GRCh38
rs757770113 Chr.2: 168901316-168901316 on Build GRCh38
rs769873771 Chr.2: 168901378-168901378 on Build GRCh38
rs774449598 Chr.2: 168901333-168901333 on Build GRCh38
rs757904389 Chr.2: 168901352-168901352 on Build GRCh38
rs368382511 Chr.2: 168901354-168901354 on Build GRCh38
rs767677848 Chr.2: 168901306-168901306 on Build GRCh38
rs577033273 Chr.2: 168901392-168901392 on Build GRCh38
rs781044423 Chr.2: 168901372-168901372 on Build GRCh38
rs748108026 Chr.2: 168901282-168901282 on Build GRCh38
rs759636288 Chr.2: 168901297-168901297 on Build GRCh38
rs774603818 Chr.2: 168901284-168901284 on Build GRCh38
rs768169038 Chr.2: 168901378-168901380 on Build GRCh38
rs903949552 Chr.2: 168901225-168901225 on Build GRCh38
rs765696432 Chr.2: 168901327-168901327 on Build GRCh38
rs759777262 Chr.2: 168901385-168901385 on Build GRCh38
rs745981571 Chr.2: 168901382-168901382 on Build GRCh38
rs536809298 Chr.2: 168901271-168901271 on Build GRCh38
rs372008743 Chr.2: 168901379-168901379 on Build GRCh38
rs891163703 Chr.2: 168901276-168901276 on Build GRCh38
rs772185727 Chr.2: 168901383-168901383 on Build GRCh38

View Full Product Details