Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42184155-42184404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCRT
Entrez Gene ID
3060
Gene Name
hypocretin neuropeptide precursor
Gene Aliases
NRCLP1, OX, PPOX
UniGene
Hs.158348
Gene Chromosome Location
Chr.17: 42184060-42185452 on Build GRCh38
Transcripts
NM_001524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
399745 c.298C>G p.R100G Chr.17: 42184252-42184252 on Build GRCh38
4420226 c.200G>T p.G67V Chr.17: 42184350-42184350 on Build GRCh38
4453245 c.269A>G p.H90R Chr.17: 42184281-42184281 on Build GRCh38
7318114 c.376G>A p.G126R Chr.17: 42184174-42184174 on Build GRCh38
4641102 c.262G>A p.G88S Chr.17: 42184288-42184288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779731661 Chr.17: 42184219-42184251 on Build GRCh38
rs1012290621 Chr.17: 42184179-42184179 on Build GRCh38
rs866782848 Chr.17: 42184274-42184274 on Build GRCh38
rs780290109 Chr.17: 42184325-42184325 on Build GRCh38
rs570258880 Chr.17: 42184222-42184222 on Build GRCh38
rs537012902 Chr.17: 42184292-42184292 on Build GRCh38
rs903829881 Chr.17: 42184281-42184281 on Build GRCh38
rs768600704 Chr.17: 42184227-42184227 on Build GRCh38
rs754648418 Chr.17: 42184332-42184332 on Build GRCh38
rs769028179 Chr.17: 42184366-42184366 on Build GRCh38
rs536458711 Chr.17: 42184208-42184208 on Build GRCh38
rs1007210598 Chr.17: 42184214-42184214 on Build GRCh38
rs534685387 Chr.17: 42184370-42184370 on Build GRCh38
rs777822783 Chr.17: 42184336-42184336 on Build GRCh38
rs537376938 Chr.17: 42184347-42184347 on Build GRCh38
rs1024019238 Chr.17: 42184193-42184193 on Build GRCh38
rs763215233 Chr.17: 42184354-42184354 on Build GRCh38
rs771882989 Chr.17: 42184334-42184334 on Build GRCh38
rs760566371 Chr.17: 42184176-42184176 on Build GRCh38
rs778354341 Chr.17: 42184333-42184333 on Build GRCh38
rs764209002 Chr.17: 42184224-42184224 on Build GRCh38
rs767907538 Chr.17: 42184387-42184387 on Build GRCh38
rs775918962 Chr.17: 42184213-42184213 on Build GRCh38
rs565917657 Chr.17: 42184206-42184206 on Build GRCh38
rs1051504874 Chr.17: 42184210-42184210 on Build GRCh38
rs757797020 Chr.17: 42184280-42184280 on Build GRCh38
rs768106966 Chr.17: 42184288-42184288 on Build GRCh38
rs746600850 Chr.17: 42184329-42184329 on Build GRCh38
rs751706776 Chr.17: 42184266-42184266 on Build GRCh38
rs548838330 Chr.17: 42184216-42184216 on Build GRCh38
rs558579797 Chr.17: 42184317-42184317 on Build GRCh38
rs992913304 Chr.17: 42184252-42184252 on Build GRCh38
rs55787720 Chr.17: 42184378-42184378 on Build GRCh38
rs948271520 Chr.17: 42184265-42184265 on Build GRCh38
rs1000855419 Chr.17: 42184337-42184337 on Build GRCh38
rs918324860 Chr.17: 42184237-42184237 on Build GRCh38
rs753435149 Chr.17: 42184329-42184329 on Build GRCh38
rs896739180 Chr.17: 42184260-42184260 on Build GRCh38
rs770691920 Chr.17: 42184345-42184345 on Build GRCh38
rs960115444 Chr.17: 42184238-42184238 on Build GRCh38
rs762013647 Chr.17: 42184382-42184382 on Build GRCh38
rs934848266 Chr.17: 42184200-42184200 on Build GRCh38
rs765363265 Chr.17: 42184202-42184202 on Build GRCh38
rs1056645139 Chr.17: 42184256-42184256 on Build GRCh38
rs970492685 Chr.17: 42184344-42184344 on Build GRCh38

View Full Product Details