Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 162383630-162383889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf226
Entrez Gene ID
400793
Gene Name
chromosome 1 open reading frame 226
Gene Aliases
-
UniGene
Hs.447011
Gene Chromosome Location
Chr.1: 162378906-162386818 on Build GRCh38
Transcripts
NM_001085375.1 NM_001135240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024509 c.907G>A p.D303N Chr.1: 162383642-162383642 on Build GRCh38
458480 c.906C>G p.L302L Chr.1: 162383641-162383641 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs946628815 Chr.1: 162383817-162383817 on Build GRCh38
rs867306710 Chr.1: 162383673-162383673 on Build GRCh38
rs930873016 Chr.1: 162383773-162383774 on Build GRCh38
rs985080064 Chr.1: 162383808-162383808 on Build GRCh38
rs964913027 Chr.1: 162383744-162383744 on Build GRCh38
rs751729454 Chr.1: 162383717-162383717 on Build GRCh38
rs952356905 Chr.1: 162383650-162383650 on Build GRCh38
rs937875284 Chr.1: 162383837-162383837 on Build GRCh38
rs753262189 Chr.1: 162383663-162383663 on Build GRCh38
rs749504474 Chr.1: 162383730-162383730 on Build GRCh38
rs769982973 Chr.1: 162383724-162383724 on Build GRCh38
rs989182003 Chr.1: 162383759-162383759 on Build GRCh38
rs370587346 Chr.1: 162383685-162383685 on Build GRCh38
rs562005793 Chr.1: 162383745-162383745 on Build GRCh38
rs367877155 Chr.1: 162383657-162383657 on Build GRCh38
rs778099497 Chr.1: 162383728-162383728 on Build GRCh38
rs200770870 Chr.1: 162383684-162383684 on Build GRCh38
rs1006484727 Chr.1: 162383674-162383674 on Build GRCh38
rs368799518 Chr.1: 162383694-162383694 on Build GRCh38
rs756830186 Chr.1: 162383666-162383666 on Build GRCh38
rs748573065 Chr.1: 162383705-162383705 on Build GRCh38
rs536148325 Chr.1: 162383687-162383687 on Build GRCh38
rs908127504 Chr.1: 162383811-162383811 on Build GRCh38
rs140889968 Chr.1: 162383769-162383769 on Build GRCh38
rs956169144 Chr.1: 162383758-162383758 on Build GRCh38
rs760397357 Chr.1: 162383651-162383651 on Build GRCh38
rs926441019 Chr.1: 162383835-162383835 on Build GRCh38
rs544066155 Chr.1: 162383751-162383751 on Build GRCh38
rs979357076 Chr.1: 162383830-162383830 on Build GRCh38

View Full Product Details