Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77433745-77434006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIPAS39
Entrez Gene ID
63894
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
Gene Aliases
C14orf133, SPE-39, SPE39, VIPAR, VPS16B, hSPE-39
UniGene
Hs.16157
Gene Chromosome Location
Chr.14: 77426675-77457640 on Build GRCh38
Transcripts
NM_001193314.1 NM_001193315.1 NM_001193316.1 NM_001193317.1 NM_022067.3 XM_011537066.1 XM_017021579.1 XM_017021580.1 XM_017021581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1232274 c.1141C>T p.R381* Chr.14: 77433880-77433880 on Build GRCh38
6318843 c.1145C>A p.A382D Chr.14: 77433876-77433876 on Build GRCh38
3815401 c.1113G>A p.L371L Chr.14: 77433908-77433908 on Build GRCh38
6001322 c.1090-10T>C p.? Chr.14: 77433941-77433941 on Build GRCh38
4052612 c.1143A>G p.R381R Chr.14: 77433878-77433878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756381649 Chr.14: 77433839-77433839 on Build GRCh38
rs940334505 Chr.14: 77433870-77433870 on Build GRCh38
rs944573317 Chr.14: 77433780-77433780 on Build GRCh38
rs762568295 Chr.14: 77433931-77433931 on Build GRCh38
rs774940891 Chr.14: 77433923-77433923 on Build GRCh38
rs146288421 Chr.14: 77433829-77433829 on Build GRCh38
rs767308056 Chr.14: 77433961-77433961 on Build GRCh38
rs772462216 Chr.14: 77433878-77433878 on Build GRCh38
rs575404280 Chr.14: 77433781-77433781 on Build GRCh38
rs761811943 Chr.14: 77433957-77433957 on Build GRCh38
rs34835605 Chr.14: 77433822-77433822 on Build GRCh38
rs753955179 Chr.14: 77433847-77433847 on Build GRCh38
rs375310819 Chr.14: 77433795-77433795 on Build GRCh38
rs1037306281 Chr.14: 77433879-77433879 on Build GRCh38
rs754164698 Chr.14: 77433926-77433926 on Build GRCh38
rs78546415 Chr.14: 77433966-77433966 on Build GRCh38
rs544468801 Chr.14: 77433785-77433785 on Build GRCh38
rs930418798 Chr.14: 77433811-77433811 on Build GRCh38
rs148380631 Chr.14: 77433851-77433851 on Build GRCh38
rs367944244 Chr.14: 77433956-77433956 on Build GRCh38
rs372598184 Chr.14: 77433948-77433948 on Build GRCh38
rs769521365 Chr.14: 77433914-77433914 on Build GRCh38
rs886043868 Chr.14: 77433857-77433857 on Build GRCh38
rs764577623 Chr.14: 77433854-77433854 on Build GRCh38
rs978317267 Chr.14: 77433824-77433824 on Build GRCh38
rs907154954 Chr.14: 77433935-77433935 on Build GRCh38
rs747290213 Chr.14: 77433897-77433897 on Build GRCh38
rs377341681 Chr.14: 77433791-77433791 on Build GRCh38
rs778181495 Chr.14: 77433880-77433880 on Build GRCh38
rs200692541 Chr.14: 77433868-77433868 on Build GRCh38
rs780098749 Chr.14: 77433843-77433843 on Build GRCh38
rs369962683 Chr.14: 77433836-77433836 on Build GRCh38
rs147548479 Chr.14: 77433891-77433891 on Build GRCh38
rs375374499 Chr.14: 77433833-77433833 on Build GRCh38
rs748646553 Chr.14: 77433872-77433872 on Build GRCh38
rs1014430582 Chr.14: 77433912-77433912 on Build GRCh38
rs924088762 Chr.14: 77433825-77433825 on Build GRCh38

View Full Product Details