Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8539748-8540003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4E
Entrez Gene ID
26253
Gene Name
C-type lectin domain family 4 member E
Gene Aliases
CLECSF9, MINCLE
UniGene
Hs.236516
Gene Chromosome Location
Chr.12: 8533305-8541328 on Build GRCh38
Transcripts
NM_014358.3 XM_011520614.2 XM_011520615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3676542 c.130+4G>T p.? Chr.12: 8539851-8539851 on Build GRCh38
3872662 c.56C>T p.S19F Chr.12: 8539929-8539929 on Build GRCh38
3872663 c.41G>A p.R14K Chr.12: 8539944-8539944 on Build GRCh38
944175 c.81T>G p.A27A Chr.12: 8539904-8539904 on Build GRCh38
6630188 c.100C>A p.L34I Chr.12: 8539885-8539885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758785904 Chr.12: 8539831-8539831 on Build GRCh38
rs757701740 Chr.12: 8539815-8539815 on Build GRCh38
rs752154371 Chr.12: 8539905-8539905 on Build GRCh38
rs1049645947 Chr.12: 8539923-8539923 on Build GRCh38
rs375495869 Chr.12: 8539951-8539951 on Build GRCh38
rs930658697 Chr.12: 8539908-8539909 on Build GRCh38
rs941637673 Chr.12: 8539775-8539775 on Build GRCh38
rs1018548251 Chr.12: 8539906-8539906 on Build GRCh38
rs979591078 Chr.12: 8539785-8539785 on Build GRCh38
rs368572057 Chr.12: 8539902-8539902 on Build GRCh38
rs367915252 Chr.12: 8539821-8539821 on Build GRCh38
rs750918854 Chr.12: 8539925-8539925 on Build GRCh38
rs749205687 Chr.12: 8539975-8539975 on Build GRCh38
rs1001076714 Chr.12: 8539920-8539920 on Build GRCh38
rs111994062 Chr.12: 8539839-8539839 on Build GRCh38
rs200497205 Chr.12: 8539971-8539971 on Build GRCh38
rs768901460 Chr.12: 8539846-8539846 on Build GRCh38
rs776936054 Chr.12: 8539976-8539976 on Build GRCh38
rs748180276 Chr.12: 8539857-8539857 on Build GRCh38
rs950318330 Chr.12: 8539870-8539870 on Build GRCh38
rs751884084 Chr.12: 8539938-8539938 on Build GRCh38
rs779412850 Chr.12: 8539817-8539817 on Build GRCh38
rs781405986 Chr.12: 8539947-8539947 on Build GRCh38
rs754427678 Chr.12: 8539812-8539812 on Build GRCh38
rs780454698 Chr.12: 8539835-8539835 on Build GRCh38
rs897389307 Chr.12: 8539778-8539778 on Build GRCh38
rs766991792 Chr.12: 8539804-8539804 on Build GRCh38
rs777615768 Chr.12: 8539974-8539974 on Build GRCh38
rs747047142 Chr.12: 8539958-8539965 on Build GRCh38
rs774039548 Chr.12: 8539872-8539872 on Build GRCh38
rs780538996 Chr.12: 8539937-8539937 on Build GRCh38
rs780305512 Chr.12: 8539883-8539884 on Build GRCh38
rs781115660 Chr.12: 8539856-8539856 on Build GRCh38
rs113313978 Chr.12: 8539783-8539783 on Build GRCh38
rs199850652 Chr.12: 8539914-8539914 on Build GRCh38
rs374178171 Chr.12: 8539777-8539777 on Build GRCh38
rs759176054 Chr.12: 8539874-8539874 on Build GRCh38
rs769005327 Chr.12: 8539966-8539966 on Build GRCh38
rs1044592923 Chr.12: 8539876-8539876 on Build GRCh38
rs769634954 Chr.12: 8539862-8539862 on Build GRCh38
rs773155374 Chr.12: 8539864-8539864 on Build GRCh38
rs865955583 Chr.12: 8539901-8539901 on Build GRCh38
rs763938533 Chr.12: 8539866-8539866 on Build GRCh38
rs760171934 Chr.12: 8539910-8539910 on Build GRCh38
rs770686515 Chr.12: 8539869-8539869 on Build GRCh38
rs758873907 Chr.12: 8539932-8539932 on Build GRCh38
rs755243353 Chr.12: 8539946-8539946 on Build GRCh38

View Full Product Details