Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7136163-7136434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN3
Entrez Gene ID
9746
Gene Name
calsyntenin 3
Gene Aliases
CDHR14, CSTN3, alcbeta
UniGene
Hs.535378
Gene Chromosome Location
Chr.12: 7130371-7158945 on Build GRCh38
Transcripts
NM_014718.3 XM_006719163.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7239100 c.815C>T p.T272I Chr.12: 7136278-7136278 on Build GRCh38
943341 c.790T>G p.L264V Chr.12: 7136253-7136253 on Build GRCh38
3465101 c.759C>T p.I253I Chr.12: 7136222-7136222 on Build GRCh38
3730637 c.743-8C>A p.? Chr.12: 7136198-7136198 on Build GRCh38
3465102 c.795C>T p.F265F Chr.12: 7136258-7136258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766305766 Chr.12: 7136333-7136333 on Build GRCh38
rs769934980 Chr.12: 7136301-7136301 on Build GRCh38
rs371197435 Chr.12: 7136340-7136340 on Build GRCh38
rs774058767 Chr.12: 7136337-7136337 on Build GRCh38
rs150654430 Chr.12: 7136405-7136405 on Build GRCh38
rs773410195 Chr.12: 7136314-7136314 on Build GRCh38
rs772924137 Chr.12: 7136186-7136186 on Build GRCh38
rs751900783 Chr.12: 7136395-7136395 on Build GRCh38
rs775797020 Chr.12: 7136204-7136204 on Build GRCh38
rs778295167 Chr.12: 7136250-7136250 on Build GRCh38
rs975425345 Chr.12: 7136400-7136400 on Build GRCh38
rs779399453 Chr.12: 7136258-7136258 on Build GRCh38
rs377471817 Chr.12: 7136315-7136315 on Build GRCh38
rs765296417 Chr.12: 7136232-7136232 on Build GRCh38
rs905722256 Chr.12: 7136282-7136282 on Build GRCh38
rs577243310 Chr.12: 7136218-7136218 on Build GRCh38
rs747369055 Chr.12: 7136279-7136279 on Build GRCh38
rs373671772 Chr.12: 7136195-7136195 on Build GRCh38
rs923016165 Chr.12: 7136412-7136412 on Build GRCh38
rs150491757 Chr.12: 7136374-7136374 on Build GRCh38
rs758870677 Chr.12: 7136375-7136375 on Build GRCh38
rs774579276 Chr.12: 7136213-7136213 on Build GRCh38
rs768782294 Chr.12: 7136284-7136284 on Build GRCh38
rs767109868 Chr.12: 7136343-7136343 on Build GRCh38
rs748437288 Chr.12: 7136404-7136404 on Build GRCh38
rs189282788 Chr.12: 7136245-7136245 on Build GRCh38
rs751947010 Chr.12: 7136265-7136265 on Build GRCh38
rs780588881 Chr.12: 7136380-7136380 on Build GRCh38
rs747255044 Chr.12: 7136206-7136206 on Build GRCh38
rs746265549 Chr.12: 7136187-7136187 on Build GRCh38
rs370896693 Chr.12: 7136409-7136409 on Build GRCh38
rs746318546 Chr.12: 7136259-7136259 on Build GRCh38
rs752282204 Chr.12: 7136370-7136370 on Build GRCh38
rs375184032 Chr.12: 7136228-7136228 on Build GRCh38
rs768043934 Chr.12: 7136241-7136241 on Build GRCh38
rs773215598 Chr.12: 7136233-7136233 on Build GRCh38
rs754622582 Chr.12: 7136246-7136246 on Build GRCh38
rs776952469 Chr.12: 7136285-7136285 on Build GRCh38
rs374604787 Chr.12: 7136396-7136396 on Build GRCh38
rs201295647 Chr.12: 7136269-7136269 on Build GRCh38
rs763654214 Chr.12: 7136371-7136371 on Build GRCh38
rs994582275 Chr.12: 7136300-7136300 on Build GRCh38
rs537697520 Chr.12: 7136240-7136240 on Build GRCh38
rs748241137 Chr.12: 7136294-7136294 on Build GRCh38
rs752230806 Chr.12: 7136251-7136251 on Build GRCh38
rs781577112 Chr.12: 7136403-7136403 on Build GRCh38

View Full Product Details