Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63236236-63236509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC7
Entrez Gene ID
79444
Gene Name
baculoviral IAP repeat containing 7
Gene Aliases
KIAP, LIVIN, ML-IAP, MLIAP, RNF50
UniGene
Hs.256126
Gene Chromosome Location
Chr.20: 63235883-63240507 on Build GRCh38
Transcripts
NM_022161.3 NM_139317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6160825 c.169G>T p.G57C Chr.20: 63236265-63236265 on Build GRCh38
1283657 c.306G>C p.E102D Chr.20: 63236402-63236402 on Build GRCh38
6247044 c.154G>T p.D52Y Chr.20: 63236250-63236250 on Build GRCh38
1190315 c.224T>C p.L75S Chr.20: 63236320-63236320 on Build GRCh38
1184744 c.211G>A p.A71T Chr.20: 63236307-63236307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047375772 Chr.20: 63236262-63236262 on Build GRCh38
rs780891467 Chr.20: 63236426-63236426 on Build GRCh38
rs780013871 Chr.20: 63236445-63236445 on Build GRCh38
rs754184374 Chr.20: 63236316-63236316 on Build GRCh38
rs149322415 Chr.20: 63236430-63236430 on Build GRCh38
rs762822972 Chr.20: 63236326-63236326 on Build GRCh38
rs776902171 Chr.20: 63236454-63236454 on Build GRCh38
rs771175273 Chr.20: 63236325-63236325 on Build GRCh38
rs1024741398 Chr.20: 63236464-63236464 on Build GRCh38
rs138408753 Chr.20: 63236317-63236317 on Build GRCh38
rs760830194 Chr.20: 63236389-63236389 on Build GRCh38
rs41282996 Chr.20: 63236274-63236274 on Build GRCh38
rs537991526 Chr.20: 63236390-63236390 on Build GRCh38
rs759072139 Chr.20: 63236460-63236460 on Build GRCh38
rs199560863 Chr.20: 63236429-63236429 on Build GRCh38
rs769686006 Chr.20: 63236363-63236363 on Build GRCh38
rs199810069 Chr.20: 63236347-63236347 on Build GRCh38
rs769713600 Chr.20: 63236300-63236300 on Build GRCh38
rs768831128 Chr.20: 63236451-63236451 on Build GRCh38
rs573942684 Chr.20: 63236458-63236458 on Build GRCh38
rs758054106 Chr.20: 63236278-63236278 on Build GRCh38
rs765669557 Chr.20: 63236321-63236321 on Build GRCh38
rs564140632 Chr.20: 63236325-63236326 on Build GRCh38
rs760696812 Chr.20: 63236263-63236264 on Build GRCh38
rs751253584 Chr.20: 63236342-63236342 on Build GRCh38
rs775833040 Chr.20: 63236309-63236309 on Build GRCh38
rs375971878 Chr.20: 63236295-63236295 on Build GRCh38
rs761142703 Chr.20: 63236310-63236310 on Build GRCh38
rs6011692 Chr.20: 63236476-63236476 on Build GRCh38
rs763786055 Chr.20: 63236443-63236445 on Build GRCh38
rs143928099 Chr.20: 63236343-63236343 on Build GRCh38
rs150688713 Chr.20: 63236459-63236459 on Build GRCh38
rs200751512 Chr.20: 63236364-63236364 on Build GRCh38
rs779681359 Chr.20: 63236284-63236284 on Build GRCh38
rs367825889 Chr.20: 63236411-63236411 on Build GRCh38
rs770943952 Chr.20: 63236443-63236443 on Build GRCh38
rs11537908 Chr.20: 63236267-63236267 on Build GRCh38
rs762235988 Chr.20: 63236396-63236396 on Build GRCh38
rs748514539 Chr.20: 63236455-63236455 on Build GRCh38
rs201104335 Chr.20: 63236307-63236307 on Build GRCh38
rs139712930 Chr.20: 63236365-63236365 on Build GRCh38
rs763405587 Chr.20: 63236403-63236403 on Build GRCh38
rs747046370 Chr.20: 63236450-63236450 on Build GRCh38
rs754589649 Chr.20: 63236412-63236412 on Build GRCh38
rs767123998 Chr.20: 63236463-63236463 on Build GRCh38
rs761627866 Chr.20: 63236459-63236459 on Build GRCh38
rs746868416 Chr.20: 63236289-63236289 on Build GRCh38
rs567181959 Chr.20: 63236328-63236328 on Build GRCh38
rs778417811 Chr.20: 63236275-63236275 on Build GRCh38
rs752491653 Chr.20: 63236428-63236428 on Build GRCh38
rs867604358 Chr.20: 63236435-63236435 on Build GRCh38
rs556501184 Chr.20: 63236409-63236409 on Build GRCh38
rs747118580 Chr.20: 63236480-63236480 on Build GRCh38
rs371634897 Chr.20: 63236420-63236420 on Build GRCh38
rs770171583 Chr.20: 63236456-63236456 on Build GRCh38
rs754269520 Chr.20: 63236376-63236376 on Build GRCh38
rs750005956 Chr.20: 63236323-63236323 on Build GRCh38
rs778907377 Chr.20: 63236479-63236479 on Build GRCh38
rs748054619 Chr.20: 63236298-63236298 on Build GRCh38
rs781313143 Chr.20: 63236489-63236489 on Build GRCh38
rs764004708 Chr.20: 63236474-63236474 on Build GRCh38
rs866304406 Chr.20: 63236344-63236344 on Build GRCh38
rs751585143 Chr.20: 63236458-63236459 on Build GRCh38
rs776444154 Chr.20: 63236296-63236296 on Build GRCh38
rs968190374 Chr.20: 63236392-63236392 on Build GRCh38
rs878882165 Chr.20: 63236261-63236261 on Build GRCh38
rs768301686 Chr.20: 63236292-63236292 on Build GRCh38
rs765794477 Chr.20: 63236402-63236402 on Build GRCh38
rs755788876 Chr.20: 63236472-63236472 on Build GRCh38
rs764398536 Chr.20: 63236313-63236313 on Build GRCh38
rs371997368 Chr.20: 63236475-63236475 on Build GRCh38
rs150610779 Chr.20: 63236357-63236357 on Build GRCh38
rs375475485 Chr.20: 63236372-63236372 on Build GRCh38
rs369947625 Chr.20: 63236485-63236485 on Build GRCh38
rs202073014 Chr.20: 63236465-63236465 on Build GRCh38
rs542383301 Chr.20: 63236490-63236490 on Build GRCh38
rs371347283 Chr.20: 63236306-63236306 on Build GRCh38
rs780860122 Chr.20: 63236351-63236351 on Build GRCh38

View Full Product Details