Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136241479-136241730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTFR2
Entrez Gene ID
113115
Gene Name
mitochondrial fission regulator 2
Gene Aliases
DUFD1, FAM54A
UniGene
Hs.121536
Gene Chromosome Location
Chr.6: 136231030-136253659 on Build GRCh38
Transcripts
NM_001099286.2 NM_001318738.1 NM_138419.4 XM_011535410.2 XM_011535412.2 XM_011535413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5357076 c.282-10delT p.? Chr.6: 136241686-136241686 on Build GRCh38
4928467 c.465G>C p.Q155H Chr.6: 136241493-136241493 on Build GRCh38
6716324 c.287G>A p.S96N Chr.6: 136241671-136241671 on Build GRCh38
3994612 c.335G>A p.R112Q Chr.6: 136241623-136241623 on Build GRCh38
370218 c.458G>T p.R153L Chr.6: 136241500-136241500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367891434 Chr.6: 136241668-136241668 on Build GRCh38
rs936073134 Chr.6: 136241573-136241573 on Build GRCh38
rs370445232 Chr.6: 136241694-136241694 on Build GRCh38
rs746485124 Chr.6: 136241685-136241685 on Build GRCh38
rs752118648 Chr.6: 136241685-136241686 on Build GRCh38
rs370847362 Chr.6: 136241679-136241679 on Build GRCh38
rs372722168 Chr.6: 136241674-136241674 on Build GRCh38
rs777828744 Chr.6: 136241564-136241564 on Build GRCh38
rs778379033 Chr.6: 136241581-136241581 on Build GRCh38
rs150447506 Chr.6: 136241623-136241623 on Build GRCh38
rs764135332 Chr.6: 136241686-136241686 on Build GRCh38
rs773807924 Chr.6: 136241619-136241619 on Build GRCh38
rs758292533 Chr.6: 136241591-136241591 on Build GRCh38
rs368008648 Chr.6: 136241546-136241546 on Build GRCh38
rs759758528 Chr.6: 136241638-136241638 on Build GRCh38
rs758109914 Chr.6: 136241684-136241684 on Build GRCh38
rs151177675 Chr.6: 136241501-136241501 on Build GRCh38
rs763133075 Chr.6: 136241574-136241575 on Build GRCh38
rs544618242 Chr.6: 136241618-136241618 on Build GRCh38
rs749465896 Chr.6: 136241681-136241681 on Build GRCh38
rs779720529 Chr.6: 136241615-136241615 on Build GRCh38
rs147722462 Chr.6: 136241624-136241624 on Build GRCh38
rs1054407867 Chr.6: 136241574-136241574 on Build GRCh38
rs267600827 Chr.6: 136241506-136241506 on Build GRCh38
rs745543926 Chr.6: 136241600-136241600 on Build GRCh38
rs561236270 Chr.6: 136241649-136241649 on Build GRCh38
rs556421355 Chr.6: 136241500-136241500 on Build GRCh38
rs374313243 Chr.6: 136241652-136241652 on Build GRCh38
rs753573464 Chr.6: 136241566-136241566 on Build GRCh38
rs756960194 Chr.6: 136241671-136241671 on Build GRCh38
rs752564130 Chr.6: 136241534-136241534 on Build GRCh38
rs766774654 Chr.6: 136241630-136241630 on Build GRCh38
rs200045608 Chr.6: 136241548-136241548 on Build GRCh38
rs757117317 Chr.6: 136241577-136241577 on Build GRCh38
rs760813346 Chr.6: 136241662-136241662 on Build GRCh38
rs140221512 Chr.6: 136241614-136241614 on Build GRCh38
rs142592349 Chr.6: 136241669-136241669 on Build GRCh38

View Full Product Details