Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52591334-52591844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT72
Entrez Gene ID
140807
Gene Name
keratin 72
Gene Aliases
K6IRS2, K6irs, KRT6, KRT6IRS2
UniGene
Hs.662013
Gene Chromosome Location
Chr.12: 52585589-52602900 on Build GRCh38
Transcripts
NM_001146225.1 NM_001146226.1 NM_080747.2 XM_017018833.1 XM_017018834.1 XM_017018835.1 XM_017018836.1 XM_017018837.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1239295 c.805A>G p.T269A Chr.12: 52591622-52591622 on Build GRCh38
940780 c.892G>A p.E298K Chr.12: 52591535-52591535 on Build GRCh38
548824 c.909C>T p.Y303Y Chr.12: 52591518-52591518 on Build GRCh38
7257409 c.849A>C p.S283S Chr.12: 52591578-52591578 on Build GRCh38
7248951 c.898C>T p.R300C Chr.12: 52591529-52591529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11613547 Chr.12: 52591382-52591382 on Build GRCh38
rs183502770 Chr.12: 52591597-52591597 on Build GRCh38
rs192300174 Chr.12: 52591748-52591748 on Build GRCh38
rs773168116 Chr.12: 52591429-52591429 on Build GRCh38
rs771506380 Chr.12: 52591424-52591424 on Build GRCh38
rs57091900 Chr.12: 52591375-52591375 on Build GRCh38
rs1048248445 Chr.12: 52591815-52591815 on Build GRCh38
rs762654484 Chr.12: 52591603-52591603 on Build GRCh38
rs756736404 Chr.12: 52591565-52591565 on Build GRCh38
rs768497145 Chr.12: 52591647-52591647 on Build GRCh38
rs377015608 Chr.12: 52591669-52591669 on Build GRCh38
rs776549204 Chr.12: 52591513-52591513 on Build GRCh38
rs762014322 Chr.12: 52591468-52591468 on Build GRCh38
rs201887205 Chr.12: 52591593-52591593 on Build GRCh38
rs374391145 Chr.12: 52591441-52591441 on Build GRCh38
rs752607357 Chr.12: 52591535-52591535 on Build GRCh38
rs747007532 Chr.12: 52591508-52591508 on Build GRCh38
rs924513420 Chr.12: 52591373-52591373 on Build GRCh38
rs755309278 Chr.12: 52591490-52591490 on Build GRCh38
rs982305715 Chr.12: 52591400-52591400 on Build GRCh38
rs908326391 Chr.12: 52591391-52591391 on Build GRCh38
rs199685214 Chr.12: 52591524-52591524 on Build GRCh38
rs906905550 Chr.12: 52591641-52591641 on Build GRCh38
rs548680308 Chr.12: 52591598-52591598 on Build GRCh38
rs555458483 Chr.12: 52591785-52591785 on Build GRCh38
rs759762067 Chr.12: 52591523-52591523 on Build GRCh38
rs368168550 Chr.12: 52591432-52591432 on Build GRCh38
rs774646841 Chr.12: 52591463-52591463 on Build GRCh38
rs745766281 Chr.12: 52591573-52591573 on Build GRCh38
rs777794753 Chr.12: 52591564-52591564 on Build GRCh38
rs1022875516 Chr.12: 52591762-52591762 on Build GRCh38
rs752396456 Chr.12: 52591549-52591549 on Build GRCh38
rs758019493 Chr.12: 52591554-52591554 on Build GRCh38
rs989721136 Chr.12: 52591445-52591445 on Build GRCh38
rs149642871 Chr.12: 52591528-52591528 on Build GRCh38
rs776259950 Chr.12: 52591446-52591446 on Build GRCh38
rs965976805 Chr.12: 52591776-52591776 on Build GRCh38
rs751015842 Chr.12: 52591469-52591469 on Build GRCh38
rs533509486 Chr.12: 52591758-52591780 on Build GRCh38
rs12824366 Chr.12: 52591509-52591509 on Build GRCh38
rs71092772 Chr.12: 52591374-52591375 on Build GRCh38
rs142172097 Chr.12: 52591609-52591609 on Build GRCh38
rs138828050 Chr.12: 52591735-52591735 on Build GRCh38
rs367576767 Chr.12: 52591536-52591536 on Build GRCh38
rs35337079 Chr.12: 52591811-52591812 on Build GRCh38
rs370711828 Chr.12: 52591435-52591435 on Build GRCh38
rs552852610 Chr.12: 52591587-52591587 on Build GRCh38
rs779959806 Chr.12: 52591579-52591579 on Build GRCh38
rs756111648 Chr.12: 52591639-52591639 on Build GRCh38
rs145259719 Chr.12: 52591484-52591484 on Build GRCh38
rs1031724684 Chr.12: 52591699-52591699 on Build GRCh38
rs141797377 Chr.12: 52591370-52591370 on Build GRCh38
rs773673423 Chr.12: 52591799-52591799 on Build GRCh38
rs777890193 Chr.12: 52591663-52591663 on Build GRCh38
rs147007594 Chr.12: 52591531-52591531 on Build GRCh38
rs760186282 Chr.12: 52591423-52591423 on Build GRCh38
rs967002772 Chr.12: 52591765-52591765 on Build GRCh38
rs866735122 Chr.12: 52591562-52591562 on Build GRCh38
rs562333020 Chr.12: 52591406-52591406 on Build GRCh38
rs763304018 Chr.12: 52591447-52591447 on Build GRCh38
rs60159645 Chr.12: 52591381-52591381 on Build GRCh38
rs150482370 Chr.12: 52591480-52591480 on Build GRCh38
rs200651000 Chr.12: 52591642-52591642 on Build GRCh38
rs576169915 Chr.12: 52591773-52591773 on Build GRCh38
rs186681670 Chr.12: 52591736-52591736 on Build GRCh38
rs769670135 Chr.12: 52591515-52591515 on Build GRCh38
rs879002578 Chr.12: 52591650-52591650 on Build GRCh38
rs778544219 Chr.12: 52591491-52591491 on Build GRCh38
rs776741905 Chr.12: 52591678-52591678 on Build GRCh38
rs866580582 Chr.12: 52591708-52591708 on Build GRCh38
rs140513368 Chr.12: 52591359-52591360 on Build GRCh38
rs112771714 Chr.12: 52591518-52591518 on Build GRCh38
rs112821912 Chr.12: 52591637-52591637 on Build GRCh38
rs957013217 Chr.12: 52591705-52591705 on Build GRCh38
rs1033861618 Chr.12: 52591539-52591539 on Build GRCh38
rs974711618 Chr.12: 52591798-52591798 on Build GRCh38
rs977402670 Chr.12: 52591373-52591382 on Build GRCh38
rs909248296 Chr.12: 52591784-52591784 on Build GRCh38
rs185861249 Chr.12: 52591774-52591774 on Build GRCh38
rs991064029 Chr.12: 52591806-52591806 on Build GRCh38
rs762095191 Chr.12: 52591713-52591713 on Build GRCh38
rs73320351 Chr.12: 52591529-52591529 on Build GRCh38
rs970457863 Chr.12: 52591392-52591392 on Build GRCh38
rs764399532 Chr.12: 52591462-52591462 on Build GRCh38
rs148734204 Chr.12: 52591505-52591505 on Build GRCh38
rs553710545 Chr.12: 52591724-52591724 on Build GRCh38
rs376773990 Chr.12: 52591517-52591517 on Build GRCh38
rs201080258 Chr.12: 52591594-52591594 on Build GRCh38
rs763669688 Chr.12: 52591604-52591604 on Build GRCh38
rs144214933 Chr.12: 52591636-52591636 on Build GRCh38
rs887993665 Chr.12: 52591437-52591437 on Build GRCh38
rs777247704 Chr.12: 52591507-52591507 on Build GRCh38
rs564060054 Chr.12: 52591816-52591816 on Build GRCh38
rs267603521 Chr.12: 52591556-52591556 on Build GRCh38
rs765538689 Chr.12: 52591543-52591543 on Build GRCh38
rs971451403 Chr.12: 52591367-52591367 on Build GRCh38
rs778450197 Chr.12: 52591417-52591417 on Build GRCh38
rs769757085 Chr.12: 52591578-52591578 on Build GRCh38
rs915237648 Chr.12: 52591448-52591448 on Build GRCh38
rs371811856 Chr.12: 52591586-52591586 on Build GRCh38
rs1011522091 Chr.12: 52591725-52591725 on Build GRCh38
rs753058952 Chr.12: 52591602-52591602 on Build GRCh38
rs769876314 Chr.12: 52591574-52591574 on Build GRCh38
rs142024844 Chr.12: 52591814-52591814 on Build GRCh38
rs766830948 Chr.12: 52591481-52591481 on Build GRCh38
rs774690274 Chr.12: 52591673-52591673 on Build GRCh38
rs561562480 Chr.12: 52591514-52591514 on Build GRCh38
rs765630761 Chr.12: 52591599-52591599 on Build GRCh38
rs201891093 Chr.12: 52591379-52591379 on Build GRCh38
rs571997286 Chr.12: 52591807-52591807 on Build GRCh38
rs147630760 Chr.12: 52591566-52591566 on Build GRCh38

View Full Product Details