Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49817842-49818351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT7
Entrez Gene ID
11143
Gene Name
lysine acetyltransferase 7
Gene Aliases
HBO1, HBOA, MYST2, ZC2HC7
UniGene
Hs.21907
Gene Chromosome Location
Chr.17: 49788619-49829096 on Build GRCh38
Transcripts
NM_001199155.1 NM_001199156.1 NM_001199157.1 NM_001199158.1 NM_007067.4 XM_011524235.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4754537 c.1145G>A p.R382H Chr.17: 49818001-49818001 on Build GRCh38
252636 c.1154T>C p.M385T Chr.17: 49818010-49818010 on Build GRCh38
356049 c.1118A>C p.K373T Chr.17: 49817974-49817974 on Build GRCh38
4788603 c.1146C>A p.R382R Chr.17: 49818002-49818002 on Build GRCh38
6580178 c.1091_1102del12 p.L365_C368delLYMC Chr.17: 49817947-49817958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982561943 Chr.17: 49818189-49818189 on Build GRCh38
rs200465294 Chr.17: 49817886-49817886 on Build GRCh38
rs936605432 Chr.17: 49818201-49818201 on Build GRCh38
rs765713870 Chr.17: 49818034-49818034 on Build GRCh38
rs201234791 Chr.17: 49817992-49817992 on Build GRCh38
rs17853073 Chr.17: 49817909-49817909 on Build GRCh38
rs778845734 Chr.17: 49818063-49818063 on Build GRCh38
rs760255504 Chr.17: 49818110-49818110 on Build GRCh38
rs764339628 Chr.17: 49818009-49818009 on Build GRCh38
rs768145602 Chr.17: 49817951-49817951 on Build GRCh38
rs572148518 Chr.17: 49818319-49818319 on Build GRCh38
rs557188497 Chr.17: 49818203-49818203 on Build GRCh38
rs754644836 Chr.17: 49818262-49818262 on Build GRCh38
rs995779960 Chr.17: 49818164-49818164 on Build GRCh38
rs777635378 Chr.17: 49818052-49818052 on Build GRCh38
rs772074802 Chr.17: 49817987-49817987 on Build GRCh38
rs754472886 Chr.17: 49818047-49818047 on Build GRCh38
rs1037018317 Chr.17: 49818143-49818143 on Build GRCh38
rs920573196 Chr.17: 49817990-49817990 on Build GRCh38
rs990089123 Chr.17: 49818298-49818298 on Build GRCh38
rs908357734 Chr.17: 49818221-49818221 on Build GRCh38
rs760864168 Chr.17: 49817993-49817993 on Build GRCh38
rs957484087 Chr.17: 49818278-49818278 on Build GRCh38
rs75933720 Chr.17: 49818255-49818255 on Build GRCh38
rs924915023 Chr.17: 49818193-49818193 on Build GRCh38
rs759605902 Chr.17: 49817978-49817978 on Build GRCh38
rs544484216 Chr.17: 49817940-49817940 on Build GRCh38
rs762138630 Chr.17: 49818029-49818029 on Build GRCh38
rs935969729 Chr.17: 49817979-49817979 on Build GRCh38
rs926526612 Chr.17: 49818281-49818281 on Build GRCh38
rs1055730965 Chr.17: 49818014-49818015 on Build GRCh38
rs754249267 Chr.17: 49817876-49817876 on Build GRCh38
rs558525453 Chr.17: 49818276-49818276 on Build GRCh38
rs548318161 Chr.17: 49818084-49818084 on Build GRCh38
rs114180924 Chr.17: 49818156-49818156 on Build GRCh38
rs750895829 Chr.17: 49818036-49818036 on Build GRCh38
rs977080686 Chr.17: 49818258-49818258 on Build GRCh38
rs1024045623 Chr.17: 49818225-49818225 on Build GRCh38
rs375776451 Chr.17: 49817887-49817887 on Build GRCh38
rs749082146 Chr.17: 49818054-49818054 on Build GRCh38
rs755801247 Chr.17: 49818051-49818051 on Build GRCh38
rs753365016 Chr.17: 49818173-49818173 on Build GRCh38
rs1012640891 Chr.17: 49818210-49818210 on Build GRCh38
rs770646330 Chr.17: 49818058-49818058 on Build GRCh38
rs200387456 Chr.17: 49818020-49818020 on Build GRCh38
rs191674320 Chr.17: 49818138-49818138 on Build GRCh38
rs769666458 Chr.17: 49817933-49817933 on Build GRCh38
rs777692678 Chr.17: 49817937-49817937 on Build GRCh38
rs754458595 Chr.17: 49817891-49817891 on Build GRCh38
rs553187021 Chr.17: 49818257-49818257 on Build GRCh38
rs144208523 Chr.17: 49818220-49818220 on Build GRCh38
rs765767355 Chr.17: 49818166-49818166 on Build GRCh38
rs528188773 Chr.17: 49818049-49818049 on Build GRCh38
rs941266204 Chr.17: 49817894-49817894 on Build GRCh38
rs1042272417 Chr.17: 49818065-49818065 on Build GRCh38
rs747837705 Chr.17: 49817918-49817918 on Build GRCh38

View Full Product Details