Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130916899-130917406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF4
Entrez Gene ID
50649
Gene Name
Rho guanine nucleotide exchange factor 4
Gene Aliases
ASEF, ASEF1, GEF4, STM6
UniGene
Hs.469935
Gene Chromosome Location
Chr.2: 130836916-131047253 on Build GRCh38
Transcripts
NM_015320.3 NM_032995.2 XM_005263681.1 XM_005263685.3 XM_011511274.2 XM_017004231.1

SNP Detail

SNP ID Chromosome Location
rs542845027 Chr.2: 130917080-130917080 on Build GRCh38
rs994418056 Chr.2: 130916921-130916921 on Build GRCh38
rs529072750 Chr.2: 130917144-130917144 on Build GRCh38
rs542709755 Chr.2: 130917274-130917274 on Build GRCh38
rs914856648 Chr.2: 130917322-130917322 on Build GRCh38
rs773975956 Chr.2: 130917253-130917253 on Build GRCh38
rs914928668 Chr.2: 130916997-130916997 on Build GRCh38
rs572153704 Chr.2: 130917061-130917061 on Build GRCh38
rs904995194 Chr.2: 130917089-130917089 on Build GRCh38
rs749132160 Chr.2: 130917054-130917054 on Build GRCh38
rs80074373 Chr.2: 130916964-130916964 on Build GRCh38
rs747068969 Chr.2: 130917212-130917212 on Build GRCh38
rs766954090 Chr.2: 130916981-130916981 on Build GRCh38
rs1025367046 Chr.2: 130916922-130916922 on Build GRCh38
rs115880351 Chr.2: 130917239-130917239 on Build GRCh38
rs779174176 Chr.2: 130917092-130917092 on Build GRCh38
rs988192420 Chr.2: 130916988-130916988 on Build GRCh38
rs532382748 Chr.2: 130917228-130917228 on Build GRCh38
rs1053699148 Chr.2: 130917052-130917052 on Build GRCh38
rs757101790 Chr.2: 130917337-130917337 on Build GRCh38
rs1025933792 Chr.2: 130917161-130917161 on Build GRCh38
rs763979019 Chr.2: 130917310-130917310 on Build GRCh38
rs1004152751 Chr.2: 130917195-130917195 on Build GRCh38
rs753771925 Chr.2: 130917323-130917323 on Build GRCh38
rs1008007393 Chr.2: 130917203-130917203 on Build GRCh38
rs777171533 Chr.2: 130917225-130917225 on Build GRCh38
rs773657792 Chr.2: 130917271-130917271 on Build GRCh38
rs112404508 Chr.2: 130917099-130917099 on Build GRCh38
rs770275136 Chr.2: 130917241-130917241 on Build GRCh38
rs76778920 Chr.2: 130917297-130917297 on Build GRCh38
rs955150728 Chr.2: 130917165-130917165 on Build GRCh38
rs1045704770 Chr.2: 130917075-130917075 on Build GRCh38
rs762216936 Chr.2: 130916946-130916946 on Build GRCh38
rs868228598 Chr.2: 130917215-130917215 on Build GRCh38
rs963769665 Chr.2: 130916920-130916920 on Build GRCh38
rs926211881 Chr.2: 130917014-130917014 on Build GRCh38
rs775435889 Chr.2: 130917178-130917178 on Build GRCh38
rs980444423 Chr.2: 130917378-130917378 on Build GRCh38
rs373185373 Chr.2: 130916951-130916951 on Build GRCh38
rs1015311202 Chr.2: 130917216-130917216 on Build GRCh38
rs780272086 Chr.2: 130917353-130917353 on Build GRCh38
rs778693388 Chr.2: 130917338-130917338 on Build GRCh38
rs202016094 Chr.2: 130917278-130917278 on Build GRCh38
rs1038220636 Chr.2: 130917120-130917120 on Build GRCh38
rs1001393966 Chr.2: 130917110-130917110 on Build GRCh38
rs564904749 Chr.2: 130917193-130917193 on Build GRCh38
rs750151523 Chr.2: 130917343-130917343 on Build GRCh38
rs868144943 Chr.2: 130917207-130917207 on Build GRCh38
rs1056256185 Chr.2: 130917136-130917136 on Build GRCh38
rs761193886 Chr.2: 130917345-130917345 on Build GRCh38
rs981519648 Chr.2: 130917339-130917339 on Build GRCh38
rs994885462 Chr.2: 130917263-130917263 on Build GRCh38
rs150760589 Chr.2: 130917124-130917124 on Build GRCh38
rs927392751 Chr.2: 130917342-130917342 on Build GRCh38
rs754598308 Chr.2: 130916967-130916967 on Build GRCh38
rs746076875 Chr.2: 130917125-130917125 on Build GRCh38
rs561182052 Chr.2: 130917107-130917107 on Build GRCh38
rs939184697 Chr.2: 130917115-130917115 on Build GRCh38
rs142228684 Chr.2: 130916941-130916941 on Build GRCh38
rs3739125 Chr.2: 130917154-130917154 on Build GRCh38
rs987679606 Chr.2: 130916978-130916978 on Build GRCh38
rs748941176 Chr.2: 130917045-130917045 on Build GRCh38
rs530244935 Chr.2: 130917295-130917295 on Build GRCh38
rs559994783 Chr.2: 130917029-130917029 on Build GRCh38
rs545046582 Chr.2: 130916987-130916987 on Build GRCh38
rs1019132094 Chr.2: 130917289-130917289 on Build GRCh38
rs879583181 Chr.2: 130916963-130916963 on Build GRCh38

View Full Product Details