Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114289557-114290122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1L2
Entrez Gene ID
84632
Gene Name
actin filament associated protein 1 like 2
Gene Aliases
CTB-1144G6.4, KIAA1914, XB130
UniGene
Hs.501106
Gene Chromosome Location
Chr.10: 114280730-114405143 on Build GRCh38
Transcripts
XM_005270233.4 XM_005270237.4 XM_005270241.4 XM_005270242.4 XM_011540286.2 XM_017016792.1 XM_017016793.1 XM_017016794.1 XM_017016795.1 XM_017016796.1 XM_017016797.1 XM_017016798.1 XM_017016804.1 XM_017016814.1 XM_017016815.1 XM_017016818.1 XM_017016829.1 XM_017016832.1 NM_001272046.1 NM_001320804.1 XM_011539754.2 XM_011539757.2 XM_017016177.1 XM_017016178.1 XM_017016179.1 XM_017016180.1

Gene Symbol
VWA2
Entrez Gene ID
340706
Gene Name
von Willebrand factor A domain containing 2
Gene Aliases
AMACO, CCSP-2, NET42
UniGene
Hs.197741
Gene Chromosome Location
Chr.10: 114239254-114294500 on Build GRCh38
Transcripts
XM_005270233.4 XM_005270237.4 XM_005270241.4 XM_005270242.4 XM_011540286.2 XM_017016792.1 XM_017016793.1 XM_017016794.1 XM_017016795.1 XM_017016796.1 XM_017016797.1 XM_017016798.1 XM_017016804.1 XM_017016814.1 XM_017016815.1 XM_017016818.1 XM_017016829.1 XM_017016832.1 NM_001272046.1 NM_001320804.1 XM_011539754.2 XM_011539757.2 XM_017016177.1 XM_017016178.1 XM_017016179.1 XM_017016180.1

SNP Detail

SNP ID Chromosome Location
rs963482559 Chr.10: 114289952-114289952 on Build GRCh38
rs1042720573 Chr.10: 114290077-114290077 on Build GRCh38
rs1016712914 Chr.10: 114289947-114289947 on Build GRCh38
rs891726688 Chr.10: 114289746-114289746 on Build GRCh38
rs566686002 Chr.10: 114289685-114289685 on Build GRCh38
rs983449567 Chr.10: 114289931-114289931 on Build GRCh38
rs994028967 Chr.10: 114289791-114289791 on Build GRCh38
rs770140668 Chr.10: 114289878-114289878 on Build GRCh38
rs923319008 Chr.10: 114289949-114289949 on Build GRCh38
rs780860103 Chr.10: 114289995-114289995 on Build GRCh38
rs776328362 Chr.10: 114289733-114289733 on Build GRCh38
rs35431260 Chr.10: 114289948-114289948 on Build GRCh38
rs1017756222 Chr.10: 114289856-114289856 on Build GRCh38
rs942401713 Chr.10: 114290101-114290101 on Build GRCh38
rs756511415 Chr.10: 114289592-114289592 on Build GRCh38
rs77692317 Chr.10: 114289995-114289995 on Build GRCh38
rs55756191 Chr.10: 114289986-114289986 on Build GRCh38
rs1001274895 Chr.10: 114289833-114289833 on Build GRCh38
rs534774896 Chr.10: 114289688-114289688 on Build GRCh38
rs1038312060 Chr.10: 114289715-114289715 on Build GRCh38
rs942149956 Chr.10: 114289602-114289602 on Build GRCh38
rs568221222 Chr.10: 114289850-114289850 on Build GRCh38
rs942069917 Chr.10: 114290021-114290021 on Build GRCh38
rs756028789 Chr.10: 114289588-114289589 on Build GRCh38
rs979952261 Chr.10: 114289632-114289632 on Build GRCh38
rs990795274 Chr.10: 114289878-114289884 on Build GRCh38
rs750055296 Chr.10: 114289585-114289585 on Build GRCh38
rs760287284 Chr.10: 114289946-114289946 on Build GRCh38
rs909193071 Chr.10: 114289963-114289963 on Build GRCh38
rs536909423 Chr.10: 114290053-114290053 on Build GRCh38
rs1033975623 Chr.10: 114289839-114289839 on Build GRCh38
rs931932052 Chr.10: 114289668-114289668 on Build GRCh38
rs767275086 Chr.10: 114289582-114289582 on Build GRCh38
rs946004020 Chr.10: 114290071-114290071 on Build GRCh38
rs796820393 Chr.10: 114289992-114289993 on Build GRCh38
rs923369878 Chr.10: 114289652-114289652 on Build GRCh38
rs114026411 Chr.10: 114289917-114289917 on Build GRCh38
rs559399071 Chr.10: 114289827-114289827 on Build GRCh38
rs887810621 Chr.10: 114289705-114289705 on Build GRCh38
rs904268291 Chr.10: 114289804-114289804 on Build GRCh38
rs925637303 Chr.10: 114290092-114290092 on Build GRCh38
rs1005876790 Chr.10: 114289709-114289710 on Build GRCh38
rs913109434 Chr.10: 114290051-114290051 on Build GRCh38
rs373662032 Chr.10: 114289675-114289675 on Build GRCh38
rs1030555491 Chr.10: 114289916-114289916 on Build GRCh38
rs997698854 Chr.10: 114289877-114289877 on Build GRCh38
rs575822389 Chr.10: 114290030-114290030 on Build GRCh38
rs980617142 Chr.10: 114289977-114289977 on Build GRCh38
rs945680589 Chr.10: 114289760-114289760 on Build GRCh38
rs77701720 Chr.10: 114289992-114289993 on Build GRCh38
rs112402661 Chr.10: 114289818-114289818 on Build GRCh38
rs752391769 Chr.10: 114289729-114289729 on Build GRCh38
rs138390990 Chr.10: 114289715-114289715 on Build GRCh38
rs909261272 Chr.10: 114289600-114289600 on Build GRCh38
rs35165936 Chr.10: 114290010-114290010 on Build GRCh38
rs897088444 Chr.10: 114290031-114290031 on Build GRCh38
rs900647314 Chr.10: 114289861-114289861 on Build GRCh38
rs927149292 Chr.10: 114289714-114289714 on Build GRCh38
rs768428060 Chr.10: 114289994-114289995 on Build GRCh38
rs1043116231 Chr.10: 114289803-114289803 on Build GRCh38
rs986252755 Chr.10: 114289953-114289953 on Build GRCh38
rs546506120 Chr.10: 114290063-114290063 on Build GRCh38
rs938488757 Chr.10: 114290023-114290023 on Build GRCh38
rs749491455 Chr.10: 114289993-114289995 on Build GRCh38
rs927418130 Chr.10: 114290006-114290006 on Build GRCh38
rs748551969 Chr.10: 114289806-114289807 on Build GRCh38

View Full Product Details