Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16439398-16439896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC75A-AS1
Entrez Gene ID
125144
Gene Name
LRRC75A antisense RNA 1
Gene Aliases
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
UniGene
Hs.368934
Gene Chromosome Location
Chr.17: 16438987-16470648 on Build GRCh38
Transcripts
NR_027158.1 NR_027159.1 NR_027160.1 NR_027161.1 NR_027162.1 NR_027163.1 NR_027164.1 NR_027165.1 NR_027166.1 NR_027167.1 NR_027168.2 NR_027169.1 NR_027170.1 NR_027171.1 NR_027172.2 NR_027173.1 NR_027174.1 NR_027175.1 NR_027176.1 NR_027177.1 NR_027178.1 NR_027179.1 NR_027667.1 NR_045021.1 NR_045022.1 NR_045023.1 NR_045024.1 NR_045025.1 NR_045026.1 NR_045027.1 NR_045028.1 NR_045029.1 NR_003043.1

Gene Symbol
SNORD49B
Entrez Gene ID
692087
Gene Name
small nucleolar RNA, C/D box 49B
Gene Aliases
U49B
UniGene
Gene Chromosome Location
Chr.17: 16439509-16439556 on Build GRCh38
Transcripts
NR_027158.1 NR_027159.1 NR_027160.1 NR_027161.1 NR_027162.1 NR_027163.1 NR_027164.1 NR_027165.1 NR_027166.1 NR_027167.1 NR_027168.2 NR_027169.1 NR_027170.1 NR_027171.1 NR_027172.2 NR_027173.1 NR_027174.1 NR_027175.1 NR_027176.1 NR_027177.1 NR_027178.1 NR_027179.1 NR_027667.1 NR_045021.1 NR_045022.1 NR_045023.1 NR_045024.1 NR_045025.1 NR_045026.1 NR_045027.1 NR_045028.1 NR_045029.1 NR_003043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3680088 c.273G>C p.W91C Chr.17: 16439414-16439414 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763171951 Chr.17: 16439574-16439574 on Build GRCh38
rs1018605909 Chr.17: 16439566-16439566 on Build GRCh38
rs774406234 Chr.17: 16439652-16439652 on Build GRCh38
rs748935210 Chr.17: 16439445-16439445 on Build GRCh38
rs374801799 Chr.17: 16439576-16439576 on Build GRCh38
rs371967345 Chr.17: 16439737-16439737 on Build GRCh38
rs765737681 Chr.17: 16439433-16439433 on Build GRCh38
rs771624816 Chr.17: 16439546-16439546 on Build GRCh38
rs750625708 Chr.17: 16439471-16439471 on Build GRCh38
rs368455014 Chr.17: 16439824-16439824 on Build GRCh38
rs191252990 Chr.17: 16439608-16439608 on Build GRCh38
rs746032277 Chr.17: 16439735-16439735 on Build GRCh38
rs376962584 Chr.17: 16439514-16439516 on Build GRCh38
rs759666912 Chr.17: 16439588-16439588 on Build GRCh38
rs755132827 Chr.17: 16439807-16439807 on Build GRCh38
rs182721071 Chr.17: 16439711-16439711 on Build GRCh38
rs868370923 Chr.17: 16439690-16439691 on Build GRCh38
rs887489363 Chr.17: 16439747-16439747 on Build GRCh38
rs3209302 Chr.17: 16439495-16439495 on Build GRCh38
rs773690186 Chr.17: 16439434-16439434 on Build GRCh38
rs777547143 Chr.17: 16439442-16439442 on Build GRCh38
rs751599652 Chr.17: 16439484-16439484 on Build GRCh38
rs771602875 Chr.17: 16439672-16439672 on Build GRCh38
rs766426684 Chr.17: 16439482-16439482 on Build GRCh38
rs201237828 Chr.17: 16439521-16439521 on Build GRCh38
rs763889880 Chr.17: 16439609-16439609 on Build GRCh38
rs753930799 Chr.17: 16439789-16439789 on Build GRCh38
rs767658365 Chr.17: 16439663-16439663 on Build GRCh38
rs112087561 Chr.17: 16439526-16439526 on Build GRCh38
rs374042866 Chr.17: 16439874-16439874 on Build GRCh38
rs371434312 Chr.17: 16439749-16439749 on Build GRCh38
rs778287123 Chr.17: 16439568-16439569 on Build GRCh38
rs765116429 Chr.17: 16439693-16439693 on Build GRCh38
rs748599552 Chr.17: 16439642-16439642 on Build GRCh38
rs552147653 Chr.17: 16439650-16439650 on Build GRCh38
rs1050312095 Chr.17: 16439525-16439525 on Build GRCh38
rs1009019875 Chr.17: 16439538-16439538 on Build GRCh38
rs777188637 Chr.17: 16439462-16439462 on Build GRCh38
rs777743762 Chr.17: 16439517-16439517 on Build GRCh38
rs117783090 Chr.17: 16439743-16439743 on Build GRCh38
rs773840309 Chr.17: 16439647-16439647 on Build GRCh38
rs11871958 Chr.17: 16439519-16439519 on Build GRCh38
rs547033065 Chr.17: 16439691-16439692 on Build GRCh38
rs756150004 Chr.17: 16439498-16439499 on Build GRCh38
rs375913111 Chr.17: 16439719-16439719 on Build GRCh38
rs753416154 Chr.17: 16439558-16439564 on Build GRCh38
rs757141743 Chr.17: 16439618-16439618 on Build GRCh38
rs572295764 Chr.17: 16439467-16439467 on Build GRCh38
rs1045007777 Chr.17: 16439580-16439580 on Build GRCh38
rs954668950 Chr.17: 16439509-16439509 on Build GRCh38
rs147936849 Chr.17: 16439653-16439653 on Build GRCh38
rs917358962 Chr.17: 16439485-16439485 on Build GRCh38
rs373556872 Chr.17: 16439595-16439595 on Build GRCh38
rs72833950 Chr.17: 16439712-16439712 on Build GRCh38
rs927518746 Chr.17: 16439864-16439864 on Build GRCh38
rs748474866 Chr.17: 16439709-16439710 on Build GRCh38
rs371862656 Chr.17: 16439460-16439460 on Build GRCh38
rs959243685 Chr.17: 16439814-16439814 on Build GRCh38
rs758066514 Chr.17: 16439700-16439700 on Build GRCh38
rs762672444 Chr.17: 16439582-16439582 on Build GRCh38
rs947703346 Chr.17: 16439792-16439792 on Build GRCh38
rs767746428 Chr.17: 16439591-16439591 on Build GRCh38
rs775920699 Chr.17: 16439740-16439740 on Build GRCh38
rs777800876 Chr.17: 16439510-16439512 on Build GRCh38
rs946157879 Chr.17: 16439656-16439656 on Build GRCh38
rs761185506 Chr.17: 16439456-16439456 on Build GRCh38
rs1010297191 Chr.17: 16439628-16439628 on Build GRCh38
rs371585303 Chr.17: 16439565-16439565 on Build GRCh38
rs761837827 Chr.17: 16439755-16439755 on Build GRCh38
rs947747272 Chr.17: 16439579-16439579 on Build GRCh38
rs921877270 Chr.17: 16439504-16439504 on Build GRCh38
rs753870813 Chr.17: 16439684-16439684 on Build GRCh38
rs763109828 Chr.17: 16439480-16439480 on Build GRCh38
rs745596414 Chr.17: 16439652-16439655 on Build GRCh38
rs779359661 Chr.17: 16439734-16439734 on Build GRCh38
rs756763398 Chr.17: 16439566-16439569 on Build GRCh38
rs980502215 Chr.17: 16439802-16439802 on Build GRCh38
rs368508925 Chr.17: 16439454-16439454 on Build GRCh38
rs778418841 Chr.17: 16439531-16439531 on Build GRCh38
rs761486650 Chr.17: 16439690-16439690 on Build GRCh38
rs984252119 Chr.17: 16439605-16439605 on Build GRCh38
rs570423795 Chr.17: 16439701-16439701 on Build GRCh38
rs375961885 Chr.17: 16439758-16439758 on Build GRCh38
rs4072454 Chr.17: 16439499-16439499 on Build GRCh38
rs563004931 Chr.17: 16439625-16439625 on Build GRCh38
rs368621303 Chr.17: 16439640-16439640 on Build GRCh38
rs200872411 Chr.17: 16439824-16439826 on Build GRCh38
rs763933638 Chr.17: 16439673-16439673 on Build GRCh38
rs368508784 Chr.17: 16439725-16439725 on Build GRCh38
rs775416626 Chr.17: 16439569-16439569 on Build GRCh38
rs755942978 Chr.17: 16439506-16439506 on Build GRCh38
rs755179854 Chr.17: 16439488-16439488 on Build GRCh38
rs765635055 Chr.17: 16439470-16439470 on Build GRCh38
rs770436459 Chr.17: 16439447-16439447 on Build GRCh38
rs112304410 Chr.17: 16439524-16439524 on Build GRCh38
rs745365628 Chr.17: 16439535-16439535 on Build GRCh38
rs1034517962 Chr.17: 16439812-16439812 on Build GRCh38
rs770308766 Chr.17: 16439749-16439749 on Build GRCh38
rs1057091480 Chr.17: 16439483-16439483 on Build GRCh38
rs370654705 Chr.17: 16439617-16439617 on Build GRCh38
rs761080899 Chr.17: 16439829-16439829 on Build GRCh38
rs756872330 Chr.17: 16439522-16439522 on Build GRCh38
rs369841329 Chr.17: 16439649-16439649 on Build GRCh38
rs775722791 Chr.17: 16439667-16439667 on Build GRCh38
rs773464187 Chr.17: 16439571-16439571 on Build GRCh38
rs569543014 Chr.17: 16439787-16439787 on Build GRCh38
rs977493437 Chr.17: 16439665-16439665 on Build GRCh38
rs537599280 Chr.17: 16439873-16439873 on Build GRCh38
rs748691628 Chr.17: 16439568-16439568 on Build GRCh38
rs759869965 Chr.17: 16439659-16439659 on Build GRCh38
rs374783041 Chr.17: 16439635-16439635 on Build GRCh38
rs558623517 Chr.17: 16439435-16439435 on Build GRCh38
rs535276258 Chr.17: 16439804-16439804 on Build GRCh38
rs527303136 Chr.17: 16439621-16439621 on Build GRCh38

View Full Product Details