Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 43376097-43376366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS20
Entrez Gene ID
80070
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 20
Gene Aliases
ADAM-TS20, ADAMTS-20, GON-1
UniGene
Hs.287554
Gene Chromosome Location
Chr.12: 43352843-43551971 on Build GRCh38
Transcripts
NM_025003.3 XM_011538754.2 XM_017019979.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589547 c.5179G>A p.D1727N Chr.12: 43376277-43376277 on Build GRCh38
3812004 c.5239T>C p.Y1747H Chr.12: 43376130-43376130 on Build GRCh38
5922940 c.5207G>A p.G1736E Chr.12: 43376249-43376249 on Build GRCh38
6136629 c.5252C>T p.P1751L Chr.12: 43376117-43376117 on Build GRCh38
7196312 c.5143T>C p.C1715R Chr.12: 43376313-43376313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781572950 Chr.12: 43376172-43376172 on Build GRCh38
rs150420241 Chr.12: 43376243-43376243 on Build GRCh38
rs778712329 Chr.12: 43376217-43376217 on Build GRCh38
rs149125994 Chr.12: 43376330-43376330 on Build GRCh38
rs889012400 Chr.12: 43376272-43376272 on Build GRCh38
rs916890159 Chr.12: 43376169-43376169 on Build GRCh38
rs375175126 Chr.12: 43376223-43376223 on Build GRCh38
rs370796585 Chr.12: 43376158-43376158 on Build GRCh38
rs940405391 Chr.12: 43376202-43376202 on Build GRCh38
rs769382541 Chr.12: 43376276-43376276 on Build GRCh38
rs751908562 Chr.12: 43376321-43376321 on Build GRCh38
rs776320896 Chr.12: 43376247-43376247 on Build GRCh38
rs145199935 Chr.12: 43376288-43376288 on Build GRCh38
rs770689269 Chr.12: 43376131-43376138 on Build GRCh38
rs771925620 Chr.12: 43376225-43376225 on Build GRCh38
rs767460567 Chr.12: 43376284-43376284 on Build GRCh38
rs1050232978 Chr.12: 43376269-43376269 on Build GRCh38
rs777583773 Chr.12: 43376229-43376229 on Build GRCh38
rs965751536 Chr.12: 43376291-43376291 on Build GRCh38
rs764545255 Chr.12: 43376320-43376320 on Build GRCh38
rs753309626 Chr.12: 43376154-43376154 on Build GRCh38
rs577453528 Chr.12: 43376233-43376233 on Build GRCh38
rs757516762 Chr.12: 43376168-43376168 on Build GRCh38
rs759789303 Chr.12: 43376203-43376204 on Build GRCh38
rs762284047 Chr.12: 43376329-43376329 on Build GRCh38
rs774551360 Chr.12: 43376151-43376151 on Build GRCh38
rs775834615 Chr.12: 43376286-43376286 on Build GRCh38
rs764571685 Chr.12: 43376162-43376162 on Build GRCh38
rs937608166 Chr.12: 43376143-43376143 on Build GRCh38
rs368350905 Chr.12: 43376306-43376306 on Build GRCh38
rs759040839 Chr.12: 43376257-43376257 on Build GRCh38
rs750474841 Chr.12: 43376178-43376178 on Build GRCh38
rs1009209461 Chr.12: 43376280-43376280 on Build GRCh38

View Full Product Details