Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190366630-190366863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP1
Entrez Gene ID
3628
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
-
UniGene
Hs.32309
Gene Chromosome Location
Chr.2: 190343470-190371665 on Build GRCh38
Transcripts
NM_001128928.1 NM_002194.3 XM_005246532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4902475 c.413C>T p.S138F Chr.2: 190366842-190366842 on Build GRCh38
5956129 c.418G>A p.E140K Chr.2: 190366847-190366847 on Build GRCh38
5679624 c.309A>G p.T103T Chr.2: 190366738-190366738 on Build GRCh38
7103130 c.298G>C p.E100Q Chr.2: 190366727-190366727 on Build GRCh38
4961766 c.353C>A p.S118Y Chr.2: 190366782-190366782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750376955 Chr.2: 190366772-190366772 on Build GRCh38
rs773675666 Chr.2: 190366678-190366678 on Build GRCh38
rs778978058 Chr.2: 190366807-190366807 on Build GRCh38
rs4940 Chr.2: 190366777-190366777 on Build GRCh38
rs201964239 Chr.2: 190366764-190366764 on Build GRCh38
rs780112337 Chr.2: 190366793-190366793 on Build GRCh38
rs746962744 Chr.2: 190366693-190366693 on Build GRCh38
rs758850283 Chr.2: 190366726-190366727 on Build GRCh38
rs747456953 Chr.2: 190366834-190366834 on Build GRCh38
rs748407786 Chr.2: 190366810-190366810 on Build GRCh38
rs774867416 Chr.2: 190366721-190366721 on Build GRCh38
rs769279803 Chr.2: 190366696-190366696 on Build GRCh38
rs762626247 Chr.2: 190366722-190366722 on Build GRCh38
rs11544940 Chr.2: 190366732-190366732 on Build GRCh38
rs761672960 Chr.2: 190366741-190366741 on Build GRCh38
rs142183759 Chr.2: 190366658-190366658 on Build GRCh38
rs531380132 Chr.2: 190366797-190366797 on Build GRCh38
rs778168268 Chr.2: 190366832-190366832 on Build GRCh38
rs772472236 Chr.2: 190366677-190366677 on Build GRCh38
rs758755066 Chr.2: 190366821-190366821 on Build GRCh38
rs768673209 Chr.2: 190366729-190366729 on Build GRCh38
rs755176350 Chr.2: 190366798-190366798 on Build GRCh38

View Full Product Details