Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6800124-6800630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD4
Entrez Gene ID
920
Gene Name
CD4 molecule
Gene Aliases
CD4mut
UniGene
Hs.631659
Gene Chromosome Location
Chr.12: 6789472-6820810 on Build GRCh38
Transcripts
NM_000616.4 NM_001195014.2 NM_001195015.2 NM_001195016.2 NM_001195017.2 XM_017020228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5524724 c.158G>A p.W53* Chr.12: 6800415-6800415 on Build GRCh38
3359905 c.58C>T p.P20S Chr.12: 6800315-6800315 on Build GRCh38
4436748 c.72G>T p.Q24H Chr.12: 6800329-6800329 on Build GRCh38
7224310 c.142A>G p.S48G Chr.12: 6800399-6800399 on Build GRCh38
3464493 c.167C>T p.S56F Chr.12: 6800424-6800424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113009163 Chr.12: 6800566-6800566 on Build GRCh38
rs781935284 Chr.12: 6800156-6800156 on Build GRCh38
rs932038127 Chr.12: 6800278-6800278 on Build GRCh38
rs951831973 Chr.12: 6800493-6800493 on Build GRCh38
rs199744985 Chr.12: 6800307-6800307 on Build GRCh38
rs782791509 Chr.12: 6800423-6800423 on Build GRCh38
rs200118536 Chr.12: 6800476-6800476 on Build GRCh38
rs372831240 Chr.12: 6800405-6800405 on Build GRCh38
rs782647911 Chr.12: 6800373-6800373 on Build GRCh38
rs916351570 Chr.12: 6800247-6800247 on Build GRCh38
rs201791637 Chr.12: 6800499-6800499 on Build GRCh38
rs781875176 Chr.12: 6800485-6800485 on Build GRCh38
rs201647213 Chr.12: 6800398-6800398 on Build GRCh38
rs782429530 Chr.12: 6800210-6800210 on Build GRCh38
rs200808735 Chr.12: 6800496-6800496 on Build GRCh38
rs782337435 Chr.12: 6800407-6800407 on Build GRCh38
rs201991979 Chr.12: 6800451-6800451 on Build GRCh38
rs782148933 Chr.12: 6800148-6800148 on Build GRCh38
rs35259686 Chr.12: 6800441-6800441 on Build GRCh38
rs782714692 Chr.12: 6800449-6800449 on Build GRCh38
rs782014370 Chr.12: 6800428-6800428 on Build GRCh38
rs3216776 Chr.12: 6800215-6800215 on Build GRCh38
rs782589833 Chr.12: 6800214-6800214 on Build GRCh38
rs782614851 Chr.12: 6800269-6800269 on Build GRCh38
rs782107070 Chr.12: 6800418-6800418 on Build GRCh38
rs782517082 Chr.12: 6800365-6800365 on Build GRCh38
rs2365568 Chr.12: 6800222-6800222 on Build GRCh38
rs782733413 Chr.12: 6800304-6800304 on Build GRCh38
rs144413418 Chr.12: 6800176-6800176 on Build GRCh38
rs201059681 Chr.12: 6800207-6800207 on Build GRCh38
rs940577841 Chr.12: 6800524-6800524 on Build GRCh38
rs1023772140 Chr.12: 6800492-6800492 on Build GRCh38
rs782537220 Chr.12: 6800225-6800225 on Build GRCh38
rs545563466 Chr.12: 6800504-6800504 on Build GRCh38
rs782804022 Chr.12: 6800201-6800201 on Build GRCh38
rs782263655 Chr.12: 6800224-6800224 on Build GRCh38
rs782252955 Chr.12: 6800146-6800146 on Build GRCh38
rs200829690 Chr.12: 6800215-6800215 on Build GRCh38
rs11064396 Chr.12: 6800269-6800269 on Build GRCh38
rs782201667 Chr.12: 6800233-6800233 on Build GRCh38
rs374341894 Chr.12: 6800391-6800391 on Build GRCh38
rs151325091 Chr.12: 6800168-6800168 on Build GRCh38
rs781974469 Chr.12: 6800264-6800264 on Build GRCh38
rs143423733 Chr.12: 6800605-6800605 on Build GRCh38
rs782118027 Chr.12: 6800214-6800215 on Build GRCh38
rs893226381 Chr.12: 6800319-6800319 on Build GRCh38
rs782569674 Chr.12: 6800228-6800228 on Build GRCh38
rs782280366 Chr.12: 6800389-6800389 on Build GRCh38
rs201308520 Chr.12: 6800297-6800297 on Build GRCh38
rs201624951 Chr.12: 6800268-6800268 on Build GRCh38
rs976021801 Chr.12: 6800526-6800526 on Build GRCh38
rs782696878 Chr.12: 6800491-6800491 on Build GRCh38
rs781864508 Chr.12: 6800181-6800181 on Build GRCh38
rs782224163 Chr.12: 6800510-6800510 on Build GRCh38
rs552669070 Chr.12: 6800183-6800183 on Build GRCh38
rs200325999 Chr.12: 6800235-6800235 on Build GRCh38
rs202129605 Chr.12: 6800331-6800331 on Build GRCh38
rs201040272 Chr.12: 6800145-6800145 on Build GRCh38
rs142478992 Chr.12: 6800149-6800149 on Build GRCh38
rs781961383 Chr.12: 6800415-6800415 on Build GRCh38
rs782602611 Chr.12: 6800350-6800350 on Build GRCh38
rs781852305 Chr.12: 6800306-6800306 on Build GRCh38
rs782173942 Chr.12: 6800270-6800270 on Build GRCh38
rs903424784 Chr.12: 6800404-6800404 on Build GRCh38
rs782693436 Chr.12: 6800271-6800271 on Build GRCh38
rs984481164 Chr.12: 6800251-6800251 on Build GRCh38
rs782457345 Chr.12: 6800348-6800348 on Build GRCh38
rs782427182 Chr.12: 6800250-6800250 on Build GRCh38
rs199890832 Chr.12: 6800433-6800433 on Build GRCh38
rs2255301 Chr.12: 6800276-6800276 on Build GRCh38
rs960429527 Chr.12: 6800180-6800180 on Build GRCh38
rs907570206 Chr.12: 6800512-6800512 on Build GRCh38
rs781900879 Chr.12: 6800204-6800204 on Build GRCh38
rs146686851 Chr.12: 6800308-6800308 on Build GRCh38
rs782075037 Chr.12: 6800294-6800294 on Build GRCh38
rs782371453 Chr.12: 6800511-6800511 on Build GRCh38
rs782459887 Chr.12: 6800502-6800502 on Build GRCh38
rs200297122 Chr.12: 6800217-6800217 on Build GRCh38
rs782648680 Chr.12: 6800514-6800514 on Build GRCh38
rs782623892 Chr.12: 6800223-6800223 on Build GRCh38
rs782244672 Chr.12: 6800265-6800265 on Build GRCh38

View Full Product Details