Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 66315024-66315540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELB
Entrez Gene ID
92797
Gene Name
DNA helicase B
Gene Aliases
DHB, hDHB
UniGene
Hs.505941
Gene Chromosome Location
Chr.12: 66302545-66343643 on Build GRCh38
Transcripts
NM_033647.4 NR_135080.1 NR_135081.1 XM_005269234.2 XM_017020230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5443635 c.1948C>G p.L650V Chr.12: 66315331-66315331 on Build GRCh38
6838348 c.1997C>T p.T666I Chr.12: 66315380-66315380 on Build GRCh38
5460974 c.1879A>G p.S627G Chr.12: 66315262-66315262 on Build GRCh38
3464315 c.1909C>T p.L637F Chr.12: 66315292-66315292 on Build GRCh38
4044239 c.1859G>A p.G620D Chr.12: 66315242-66315242 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368884118 Chr.12: 66315218-66315218 on Build GRCh38
rs928087356 Chr.12: 66315220-66315220 on Build GRCh38
rs755953195 Chr.12: 66315407-66315408 on Build GRCh38
rs771930143 Chr.12: 66315326-66315326 on Build GRCh38
rs550639487 Chr.12: 66315134-66315134 on Build GRCh38
rs769466347 Chr.12: 66315289-66315289 on Build GRCh38
rs777162356 Chr.12: 66315252-66315252 on Build GRCh38
rs530821733 Chr.12: 66315067-66315067 on Build GRCh38
rs370786848 Chr.12: 66315416-66315416 on Build GRCh38
rs142976654 Chr.12: 66315309-66315309 on Build GRCh38
rs1004772765 Chr.12: 66315347-66315347 on Build GRCh38
rs867452156 Chr.12: 66315333-66315333 on Build GRCh38
rs7137719 Chr.12: 66315368-66315368 on Build GRCh38
rs748795758 Chr.12: 66315291-66315291 on Build GRCh38
rs76473015 Chr.12: 66315244-66315244 on Build GRCh38
rs767560163 Chr.12: 66315288-66315289 on Build GRCh38
rs780475854 Chr.12: 66315374-66315374 on Build GRCh38
rs762129088 Chr.12: 66315361-66315361 on Build GRCh38
rs765360177 Chr.12: 66315364-66315364 on Build GRCh38
rs752454626 Chr.12: 66315223-66315223 on Build GRCh38
rs752601174 Chr.12: 66315293-66315293 on Build GRCh38
rs745406341 Chr.12: 66315411-66315411 on Build GRCh38
rs760436338 Chr.12: 66315344-66315344 on Build GRCh38
rs974486966 Chr.12: 66315068-66315068 on Build GRCh38
rs752654602 Chr.12: 66315116-66315116 on Build GRCh38
rs781543570 Chr.12: 66315386-66315386 on Build GRCh38
rs377623347 Chr.12: 66315387-66315387 on Build GRCh38
rs778184316 Chr.12: 66315417-66315418 on Build GRCh38
rs989926960 Chr.12: 66315133-66315135 on Build GRCh38
rs935709978 Chr.12: 66315143-66315143 on Build GRCh38
rs777463984 Chr.12: 66315290-66315290 on Build GRCh38
rs989767183 Chr.12: 66315148-66315148 on Build GRCh38
rs117680200 Chr.12: 66315342-66315342 on Build GRCh38
rs750874492 Chr.12: 66315271-66315271 on Build GRCh38
rs778443352 Chr.12: 66315394-66315394 on Build GRCh38
rs989484290 Chr.12: 66315455-66315455 on Build GRCh38
rs759398411 Chr.12: 66315209-66315209 on Build GRCh38
rs148447233 Chr.12: 66315302-66315302 on Build GRCh38
rs144507658 Chr.12: 66315275-66315275 on Build GRCh38
rs1186028 Chr.12: 66315098-66315098 on Build GRCh38
rs745495409 Chr.12: 66315453-66315453 on Build GRCh38
rs1038470391 Chr.12: 66315336-66315336 on Build GRCh38
rs764397288 Chr.12: 66315345-66315345 on Build GRCh38
rs775346203 Chr.12: 66315418-66315418 on Build GRCh38
rs995534218 Chr.12: 66315314-66315314 on Build GRCh38
rs754177530 Chr.12: 66315355-66315355 on Build GRCh38
rs373487548 Chr.12: 66315239-66315239 on Build GRCh38
rs780427434 Chr.12: 66315279-66315279 on Build GRCh38
rs772020383 Chr.12: 66315412-66315412 on Build GRCh38
rs760513321 Chr.12: 66315422-66315422 on Build GRCh38
rs61753953 Chr.12: 66315268-66315268 on Build GRCh38
rs370596928 Chr.12: 66315284-66315284 on Build GRCh38
rs915322300 Chr.12: 66315182-66315182 on Build GRCh38
rs913515503 Chr.12: 66315167-66315167 on Build GRCh38
rs760908487 Chr.12: 66315235-66315235 on Build GRCh38
rs766294170 Chr.12: 66315208-66315208 on Build GRCh38
rs750502116 Chr.12: 66315366-66315366 on Build GRCh38
rs773861620 Chr.12: 66315319-66315319 on Build GRCh38
rs921599980 Chr.12: 66315075-66315075 on Build GRCh38
rs753324397 Chr.12: 66315097-66315097 on Build GRCh38
rs141108737 Chr.12: 66315379-66315379 on Build GRCh38
rs762503840 Chr.12: 66315207-66315207 on Build GRCh38
rs956694160 Chr.12: 66315132-66315132 on Build GRCh38
rs751942873 Chr.12: 66315377-66315377 on Build GRCh38
rs779556828 Chr.12: 66315255-66315255 on Build GRCh38
rs201815365 Chr.12: 66315370-66315370 on Build GRCh38
rs948159638 Chr.12: 66315189-66315189 on Build GRCh38
rs770590380 Chr.12: 66315391-66315391 on Build GRCh38
rs772678493 Chr.12: 66315191-66315191 on Build GRCh38
rs75324469 Chr.12: 66315325-66315325 on Build GRCh38

View Full Product Details