Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54522368-54522916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGRRF1
Entrez Gene ID
10668
Gene Name
cell growth regulator with ring finger domain 1
Gene Aliases
CGR19, RNF197
UniGene
Hs.59106
Gene Chromosome Location
Chr.14: 54509869-54539310 on Build GRCh38
Transcripts
NM_006568.2 XM_011536362.2 XM_017020931.1 XM_017020932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
698353 c.230C>G p.S77* Chr.14: 54522579-54522579 on Build GRCh38
5654928 c.106T>G p.F36V Chr.14: 54522455-54522455 on Build GRCh38
2245113 c.223C>T p.P75S Chr.14: 54522572-54522572 on Build GRCh38
956301 c.164G>T p.R55I Chr.14: 54522513-54522513 on Build GRCh38
70164 c.206G>C p.G69A Chr.14: 54522555-54522555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014905120 Chr.14: 54522711-54522711 on Build GRCh38
rs778885140 Chr.14: 54522637-54522638 on Build GRCh38
rs753729580 Chr.14: 54522536-54522536 on Build GRCh38
rs917796583 Chr.14: 54522821-54522821 on Build GRCh38
rs776333833 Chr.14: 54522448-54522448 on Build GRCh38
rs758991836 Chr.14: 54522781-54522781 on Build GRCh38
rs769742557 Chr.14: 54522454-54522454 on Build GRCh38
rs201744333 Chr.14: 54522490-54522490 on Build GRCh38
rs903565897 Chr.14: 54522647-54522647 on Build GRCh38
rs570432822 Chr.14: 54522736-54522736 on Build GRCh38
rs773381128 Chr.14: 54522425-54522451 on Build GRCh38
rs372053282 Chr.14: 54522633-54522633 on Build GRCh38
rs765945117 Chr.14: 54522714-54522714 on Build GRCh38
rs910522980 Chr.14: 54522812-54522812 on Build GRCh38
rs990759826 Chr.14: 54522809-54522809 on Build GRCh38
rs558312415 Chr.14: 54522395-54522399 on Build GRCh38
rs534447279 Chr.14: 54522766-54522766 on Build GRCh38
rs770423114 Chr.14: 54522613-54522613 on Build GRCh38
rs774039810 Chr.14: 54522499-54522499 on Build GRCh38
rs764485821 Chr.14: 54522509-54522509 on Build GRCh38
rs546310518 Chr.14: 54522828-54522828 on Build GRCh38
rs879841880 Chr.14: 54522749-54522749 on Build GRCh38
rs759596574 Chr.14: 54522471-54522473 on Build GRCh38
rs758543565 Chr.14: 54522541-54522541 on Build GRCh38
rs769648791 Chr.14: 54522576-54522576 on Build GRCh38
rs759177058 Chr.14: 54522502-54522502 on Build GRCh38
rs751607005 Chr.14: 54522466-54522471 on Build GRCh38
rs1042015868 Chr.14: 54522845-54522845 on Build GRCh38
rs755207853 Chr.14: 54522559-54522559 on Build GRCh38
rs993594298 Chr.14: 54522739-54522739 on Build GRCh38
rs146786181 Chr.14: 54522567-54522567 on Build GRCh38
rs772666262 Chr.14: 54522473-54522473 on Build GRCh38
rs993046351 Chr.14: 54522408-54522408 on Build GRCh38
rs1015705853 Chr.14: 54522710-54522710 on Build GRCh38
rs764075753 Chr.14: 54522473-54522481 on Build GRCh38
rs576783144 Chr.14: 54522636-54522636 on Build GRCh38
rs780989272 Chr.14: 54522573-54522573 on Build GRCh38
rs78593805 Chr.14: 54522865-54522865 on Build GRCh38
rs551998169 Chr.14: 54522701-54522701 on Build GRCh38
rs986772131 Chr.14: 54522706-54522706 on Build GRCh38
rs763182778 Chr.14: 54522444-54522445 on Build GRCh38
rs748097604 Chr.14: 54522451-54522451 on Build GRCh38
rs962177894 Chr.14: 54522430-54522430 on Build GRCh38
rs780069132 Chr.14: 54522548-54522548 on Build GRCh38
rs902298575 Chr.14: 54522856-54522856 on Build GRCh38
rs937929844 Chr.14: 54522397-54522397 on Build GRCh38
rs562029606 Chr.14: 54522442-54522442 on Build GRCh38
rs773772495 Chr.14: 54522617-54522617 on Build GRCh38
rs1024538542 Chr.14: 54522731-54522731 on Build GRCh38
rs533354499 Chr.14: 54522659-54522659 on Build GRCh38
rs771655842 Chr.14: 54522625-54522625 on Build GRCh38
rs373707670 Chr.14: 54522420-54522420 on Build GRCh38
rs902246338 Chr.14: 54522590-54522590 on Build GRCh38
rs747966078 Chr.14: 54522575-54522575 on Build GRCh38
rs1001910032 Chr.14: 54522655-54522655 on Build GRCh38
rs973931317 Chr.14: 54522740-54522740 on Build GRCh38
rs551251133 Chr.14: 54522550-54522550 on Build GRCh38
rs750967478 Chr.14: 54522792-54522792 on Build GRCh38
rs893410258 Chr.14: 54522656-54522656 on Build GRCh38
rs766636003 Chr.14: 54522454-54522468 on Build GRCh38
rs955411082 Chr.14: 54522691-54522691 on Build GRCh38
rs759203392 Chr.14: 54522624-54522624 on Build GRCh38
rs1024881997 Chr.14: 54522428-54522428 on Build GRCh38
rs762450985 Chr.14: 54522476-54522476 on Build GRCh38
rs765844063 Chr.14: 54522536-54522536 on Build GRCh38
rs368690111 Chr.14: 54522597-54522597 on Build GRCh38
rs976417505 Chr.14: 54522844-54522844 on Build GRCh38
rs765755970 Chr.14: 54522643-54522643 on Build GRCh38
rs750473196 Chr.14: 54522539-54522539 on Build GRCh38
rs910502702 Chr.14: 54522562-54522562 on Build GRCh38
rs949072473 Chr.14: 54522823-54522823 on Build GRCh38
rs754178720 Chr.14: 54522513-54522513 on Build GRCh38
rs773143538 Chr.14: 54522587-54522587 on Build GRCh38
rs757801242 Chr.14: 54522529-54522529 on Build GRCh38
rs1028956061 Chr.14: 54522667-54522667 on Build GRCh38
rs540614841 Chr.14: 54522399-54522399 on Build GRCh38
rs58196816 Chr.14: 54522634-54522634 on Build GRCh38

View Full Product Details