Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30626523-30627038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP11B
Entrez Gene ID
89839
Gene Name
Rho GTPase activating protein 11B
Gene Aliases
B'-T, FAM7B1
UniGene
Hs.659621
Gene Chromosome Location
Chr.15: 30626676-30638810 on Build GRCh38
Transcripts
NM_001039841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
470485 c.33G>T p.L11L Chr.15: 30626853-30626853 on Build GRCh38
6861221 c.20T>G p.V7G Chr.15: 30626840-30626840 on Build GRCh38
4053919 c.5G>A p.W2* Chr.15: 30626825-30626825 on Build GRCh38
700211 c.85C>T p.Q29* Chr.15: 30626905-30626905 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762702575 Chr.15: 30626959-30626959 on Build GRCh38
rs545687593 Chr.15: 30626705-30626705 on Build GRCh38
rs374688463 Chr.15: 30626794-30626794 on Build GRCh38
rs980562569 Chr.15: 30626762-30626762 on Build GRCh38
rs201169941 Chr.15: 30626807-30626807 on Build GRCh38
rs114851315 Chr.15: 30626570-30626570 on Build GRCh38
rs904606812 Chr.15: 30626551-30626551 on Build GRCh38
rs995277515 Chr.15: 30626709-30626709 on Build GRCh38
rs760282309 Chr.15: 30626777-30626777 on Build GRCh38
rs776018280 Chr.15: 30626859-30626859 on Build GRCh38
rs766049688 Chr.15: 30626994-30626994 on Build GRCh38
rs774834205 Chr.15: 30626776-30626776 on Build GRCh38
rs960083325 Chr.15: 30626680-30626680 on Build GRCh38
rs1026638176 Chr.15: 30626721-30626721 on Build GRCh38
rs780077658 Chr.15: 30626816-30626816 on Build GRCh38
rs778020898 Chr.15: 30626900-30626900 on Build GRCh38
rs542660204 Chr.15: 30626589-30626589 on Build GRCh38
rs761540412 Chr.15: 30626860-30626860 on Build GRCh38
rs765065739 Chr.15: 30626806-30626806 on Build GRCh38
rs111471393 Chr.15: 30626945-30626945 on Build GRCh38
rs375258221 Chr.15: 30626827-30626827 on Build GRCh38
rs377012511 Chr.15: 30626857-30626857 on Build GRCh38
rs767395684 Chr.15: 30626975-30626975 on Build GRCh38
rs145555407 Chr.15: 30626947-30626947 on Build GRCh38
rs367843325 Chr.15: 30626968-30626968 on Build GRCh38
rs891377286 Chr.15: 30626609-30626609 on Build GRCh38
rs747696819 Chr.15: 30626929-30626929 on Build GRCh38
rs746328211 Chr.15: 30626921-30626921 on Build GRCh38
rs781140366 Chr.15: 30626834-30626834 on Build GRCh38
rs763438128 Chr.15: 30626771-30626771 on Build GRCh38
rs776197616 Chr.15: 30626925-30626925 on Build GRCh38
rs373698991 Chr.15: 30626881-30626881 on Build GRCh38
rs766143966 Chr.15: 30626967-30626967 on Build GRCh38
rs749536238 Chr.15: 30626904-30626904 on Build GRCh38
rs745660543 Chr.15: 30626980-30626983 on Build GRCh38
rs778936202 Chr.15: 30626918-30626918 on Build GRCh38
rs186081926 Chr.15: 30626817-30626817 on Build GRCh38
rs772511633 Chr.15: 30626924-30626924 on Build GRCh38
rs371649115 Chr.15: 30626876-30626876 on Build GRCh38
rs781297338 Chr.15: 30626800-30626801 on Build GRCh38
rs769546429 Chr.15: 30626931-30626931 on Build GRCh38
rs970627550 Chr.15: 30626781-30626781 on Build GRCh38
rs367872172 Chr.15: 30626875-30626875 on Build GRCh38
rs759990878 Chr.15: 30626849-30626849 on Build GRCh38
rs575988559 Chr.15: 30626683-30626683 on Build GRCh38
rs752841959 Chr.15: 30626896-30626896 on Build GRCh38
rs781322187 Chr.15: 30626890-30626890 on Build GRCh38
rs189359089 Chr.15: 30626848-30626848 on Build GRCh38
rs896904544 Chr.15: 30626685-30626685 on Build GRCh38
rs200888956 Chr.15: 30626840-30626840 on Build GRCh38
rs749695760 Chr.15: 30626845-30626845 on Build GRCh38
rs565414781 Chr.15: 30626749-30626749 on Build GRCh38
rs889363319 Chr.15: 30626853-30626853 on Build GRCh38
rs771494259 Chr.15: 30626774-30626774 on Build GRCh38
rs756139467 Chr.15: 30626995-30626995 on Build GRCh38
rs113388235 Chr.15: 30626912-30626912 on Build GRCh38
rs556130093 Chr.15: 30626594-30626594 on Build GRCh38
rs751449533 Chr.15: 30626887-30626887 on Build GRCh38
rs541252228 Chr.15: 30626843-30626843 on Build GRCh38
rs770061069 Chr.15: 30626842-30626842 on Build GRCh38
rs1024848720 Chr.15: 30626731-30626731 on Build GRCh38
rs758402948 Chr.15: 30626809-30626809 on Build GRCh38
rs1025273012 Chr.15: 30626755-30626755 on Build GRCh38
rs752754000 Chr.15: 30626986-30626986 on Build GRCh38
rs770978363 Chr.15: 30626908-30626909 on Build GRCh38
rs753487483 Chr.15: 30626800-30626800 on Build GRCh38
rs554274621 Chr.15: 30626560-30626560 on Build GRCh38
rs1008364241 Chr.15: 30626614-30626614 on Build GRCh38
rs766121048 Chr.15: 30626886-30626886 on Build GRCh38
rs894774603 Chr.15: 30626744-30626744 on Build GRCh38
rs755058888 Chr.15: 30626784-30626785 on Build GRCh38
rs768145878 Chr.15: 30626858-30626858 on Build GRCh38
rs933004817 Chr.15: 30626839-30626839 on Build GRCh38
rs998974949 Chr.15: 30626569-30626569 on Build GRCh38
rs144751048 Chr.15: 30626888-30626888 on Build GRCh38
rs370060705 Chr.15: 30626695-30626695 on Build GRCh38
rs761560945 Chr.15: 30626802-30626802 on Build GRCh38
rs749247947 Chr.15: 30626842-30626846 on Build GRCh38
rs757576065 Chr.15: 30626910-30626910 on Build GRCh38

View Full Product Details