Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 9119332-9119831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFV2
Entrez Gene ID
4729
Gene Name
NADH:ubiquinone oxidoreductase core subunit V2
Gene Aliases
CI-24k
UniGene
Hs.464572
Gene Chromosome Location
Chr.18: 9102623-9134345 on Build GRCh38
Transcripts
NM_021074.4 XM_017025782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4131120 c.201A>T p.V67V Chr.18: 9119491-9119491 on Build GRCh38
4913341 c.216A>G p.E72E Chr.18: 9119506-9119506 on Build GRCh38
1216932 c.175A>C p.N59H Chr.18: 9119380-9119380 on Build GRCh38
7412997 c.147T>G p.P49P Chr.18: 9119352-9119352 on Build GRCh38
3147953 c.184A>C p.R62R Chr.18: 9119474-9119474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899835997 Chr.18: 9119736-9119736 on Build GRCh38
rs373173197 Chr.18: 9119441-9119441 on Build GRCh38
rs751364764 Chr.18: 9119572-9119572 on Build GRCh38
rs374325723 Chr.18: 9119655-9119655 on Build GRCh38
rs756964058 Chr.18: 9119415-9119416 on Build GRCh38
rs936561432 Chr.18: 9119383-9119383 on Build GRCh38
rs777647699 Chr.18: 9119431-9119431 on Build GRCh38
rs745935737 Chr.18: 9119456-9119456 on Build GRCh38
rs41274298 Chr.18: 9119407-9119407 on Build GRCh38
rs759046300 Chr.18: 9119636-9119636 on Build GRCh38
rs558432062 Chr.18: 9119685-9119685 on Build GRCh38
rs1027685770 Chr.18: 9119800-9119800 on Build GRCh38
rs765008316 Chr.18: 9119431-9119432 on Build GRCh38
rs925339810 Chr.18: 9119753-9119753 on Build GRCh38
rs758211188 Chr.18: 9119428-9119428 on Build GRCh38
rs776058736 Chr.18: 9119480-9119480 on Build GRCh38
rs980834459 Chr.18: 9119752-9119752 on Build GRCh38
rs182354030 Chr.18: 9119596-9119596 on Build GRCh38
rs1004420665 Chr.18: 9119807-9119807 on Build GRCh38
rs753671571 Chr.18: 9119362-9119362 on Build GRCh38
rs926298501 Chr.18: 9119368-9119368 on Build GRCh38
rs760351418 Chr.18: 9119497-9119497 on Build GRCh38
rs770629582 Chr.18: 9119623-9119623 on Build GRCh38
rs774649439 Chr.18: 9119493-9119493 on Build GRCh38
rs371902664 Chr.18: 9119545-9119545 on Build GRCh38
rs747954229 Chr.18: 9119419-9119419 on Build GRCh38
rs367649556 Chr.18: 9119380-9119380 on Build GRCh38
rs754499401 Chr.18: 9119516-9119516 on Build GRCh38
rs376294168 Chr.18: 9119446-9119446 on Build GRCh38
rs41274300 Chr.18: 9119491-9119491 on Build GRCh38
rs765985175 Chr.18: 9119508-9119508 on Build GRCh38
rs958136280 Chr.18: 9119766-9119766 on Build GRCh38
rs749823954 Chr.18: 9119598-9119598 on Build GRCh38
rs540108131 Chr.18: 9119625-9119625 on Build GRCh38
rs898940223 Chr.18: 9119570-9119570 on Build GRCh38
rs752688140 Chr.18: 9119540-9119540 on Build GRCh38
rs764821161 Chr.18: 9119638-9119638 on Build GRCh38
rs771233944 Chr.18: 9119447-9119447 on Build GRCh38
rs758025899 Chr.18: 9119616-9119617 on Build GRCh38
rs761235301 Chr.18: 9119482-9119482 on Build GRCh38
rs186788173 Chr.18: 9119619-9119619 on Build GRCh38
rs753396575 Chr.18: 9119513-9119513 on Build GRCh38
rs981230307 Chr.18: 9119374-9119374 on Build GRCh38
rs1022887324 Chr.18: 9119701-9119701 on Build GRCh38
rs1052625937 Chr.18: 9119404-9119404 on Build GRCh38
rs781548548 Chr.18: 9119594-9119594 on Build GRCh38
rs551027823 Chr.18: 9119617-9119617 on Build GRCh38
rs573316973 Chr.18: 9119765-9119765 on Build GRCh38
rs769920941 Chr.18: 9119597-9119597 on Build GRCh38
rs368515386 Chr.18: 9119525-9119525 on Build GRCh38
rs755933742 Chr.18: 9119678-9119678 on Build GRCh38
rs765897035 Chr.18: 9119372-9119372 on Build GRCh38
rs945250352 Chr.18: 9119412-9119412 on Build GRCh38
rs995740849 Chr.18: 9119746-9119746 on Build GRCh38
rs776887143 Chr.18: 9119453-9119453 on Build GRCh38
rs757598928 Chr.18: 9119582-9119582 on Build GRCh38
rs769112162 Chr.18: 9119610-9119610 on Build GRCh38
rs770259847 Chr.18: 9119475-9119475 on Build GRCh38
rs1052026719 Chr.18: 9119489-9119489 on Build GRCh38
rs951525552 Chr.18: 9119802-9119802 on Build GRCh38
rs151299130 Chr.18: 9119359-9119359 on Build GRCh38
rs750082630 Chr.18: 9119491-9119493 on Build GRCh38
rs370594505 Chr.18: 9119392-9119392 on Build GRCh38

View Full Product Details