Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40860862-40861358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CITED4
Entrez Gene ID
163732
Gene Name
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4
Gene Aliases
-
UniGene
Hs.355820
Gene Chromosome Location
Chr.1: 40861056-40862346 on Build GRCh38
Transcripts
NM_133467.2

SNP Detail

SNP ID Chromosome Location
rs181619603 Chr.1: 40860932-40860932 on Build GRCh38
rs1016552622 Chr.1: 40860898-40860898 on Build GRCh38
rs1025484035 Chr.1: 40861281-40861281 on Build GRCh38
rs961951752 Chr.1: 40860905-40860905 on Build GRCh38
rs914678782 Chr.1: 40861328-40861328 on Build GRCh38
rs908551226 Chr.1: 40860899-40860899 on Build GRCh38
rs939049940 Chr.1: 40861270-40861270 on Build GRCh38
rs927784438 Chr.1: 40861225-40861225 on Build GRCh38
rs762720601 Chr.1: 40861062-40861062 on Build GRCh38
rs766179331 Chr.1: 40861093-40861093 on Build GRCh38
rs566959677 Chr.1: 40861026-40861026 on Build GRCh38
rs963050857 Chr.1: 40861177-40861177 on Build GRCh38
rs932905825 Chr.1: 40861336-40861336 on Build GRCh38
rs980640596 Chr.1: 40861223-40861223 on Build GRCh38
rs893129400 Chr.1: 40861066-40861066 on Build GRCh38
rs115603027 Chr.1: 40860960-40860960 on Build GRCh38
rs990215622 Chr.1: 40861317-40861317 on Build GRCh38
rs773167696 Chr.1: 40861001-40861001 on Build GRCh38
rs1054578913 Chr.1: 40861046-40861046 on Build GRCh38
rs1034933541 Chr.1: 40861176-40861176 on Build GRCh38
rs1004259871 Chr.1: 40861164-40861164 on Build GRCh38
rs1011131060 Chr.1: 40861057-40861057 on Build GRCh38
rs1044823537 Chr.1: 40860962-40860962 on Build GRCh38
rs538505146 Chr.1: 40861290-40861290 on Build GRCh38
rs752204726 Chr.1: 40861278-40861278 on Build GRCh38
rs911040934 Chr.1: 40861295-40861295 on Build GRCh38
rs12140441 Chr.1: 40860925-40860925 on Build GRCh38
rs938996324 Chr.1: 40860993-40860993 on Build GRCh38
rs899981749 Chr.1: 40861085-40861085 on Build GRCh38
rs963940634 Chr.1: 40861309-40861309 on Build GRCh38
rs946399566 Chr.1: 40861284-40861284 on Build GRCh38
rs116153784 Chr.1: 40861195-40861195 on Build GRCh38
rs542218869 Chr.1: 40860947-40860947 on Build GRCh38
rs551824550 Chr.1: 40861324-40861324 on Build GRCh38
rs12044255 Chr.1: 40861214-40861214 on Build GRCh38
rs997456100 Chr.1: 40861109-40861109 on Build GRCh38
rs907460611 Chr.1: 40861125-40861125 on Build GRCh38
rs973324102 Chr.1: 40861192-40861192 on Build GRCh38
rs891168813 Chr.1: 40861163-40861163 on Build GRCh38
rs1057641 Chr.1: 40861244-40861244 on Build GRCh38
rs918525304 Chr.1: 40860916-40860916 on Build GRCh38
rs549290196 Chr.1: 40861199-40861199 on Build GRCh38
rs1015519648 Chr.1: 40861174-40861174 on Build GRCh38
rs765038255 Chr.1: 40860942-40860942 on Build GRCh38
rs987904613 Chr.1: 40861273-40861273 on Build GRCh38
rs1029698352 Chr.1: 40861142-40861142 on Build GRCh38
rs962333985 Chr.1: 40861272-40861272 on Build GRCh38
rs962384565 Chr.1: 40860904-40860904 on Build GRCh38
rs1043972152 Chr.1: 40861288-40861288 on Build GRCh38
rs559254344 Chr.1: 40861124-40861124 on Build GRCh38
rs892136821 Chr.1: 40861021-40861021 on Build GRCh38
rs946055713 Chr.1: 40861094-40861094 on Build GRCh38
rs927733639 Chr.1: 40860978-40860978 on Build GRCh38
rs1016666053 Chr.1: 40861227-40861227 on Build GRCh38
rs751180475 Chr.1: 40861110-40861110 on Build GRCh38

View Full Product Details