Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1000424-1000944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN3B
Entrez Gene ID
116444
Gene Name
glutamate ionotropic receptor NMDA type subunit 3B
Gene Aliases
GluN3B, NR3B
UniGene
Hs.660378
Gene Chromosome Location
Chr.19: 1000419-1009814 on Build GRCh38
Transcripts
NM_138690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4131123 c.349C>T p.H117Y Chr.19: 1000786-1000786 on Build GRCh38
5449166 c.93G>A p.A31A Chr.19: 1000530-1000530 on Build GRCh38
4968783 c.47_48insCGCTGG p.G16_P17insAG Chr.19: 1000484-1000485 on Build GRCh38
6405759 c.13C>T p.R5W Chr.19: 1000450-1000450 on Build GRCh38
5636405 c.8T>C p.F3S Chr.19: 1000445-1000445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566592705 Chr.19: 1000854-1000854 on Build GRCh38
rs1037006531 Chr.19: 1000567-1000567 on Build GRCh38
rs577371675 Chr.19: 1000724-1000724 on Build GRCh38
rs1019062902 Chr.19: 1000593-1000593 on Build GRCh38
rs985279510 Chr.19: 1000446-1000446 on Build GRCh38
rs139242998 Chr.19: 1000718-1000718 on Build GRCh38
rs956634776 Chr.19: 1000636-1000636 on Build GRCh38
rs546823313 Chr.19: 1000677-1000677 on Build GRCh38
rs764232329 Chr.19: 1000461-1000461 on Build GRCh38
rs542744852 Chr.19: 1000531-1000531 on Build GRCh38
rs558871680 Chr.19: 1000469-1000469 on Build GRCh38
rs1030800158 Chr.19: 1000831-1000831 on Build GRCh38
rs190077992 Chr.19: 1000480-1000480 on Build GRCh38
rs935727883 Chr.19: 1000600-1000614 on Build GRCh38
rs1007587890 Chr.19: 1000758-1000758 on Build GRCh38
rs550863693 Chr.19: 1000743-1000743 on Build GRCh38
rs770128506 Chr.19: 1000731-1000731 on Build GRCh38
rs975512370 Chr.19: 1000893-1000893 on Build GRCh38
rs532635834 Chr.19: 1000725-1000725 on Build GRCh38
rs919425804 Chr.19: 1000916-1000916 on Build GRCh38
rs748958926 Chr.19: 1000612-1000612 on Build GRCh38
rs372053516 Chr.19: 1000467-1000467 on Build GRCh38
rs112236900 Chr.19: 1000569-1000569 on Build GRCh38
rs972189690 Chr.19: 1000909-1000909 on Build GRCh38
rs1036547423 Chr.19: 1000702-1000702 on Build GRCh38
rs754448875 Chr.19: 1000632-1000632 on Build GRCh38
rs1008054623 Chr.19: 1000591-1000591 on Build GRCh38
rs960497376 Chr.19: 1000900-1000901 on Build GRCh38
rs764099116 Chr.19: 1000787-1000787 on Build GRCh38
rs939495536 Chr.19: 1000696-1000696 on Build GRCh38
rs756032796 Chr.19: 1000824-1000824 on Build GRCh38
rs768904578 Chr.19: 1000467-1000468 on Build GRCh38
rs1018530107 Chr.19: 1000464-1000464 on Build GRCh38
rs989122924 Chr.19: 1000497-1000497 on Build GRCh38
rs889167389 Chr.19: 1000581-1000586 on Build GRCh38
rs749623371 Chr.19: 1000449-1000450 on Build GRCh38
rs980919866 Chr.19: 1000672-1000672 on Build GRCh38
rs947814832 Chr.19: 1000528-1000528 on Build GRCh38
rs544534250 Chr.19: 1000478-1000478 on Build GRCh38
rs980484474 Chr.19: 1000878-1000878 on Build GRCh38
rs964177320 Chr.19: 1000886-1000886 on Build GRCh38
rs994812003 Chr.19: 1000573-1000573 on Build GRCh38
rs886877480 Chr.19: 1000838-1000838 on Build GRCh38
rs2301813 Chr.19: 1000800-1000800 on Build GRCh38
rs1016565759 Chr.19: 1000853-1000853 on Build GRCh38
rs969445021 Chr.19: 1000867-1000867 on Build GRCh38
rs914963662 Chr.19: 1000517-1000517 on Build GRCh38
rs1049101165 Chr.19: 1000580-1000581 on Build GRCh38
rs577245079 Chr.19: 1000477-1000477 on Build GRCh38
rs1022201406 Chr.19: 1000864-1000864 on Build GRCh38
rs12986002 Chr.19: 1000786-1000786 on Build GRCh38
rs767603391 Chr.19: 1000812-1000812 on Build GRCh38
rs554367291 Chr.19: 1000815-1000815 on Build GRCh38
rs766207454 Chr.19: 1000847-1000847 on Build GRCh38
rs925903057 Chr.19: 1000492-1000492 on Build GRCh38
rs747492431 Chr.19: 1000667-1000667 on Build GRCh38
rs530938555 Chr.19: 1000491-1000491 on Build GRCh38
rs761687154 Chr.19: 1000807-1000807 on Build GRCh38
rs1035156351 Chr.19: 1000880-1000880 on Build GRCh38
rs989071947 Chr.19: 1000661-1000661 on Build GRCh38
rs969117858 Chr.19: 1000671-1000672 on Build GRCh38
rs774860708 Chr.19: 1000840-1000841 on Build GRCh38
rs144440136 Chr.19: 1000666-1000666 on Build GRCh38
rs757164134 Chr.19: 1000472-1000472 on Build GRCh38
rs998053758 Chr.19: 1000601-1000601 on Build GRCh38
rs925376109 Chr.19: 1000911-1000911 on Build GRCh38
rs928117882 Chr.19: 1000558-1000558 on Build GRCh38

View Full Product Details