Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233743052-233743544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB3
Entrez Gene ID
414061
Gene Name
DnaJ heat shock protein family (Hsp40) member B3
Gene Aliases
HCG3
UniGene
Hs.725533
Gene Chromosome Location
Chr.2: 233742750-233744015 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A10
Entrez Gene ID
54575
Gene Name
UDP glucuronosyltransferase family 1 member A10
Gene Aliases
UDPGT, UGT-1J, UGT1-10, UGT1.10, UGT1J
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233636477-233773305 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A3
Entrez Gene ID
54659
Gene Name
UDP glucuronosyltransferase family 1 member A3
Gene Aliases
UDPGT, UDPGT 1-3, UGT-1C, UGT1-03, UGT1.3, UGT1A3S, UGT1C
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233729127-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A4
Entrez Gene ID
54657
Gene Name
UDP glucuronosyltransferase family 1 member A4
Gene Aliases
HUG-BR2, UDPGT, UDPGT 1-4, UGT-1D, UGT1-04, UGT1.4, UGT1A4S, UGT1D
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233718792-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A5
Entrez Gene ID
54579
Gene Name
UDP glucuronosyltransferase family 1 member A5
Gene Aliases
UDPGT, UDPGT 1-5, UGT1E
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233712992-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A6
Entrez Gene ID
54578
Gene Name
UDP glucuronosyltransferase family 1 member A6
Gene Aliases
GNT1, HLUGP, HLUGP1, UDPGT, UDPGT 1-6, UGT1, UGT1A6S, UGT1F
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233691675-233773305 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A7
Entrez Gene ID
54577
Gene Name
UDP glucuronosyltransferase family 1 member A7
Gene Aliases
UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233681938-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A8
Entrez Gene ID
54576
Gene Name
UDP glucuronosyltransferase family 1 member A8
Gene Aliases
UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233617645-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A9
Entrez Gene ID
54600
Gene Name
UDP glucuronosyltransferase family 1 member A9
Gene Aliases
HLUGP4, LUGP4, UDPGT, UDPGT 1-9, UGT-1I, UGT1-09, UGT1-9, UGT1.9, UGT1A9S, UGT1AI, UGT1I
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233671898-233773305 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

SNP Detail

SNP ID Chromosome Location
rs943372310 Chr.2: 233743386-233743386 on Build GRCh38
rs764022501 Chr.2: 233743262-233743262 on Build GRCh38
rs35742148 Chr.2: 233743416-233743417 on Build GRCh38
rs926867030 Chr.2: 233743086-233743086 on Build GRCh38
rs1039034416 Chr.2: 233743402-233743402 on Build GRCh38
rs779473501 Chr.2: 233743432-233743432 on Build GRCh38
rs891586644 Chr.2: 233743447-233743449 on Build GRCh38
rs904424565 Chr.2: 233743406-233743406 on Build GRCh38
rs191127881 Chr.2: 233743160-233743160 on Build GRCh38
rs1133491 Chr.2: 233743219-233743219 on Build GRCh38
rs890417229 Chr.2: 233743173-233743173 on Build GRCh38
rs61736688 Chr.2: 233743458-233743458 on Build GRCh38
rs77486749 Chr.2: 233743142-233743142 on Build GRCh38
rs751434159 Chr.2: 233743230-233743230 on Build GRCh38
rs28900375 Chr.2: 233743176-233743176 on Build GRCh38
rs17863796 Chr.2: 233743234-233743234 on Build GRCh38
rs3796088 Chr.2: 233743154-233743154 on Build GRCh38
rs1054800 Chr.2: 233743076-233743076 on Build GRCh38
rs185061073 Chr.2: 233743506-233743506 on Build GRCh38
rs912011839 Chr.2: 233743365-233743365 on Build GRCh38
rs577178509 Chr.2: 233743419-233743419 on Build GRCh38
rs904489240 Chr.2: 233743187-233743187 on Build GRCh38
rs538069492 Chr.2: 233743270-233743270 on Build GRCh38
rs771301493 Chr.2: 233743449-233743449 on Build GRCh38
rs561592324 Chr.2: 233743242-233743242 on Build GRCh38
rs553930129 Chr.2: 233743379-233743379 on Build GRCh38
rs539413664 Chr.2: 233743435-233743435 on Build GRCh38
rs542057655 Chr.2: 233743464-233743464 on Build GRCh38
rs980978105 Chr.2: 233743077-233743077 on Build GRCh38
rs1007522994 Chr.2: 233743174-233743174 on Build GRCh38
rs777952241 Chr.2: 233743521-233743521 on Build GRCh38
rs753191882 Chr.2: 233743514-233743514 on Build GRCh38
rs1013981609 Chr.2: 233743205-233743205 on Build GRCh38
rs397839834 Chr.2: 233743224-233743224 on Build GRCh38
rs758496826 Chr.2: 233743438-233743438 on Build GRCh38
rs759007678 Chr.2: 233743520-233743520 on Build GRCh38
rs747196362 Chr.2: 233743442-233743442 on Build GRCh38
rs372766492 Chr.2: 233743462-233743462 on Build GRCh38
rs568503727 Chr.2: 233743334-233743334 on Build GRCh38
rs755334550 Chr.2: 233743431-233743431 on Build GRCh38
rs987483436 Chr.2: 233743352-233743352 on Build GRCh38
rs199947040 Chr.2: 233743509-233743509 on Build GRCh38
rs1016583348 Chr.2: 233743495-233743495 on Build GRCh38
rs569147758 Chr.2: 233743247-233743247 on Build GRCh38
rs192671736 Chr.2: 233743440-233743440 on Build GRCh38
rs761358710 Chr.2: 233743505-233743505 on Build GRCh38
rs974078775 Chr.2: 233743284-233743284 on Build GRCh38
rs879890998 Chr.2: 233743223-233743223 on Build GRCh38
rs78331994 Chr.2: 233743143-233743143 on Build GRCh38
rs546323978 Chr.2: 233743185-233743185 on Build GRCh38
rs560095677 Chr.2: 233743192-233743192 on Build GRCh38
rs187768174 Chr.2: 233743103-233743103 on Build GRCh38
rs919411761 Chr.2: 233743303-233743303 on Build GRCh38
rs773869804 Chr.2: 233743484-233743484 on Build GRCh38
rs750114193 Chr.2: 233743508-233743508 on Build GRCh38
rs762879751 Chr.2: 233743474-233743474 on Build GRCh38
rs751668012 Chr.2: 233743524-233743524 on Build GRCh38
rs879874451 Chr.2: 233743209-233743209 on Build GRCh38
rs919324462 Chr.2: 233743135-233743135 on Build GRCh38
rs548391374 Chr.2: 233743280-233743280 on Build GRCh38
rs534297671 Chr.2: 233743335-233743335 on Build GRCh38
rs182109098 Chr.2: 233743171-233743171 on Build GRCh38
rs942184123 Chr.2: 233743088-233743088 on Build GRCh38
rs773487683 Chr.2: 233743474-233743475 on Build GRCh38
rs187896113 Chr.2: 233743204-233743204 on Build GRCh38
rs781319015 Chr.2: 233743455-233743455 on Build GRCh38
rs768112236 Chr.2: 233743482-233743482 on Build GRCh38

View Full Product Details