Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45878202-45878713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSWIM3
Entrez Gene ID
140831
Gene Name
zinc finger SWIM-type containing 3
Gene Aliases
C20orf164, PPP1R174
UniGene
Hs.292135
Gene Chromosome Location
Chr.20: 45857581-45879131 on Build GRCh38
Transcripts
NM_080752.3 NR_037628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1681718 c.1919G>C p.R640P Chr.20: 45878495-45878495 on Build GRCh38
3707800 c.1675T>G p.L559V Chr.20: 45878251-45878251 on Build GRCh38
7279535 c.1719G>A p.M573I Chr.20: 45878295-45878295 on Build GRCh38
5797232 c.2066A>G p.H689R Chr.20: 45878642-45878642 on Build GRCh38
1736799 c.1825C>T p.R609W Chr.20: 45878401-45878401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775442140 Chr.20: 45878532-45878532 on Build GRCh38
rs781502902 Chr.20: 45878304-45878304 on Build GRCh38
rs141779185 Chr.20: 45878575-45878575 on Build GRCh38
rs747605406 Chr.20: 45878511-45878511 on Build GRCh38
rs757046850 Chr.20: 45878553-45878555 on Build GRCh38
rs754235021 Chr.20: 45878490-45878490 on Build GRCh38
rs757139461 Chr.20: 45878614-45878614 on Build GRCh38
rs777793054 Chr.20: 45878242-45878242 on Build GRCh38
rs369388471 Chr.20: 45878617-45878617 on Build GRCh38
rs768713497 Chr.20: 45878656-45878656 on Build GRCh38
rs1038020173 Chr.20: 45878509-45878509 on Build GRCh38
rs200330798 Chr.20: 45878342-45878342 on Build GRCh38
rs774688845 Chr.20: 45878526-45878526 on Build GRCh38
rs372827829 Chr.20: 45878635-45878635 on Build GRCh38
rs757702054 Chr.20: 45878266-45878266 on Build GRCh38
rs766108911 Chr.20: 45878494-45878494 on Build GRCh38
rs754132693 Chr.20: 45878389-45878389 on Build GRCh38
rs781451177 Chr.20: 45878625-45878625 on Build GRCh38
rs911326872 Chr.20: 45878545-45878545 on Build GRCh38
rs754428062 Chr.20: 45878406-45878406 on Build GRCh38
rs375272202 Chr.20: 45878559-45878559 on Build GRCh38
rs748031815 Chr.20: 45878410-45878410 on Build GRCh38
rs766563720 Chr.20: 45878301-45878301 on Build GRCh38
rs777811141 Chr.20: 45878516-45878516 on Build GRCh38
rs777906961 Chr.20: 45878601-45878601 on Build GRCh38
rs17446511 Chr.20: 45878668-45878668 on Build GRCh38
rs749029309 Chr.20: 45878662-45878662 on Build GRCh38
rs748403812 Chr.20: 45878247-45878247 on Build GRCh38
rs751680946 Chr.20: 45878378-45878378 on Build GRCh38
rs751527870 Chr.20: 45878302-45878302 on Build GRCh38
rs749042009 Chr.20: 45878454-45878454 on Build GRCh38
rs768109084 Chr.20: 45878383-45878383 on Build GRCh38
rs755652199 Chr.20: 45878597-45878597 on Build GRCh38
rs763274124 Chr.20: 45878366-45878366 on Build GRCh38
rs1054538733 Chr.20: 45878564-45878564 on Build GRCh38
rs769789787 Chr.20: 45878420-45878420 on Build GRCh38
rs766649703 Chr.20: 45878370-45878370 on Build GRCh38
rs745420059 Chr.20: 45878257-45878257 on Build GRCh38
rs755365506 Chr.20: 45878231-45878231 on Build GRCh38
rs751361964 Chr.20: 45878428-45878428 on Build GRCh38
rs560824324 Chr.20: 45878382-45878382 on Build GRCh38
rs548540451 Chr.20: 45878541-45878541 on Build GRCh38
rs745501714 Chr.20: 45878334-45878334 on Build GRCh38
rs774404420 Chr.20: 45878457-45878457 on Build GRCh38
rs745616813 Chr.20: 45878629-45878629 on Build GRCh38
rs762312038 Chr.20: 45878681-45878681 on Build GRCh38
rs540195429 Chr.20: 45878398-45878398 on Build GRCh38
rs544406910 Chr.20: 45878303-45878303 on Build GRCh38
rs754335703 Chr.20: 45878618-45878618 on Build GRCh38
rs752286337 Chr.20: 45878581-45878581 on Build GRCh38
rs756524032 Chr.20: 45878385-45878385 on Build GRCh38
rs776406063 Chr.20: 45878360-45878360 on Build GRCh38
rs780067523 Chr.20: 45878401-45878401 on Build GRCh38
rs148099263 Chr.20: 45878556-45878556 on Build GRCh38
rs150906252 Chr.20: 45878402-45878402 on Build GRCh38
rs896620485 Chr.20: 45878243-45878243 on Build GRCh38
rs1009935259 Chr.20: 45878448-45878448 on Build GRCh38
rs961557879 Chr.20: 45878508-45878508 on Build GRCh38
rs751159706 Chr.20: 45878495-45878495 on Build GRCh38
rs773156129 Chr.20: 45878449-45878449 on Build GRCh38
rs753812522 Chr.20: 45878608-45878608 on Build GRCh38
rs374425711 Chr.20: 45878249-45878249 on Build GRCh38
rs145329268 Chr.20: 45878363-45878363 on Build GRCh38
rs6032571 Chr.20: 45878422-45878422 on Build GRCh38
rs779285020 Chr.20: 45878306-45878306 on Build GRCh38
rs761477278 Chr.20: 45878279-45878279 on Build GRCh38
rs145639872 Chr.20: 45878531-45878531 on Build GRCh38
rs116532261 Chr.20: 45878505-45878505 on Build GRCh38
rs771307649 Chr.20: 45878456-45878456 on Build GRCh38
rs761172793 Chr.20: 45878479-45878479 on Build GRCh38
rs770609478 Chr.20: 45878520-45878520 on Build GRCh38
rs149462189 Chr.20: 45878688-45878688 on Build GRCh38
rs141913268 Chr.20: 45878491-45878491 on Build GRCh38
rs750733599 Chr.20: 45878569-45878569 on Build GRCh38
rs142105449 Chr.20: 45878384-45878384 on Build GRCh38
rs529895998 Chr.20: 45878517-45878517 on Build GRCh38
rs759710196 Chr.20: 45878458-45878458 on Build GRCh38
rs775459561 Chr.20: 45878265-45878265 on Build GRCh38
rs756438068 Chr.20: 45878317-45878317 on Build GRCh38
rs750376917 Chr.20: 45878608-45878609 on Build GRCh38
rs201805977 Chr.20: 45878622-45878622 on Build GRCh38
rs200927989 Chr.20: 45878326-45878326 on Build GRCh38
rs747145684 Chr.20: 45878655-45878655 on Build GRCh38
rs140603620 Chr.20: 45878271-45878271 on Build GRCh38
rs139396693 Chr.20: 45878473-45878473 on Build GRCh38
rs756141994 Chr.20: 45878512-45878512 on Build GRCh38
rs747505197 Chr.20: 45878228-45878228 on Build GRCh38
rs936114296 Chr.20: 45878561-45878561 on Build GRCh38
rs772203694 Chr.20: 45878640-45878640 on Build GRCh38
rs542143044 Chr.20: 45878359-45878359 on Build GRCh38
rs537547455 Chr.20: 45878568-45878568 on Build GRCh38
rs780221869 Chr.20: 45878654-45878654 on Build GRCh38
rs201321759 Chr.20: 45878576-45878576 on Build GRCh38
rs777104903 Chr.20: 45878548-45878548 on Build GRCh38
rs368149535 Chr.20: 45878469-45878469 on Build GRCh38
rs967978052 Chr.20: 45878624-45878624 on Build GRCh38
rs781249493 Chr.20: 45878407-45878407 on Build GRCh38
rs764523680 Chr.20: 45878487-45878487 on Build GRCh38
rs149198668 Chr.20: 45878365-45878365 on Build GRCh38
rs200818698 Chr.20: 45878336-45878336 on Build GRCh38
rs373046431 Chr.20: 45878280-45878280 on Build GRCh38
rs553363961 Chr.20: 45878255-45878255 on Build GRCh38
rs771545352 Chr.20: 45878262-45878262 on Build GRCh38
rs763185273 Chr.20: 45878293-45878293 on Build GRCh38
rs778495385 Chr.20: 45878254-45878254 on Build GRCh38
rs150493098 Chr.20: 45878349-45878349 on Build GRCh38
rs780574081 Chr.20: 45878507-45878507 on Build GRCh38
rs764627937 Chr.20: 45878542-45878542 on Build GRCh38
rs767424682 Chr.20: 45878600-45878602 on Build GRCh38
rs746457594 Chr.20: 45878350-45878350 on Build GRCh38

View Full Product Details