Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84644958-84645504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSX2IP
Entrez Gene ID
117178
Gene Name
SSX family member 2 interacting protein
Gene Aliases
ADIP
UniGene
Hs.22587
Gene Chromosome Location
Chr.1: 84643707-84690757 on Build GRCh38
Transcripts
NM_001166293.1 NM_001166294.1 NM_001166295.1 NM_001166417.1 NM_014021.3 XM_005270427.1 XM_005270428.1 XM_005270429.1 XM_006710331.1 XM_011540610.1 XM_017000234.1 XM_017000235.1 XM_017000236.1 XM_017000237.1

SNP Detail

SNP ID Chromosome Location
rs951184214 Chr.1: 84645478-84645478 on Build GRCh38
rs369423311 Chr.1: 84645085-84645088 on Build GRCh38
rs920795948 Chr.1: 84645179-84645179 on Build GRCh38
rs970743740 Chr.1: 84645068-84645068 on Build GRCh38
rs989620585 Chr.1: 84645208-84645208 on Build GRCh38
rs929499827 Chr.1: 84645194-84645194 on Build GRCh38
rs1029592699 Chr.1: 84645063-84645063 on Build GRCh38
rs745731936 Chr.1: 84645442-84645442 on Build GRCh38
rs974113286 Chr.1: 84645462-84645462 on Build GRCh38
rs772201018 Chr.1: 84645471-84645471 on Build GRCh38
rs796816536 Chr.1: 84645235-84645235 on Build GRCh38
rs962398268 Chr.1: 84645461-84645461 on Build GRCh38
rs902562660 Chr.1: 84645422-84645422 on Build GRCh38
rs1005280599 Chr.1: 84645435-84645435 on Build GRCh38
rs189658260 Chr.1: 84645323-84645323 on Build GRCh38
rs888208502 Chr.1: 84645397-84645397 on Build GRCh38
rs552531937 Chr.1: 84645025-84645025 on Build GRCh38
rs529346829 Chr.1: 84645307-84645307 on Build GRCh38
rs958081261 Chr.1: 84645138-84645138 on Build GRCh38
rs561170318 Chr.1: 84645295-84645295 on Build GRCh38
rs567456088 Chr.1: 84645150-84645150 on Build GRCh38
rs572552045 Chr.1: 84645030-84645030 on Build GRCh38
rs115064597 Chr.1: 84645064-84645064 on Build GRCh38
rs895130224 Chr.1: 84645374-84645374 on Build GRCh38
rs2911613 Chr.1: 84645441-84645441 on Build GRCh38
rs879253133 Chr.1: 84645299-84645299 on Build GRCh38
rs551800248 Chr.1: 84645467-84645467 on Build GRCh38
rs370591298 Chr.1: 84645056-84645056 on Build GRCh38
rs578236241 Chr.1: 84645323-84645324 on Build GRCh38
rs1049386996 Chr.1: 84645392-84645392 on Build GRCh38
rs945643338 Chr.1: 84645315-84645315 on Build GRCh38
rs922296628 Chr.1: 84645355-84645355 on Build GRCh38
rs999368605 Chr.1: 84645424-84645425 on Build GRCh38
rs377225081 Chr.1: 84645337-84645337 on Build GRCh38
rs1041287118 Chr.1: 84645325-84645325 on Build GRCh38
rs1043807959 Chr.1: 84645422-84645426 on Build GRCh38
rs575515949 Chr.1: 84645106-84645106 on Build GRCh38
rs181223156 Chr.1: 84645376-84645376 on Build GRCh38
rs372944509 Chr.1: 84645102-84645102 on Build GRCh38
rs377287868 Chr.1: 84645246-84645246 on Build GRCh38
rs910860414 Chr.1: 84645107-84645107 on Build GRCh38
rs973847517 Chr.1: 84645140-84645140 on Build GRCh38
rs1055075444 Chr.1: 84645345-84645345 on Build GRCh38
rs544042724 Chr.1: 84645241-84645241 on Build GRCh38
rs955286518 Chr.1: 84645438-84645438 on Build GRCh38
rs1014619635 Chr.1: 84645453-84645453 on Build GRCh38
rs926712503 Chr.1: 84645124-84645124 on Build GRCh38
rs748412841 Chr.1: 84645258-84645258 on Build GRCh38
rs1014152032 Chr.1: 84645023-84645023 on Build GRCh38
rs961606686 Chr.1: 84645012-84645012 on Build GRCh38

View Full Product Details