Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84656269-84656783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSX2IP
Entrez Gene ID
117178
Gene Name
SSX family member 2 interacting protein
Gene Aliases
ADIP
UniGene
Hs.22587
Gene Chromosome Location
Chr.1: 84643707-84690757 on Build GRCh38
Transcripts
NM_001166293.1 NM_001166294.1 NM_001166295.1 NM_001166417.1 NM_014021.3 XM_005270427.1 XM_005270428.1 XM_005270429.1 XM_006710331.1 XM_011540610.1 XM_011540612.2 XM_017000234.1 XM_017000235.1 XM_017000236.1 XM_017000237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842455 c.1188G>A p.M396I Chr.1: 84656375-84656375 on Build GRCh38
6397926 c.1079T>G p.V360G Chr.1: 84656484-84656484 on Build GRCh38
4577605 c.1155A>G p.K385K Chr.1: 84656408-84656408 on Build GRCh38
5881801 c.1168A>G p.I390V Chr.1: 84656395-84656395 on Build GRCh38
1344552 c.1158C>T p.L386L Chr.1: 84656405-84656405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753234152 Chr.1: 84656448-84656448 on Build GRCh38
rs2994441 Chr.1: 84656300-84656300 on Build GRCh38
rs547783435 Chr.1: 84656301-84656301 on Build GRCh38
rs532073248 Chr.1: 84656634-84656634 on Build GRCh38
rs936005194 Chr.1: 84656744-84656744 on Build GRCh38
rs1014858942 Chr.1: 84656581-84656581 on Build GRCh38
rs370153105 Chr.1: 84656404-84656404 on Build GRCh38
rs916437502 Chr.1: 84656720-84656720 on Build GRCh38
rs960786218 Chr.1: 84656615-84656615 on Build GRCh38
rs765786190 Chr.1: 84656441-84656441 on Build GRCh38
rs893349622 Chr.1: 84656658-84656658 on Build GRCh38
rs780707067 Chr.1: 84656499-84656500 on Build GRCh38
rs187802459 Chr.1: 84656548-84656548 on Build GRCh38
rs578130929 Chr.1: 84656563-84656563 on Build GRCh38
rs1004736228 Chr.1: 84656566-84656566 on Build GRCh38
rs751745089 Chr.1: 84656472-84656472 on Build GRCh38
rs768364463 Chr.1: 84656480-84656481 on Build GRCh38
rs928780706 Chr.1: 84656620-84656620 on Build GRCh38
rs540155128 Chr.1: 84656498-84656498 on Build GRCh38
rs777916573 Chr.1: 84656314-84656314 on Build GRCh38
rs1030116463 Chr.1: 84656362-84656362 on Build GRCh38
rs756236110 Chr.1: 84656489-84656489 on Build GRCh38
rs770611212 Chr.1: 84656525-84656525 on Build GRCh38
rs781364723 Chr.1: 84656357-84656357 on Build GRCh38
rs771240941 Chr.1: 84656551-84656551 on Build GRCh38
rs949138122 Chr.1: 84656599-84656599 on Build GRCh38
rs980210657 Chr.1: 84656726-84656726 on Build GRCh38
rs183116917 Chr.1: 84656303-84656303 on Build GRCh38
rs768674134 Chr.1: 84656373-84656373 on Build GRCh38
rs759018697 Chr.1: 84656437-84656437 on Build GRCh38
rs771141736 Chr.1: 84656353-84656353 on Build GRCh38
rs771766564 Chr.1: 84656519-84656519 on Build GRCh38
rs764453235 Chr.1: 84656464-84656464 on Build GRCh38
rs12116963 Chr.1: 84656588-84656588 on Build GRCh38
rs771883431 Chr.1: 84656402-84656402 on Build GRCh38
rs28667025 Chr.1: 84656725-84656725 on Build GRCh38
rs925924579 Chr.1: 84656732-84656732 on Build GRCh38
rs749251388 Chr.1: 84656550-84656550 on Build GRCh38
rs760304925 Chr.1: 84656427-84656427 on Build GRCh38
rs555457607 Chr.1: 84656623-84656623 on Build GRCh38
rs947438670 Chr.1: 84656671-84656671 on Build GRCh38
rs997998899 Chr.1: 84656709-84656709 on Build GRCh38
rs375495640 Chr.1: 84656359-84656359 on Build GRCh38
rs1056074639 Chr.1: 84656652-84656652 on Build GRCh38
rs748110896 Chr.1: 84656510-84656510 on Build GRCh38
rs747996869 Chr.1: 84656514-84656515 on Build GRCh38
rs1042092042 Chr.1: 84656672-84656676 on Build GRCh38
rs536068181 Chr.1: 84656622-84656622 on Build GRCh38
rs563348781 Chr.1: 84656478-84656478 on Build GRCh38
rs761558074 Chr.1: 84656395-84656395 on Build GRCh38
rs981646352 Chr.1: 84656617-84656617 on Build GRCh38
rs774072260 Chr.1: 84656745-84656745 on Build GRCh38
rs745981359 Chr.1: 84656488-84656488 on Build GRCh38
rs765899069 Chr.1: 84656431-84656431 on Build GRCh38
rs903304598 Chr.1: 84656699-84656699 on Build GRCh38
rs556793574 Chr.1: 84656545-84656545 on Build GRCh38
rs753308166 Chr.1: 84656436-84656436 on Build GRCh38
rs773239996 Chr.1: 84656319-84656319 on Build GRCh38
rs951739450 Chr.1: 84656533-84656533 on Build GRCh38
rs373998289 Chr.1: 84656343-84656343 on Build GRCh38
rs12116932 Chr.1: 84656515-84656515 on Build GRCh38
rs1040489265 Chr.1: 84656470-84656470 on Build GRCh38
rs758756514 Chr.1: 84656454-84656454 on Build GRCh38
rs774399474 Chr.1: 84656390-84656390 on Build GRCh38
rs781452200 Chr.1: 84656479-84656479 on Build GRCh38

View Full Product Details